Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 76943324-76943863 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RCN2
Entrez Gene ID
5955
Gene Name
reticulocalbin 2
Gene Aliases
E6BP, ERC-55, ERC55, TCBP49
UniGene
Hs.79088
Gene Chromosome Location
Chr.15: 76931621-76950260 on Build GRCh38
Transcripts
NM_001271837.1 NM_002902.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
965149 c.448C>A p.L150I Chr.15: 76943758-76943758 on Build GRCh38
4972154 c.516A>C p.E172D Chr.15: 76943826-76943826 on Build GRCh38
1939982 c.496G>T p.G166C Chr.15: 76943806-76943806 on Build GRCh38
6400229 c.457A>T p.K153* Chr.15: 76943767-76943767 on Build GRCh38
471093 c.449T>C p.L150P Chr.15: 76943759-76943759 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs543979622 Chr.15: 76943641-76943641 on Build GRCh38
rs370184847 Chr.15: 76943375-76943375 on Build GRCh38
rs959279593 Chr.15: 76943570-76943570 on Build GRCh38
rs996174396 Chr.15: 76943358-76943358 on Build GRCh38
rs946435491 Chr.15: 76943588-76943588 on Build GRCh38
rs1025855166 Chr.15: 76943722-76943722 on Build GRCh38
rs74024642 Chr.15: 76943519-76943519 on Build GRCh38
rs900803683 Chr.15: 76943710-76943710 on Build GRCh38
rs965142965 Chr.15: 76943408-76943408 on Build GRCh38
rs549157670 Chr.15: 76943447-76943447 on Build GRCh38
rs900505008 Chr.15: 76943354-76943354 on Build GRCh38
rs1005363107 Chr.15: 76943783-76943783 on Build GRCh38
rs757910578 Chr.15: 76943832-76943832 on Build GRCh38
rs932247424 Chr.15: 76943754-76943754 on Build GRCh38
rs142859413 Chr.15: 76943811-76943811 on Build GRCh38
rs909815094 Chr.15: 76943455-76943455 on Build GRCh38
rs774911517 Chr.15: 76943779-76943779 on Build GRCh38
rs768143673 Chr.15: 76943800-76943800 on Build GRCh38
rs745691458 Chr.15: 76943735-76943735 on Build GRCh38
rs1004984314 Chr.15: 76943454-76943454 on Build GRCh38
rs946228590 Chr.15: 76943628-76943628 on Build GRCh38
rs775628097 Chr.15: 76943742-76943742 on Build GRCh38
rs1017923992 Chr.15: 76943380-76943380 on Build GRCh38
rs75550048 Chr.15: 76943700-76943700 on Build GRCh38
rs997842382 Chr.15: 76943715-76943715 on Build GRCh38
rs763061885 Chr.15: 76943762-76943762 on Build GRCh38
rs770866416 Chr.15: 76943708-76943708 on Build GRCh38
rs749591043 Chr.15: 76943664-76943664 on Build GRCh38
rs768688268 Chr.15: 76943775-76943775 on Build GRCh38
rs540583456 Chr.15: 76943398-76943398 on Build GRCh38
rs914924027 Chr.15: 76943579-76943579 on Build GRCh38
rs747682498 Chr.15: 76943595-76943595 on Build GRCh38
rs762353999 Chr.15: 76943780-76943780 on Build GRCh38
rs181911802 Chr.15: 76943699-76943699 on Build GRCh38
rs763101513 Chr.15: 76943532-76943532 on Build GRCh38
rs186289825 Chr.15: 76943741-76943741 on Build GRCh38
rs982428896 Chr.15: 76943535-76943535 on Build GRCh38
rs144772621 Chr.15: 76943836-76943836 on Build GRCh38
rs938773372 Chr.15: 76943558-76943558 on Build GRCh38
rs780895844 Chr.15: 76943727-76943727 on Build GRCh38
rs969319173 Chr.15: 76943512-76943512 on Build GRCh38
rs78925049 Chr.15: 76943763-76943763 on Build GRCh38
rs990452758 Chr.15: 76943572-76943572 on Build GRCh38
rs541675469 Chr.15: 76943482-76943482 on Build GRCh38
rs904784337 Chr.15: 76943652-76943652 on Build GRCh38
rs759462694 Chr.15: 76943370-76943370 on Build GRCh38
rs75116689 Chr.15: 76943560-76943560 on Build GRCh38
rs752356817 Chr.15: 76943827-76943827 on Build GRCh38
rs189618065 Chr.15: 76943431-76943431 on Build GRCh38
rs951323480 Chr.15: 76943436-76943436 on Build GRCh38
rs756276976 Chr.15: 76943559-76943559 on Build GRCh38
rs886845272 Chr.15: 76943724-76943724 on Build GRCh38
rs191064420 Chr.15: 76943812-76943812 on Build GRCh38
rs1043215433 Chr.15: 76943638-76943638 on Build GRCh38
rs765261872 Chr.15: 76943411-76943411 on Build GRCh38

View Full Product Details