Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 76625707-76626199 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ST6GALNAC1
Entrez Gene ID
55808
Gene Name
ST6 N-acetylgalactosaminide alpha-2,6-sialyltransferase 1
Gene Aliases
HSY11339, SIAT7A, ST6GalNAcI, STYI
UniGene
Hs.105352
Gene Chromosome Location
Chr.17: 76617769-76643838 on Build GRCh38
Transcripts
NM_001289107.1 NM_018414.4 NR_110309.1 XM_011524995.2 XM_011524996.2 XM_017024842.1 XM_017024843.1 XM_017024844.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
984742 c.1517C>A p.S506Y Chr.17: 76625907-76625907 on Build GRCh38
3522589 c.1490G>A p.R497Q Chr.17: 76626021-76626021 on Build GRCh38
984741 c.1534G>A p.A512T Chr.17: 76625890-76625890 on Build GRCh38
3362379 c.1461G>A p.R487R Chr.17: 76626050-76626050 on Build GRCh38
6081972 c.1419C>T p.H473H Chr.17: 76626092-76626092 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs891061206 Chr.17: 76626039-76626039 on Build GRCh38
rs371628668 Chr.17: 76625798-76625798 on Build GRCh38
rs961714611 Chr.17: 76625825-76625825 on Build GRCh38
rs202064484 Chr.17: 76625854-76625854 on Build GRCh38
rs116419615 Chr.17: 76625858-76625858 on Build GRCh38
rs776350615 Chr.17: 76626053-76626053 on Build GRCh38
rs779385918 Chr.17: 76625831-76625831 on Build GRCh38
rs758251282 Chr.17: 76625787-76625787 on Build GRCh38
rs946735747 Chr.17: 76625970-76625970 on Build GRCh38
rs1035880750 Chr.17: 76626164-76626164 on Build GRCh38
rs755801440 Chr.17: 76625943-76625943 on Build GRCh38
rs776670843 Chr.17: 76625981-76625981 on Build GRCh38
rs752292467 Chr.17: 76625931-76625931 on Build GRCh38
rs746178478 Chr.17: 76625952-76625952 on Build GRCh38
rs202185117 Chr.17: 76625989-76625989 on Build GRCh38
rs200331134 Chr.17: 76625880-76625880 on Build GRCh38
rs141702482 Chr.17: 76625872-76625872 on Build GRCh38
rs981337695 Chr.17: 76625873-76625873 on Build GRCh38
rs374878476 Chr.17: 76626056-76626056 on Build GRCh38
rs1002000581 Chr.17: 76626088-76626088 on Build GRCh38
rs202151074 Chr.17: 76626004-76626004 on Build GRCh38
rs781690876 Chr.17: 76626067-76626067 on Build GRCh38
rs1018749411 Chr.17: 76625738-76625738 on Build GRCh38
rs763771311 Chr.17: 76626032-76626032 on Build GRCh38
rs772094619 Chr.17: 76626114-76626114 on Build GRCh38
rs766365379 Chr.17: 76625790-76625790 on Build GRCh38
rs141085732 Chr.17: 76626045-76626045 on Build GRCh38
rs576130406 Chr.17: 76625747-76625747 on Build GRCh38
rs777775479 Chr.17: 76626084-76626084 on Build GRCh38
rs767128837 Chr.17: 76625928-76625928 on Build GRCh38
rs775573155 Chr.17: 76626116-76626116 on Build GRCh38
rs764782480 Chr.17: 76626162-76626162 on Build GRCh38
rs753329733 Chr.17: 76626146-76626146 on Build GRCh38
rs751531292 Chr.17: 76625922-76625922 on Build GRCh38
rs758987790 Chr.17: 76626059-76626059 on Build GRCh38
rs758792835 Chr.17: 76625954-76625954 on Build GRCh38
rs772947097 Chr.17: 76625901-76625901 on Build GRCh38
rs760203239 Chr.17: 76626130-76626130 on Build GRCh38
rs766181018 Chr.17: 76625998-76625998 on Build GRCh38
rs774077279 Chr.17: 76626003-76626003 on Build GRCh38
rs773491014 Chr.17: 76625988-76625988 on Build GRCh38
rs774170556 Chr.17: 76626099-76626099 on Build GRCh38
rs201736626 Chr.17: 76626022-76626022 on Build GRCh38
rs151127625 Chr.17: 76626021-76626021 on Build GRCh38
rs200612139 Chr.17: 76626098-76626098 on Build GRCh38
rs780931359 Chr.17: 76625812-76625812 on Build GRCh38
rs374505713 Chr.17: 76625799-76625799 on Build GRCh38
rs772562940 Chr.17: 76625961-76625961 on Build GRCh38
rs542407048 Chr.17: 76626043-76626043 on Build GRCh38
rs755893812 Chr.17: 76625826-76625826 on Build GRCh38
rs1043759207 Chr.17: 76625975-76625975 on Build GRCh38
rs1021572626 Chr.17: 76625781-76625781 on Build GRCh38
rs766337122 Chr.17: 76625942-76625944 on Build GRCh38
rs531716573 Chr.17: 76626066-76626066 on Build GRCh38
rs759451622 Chr.17: 76626070-76626070 on Build GRCh38
rs746991796 Chr.17: 76625859-76625859 on Build GRCh38
rs146451248 Chr.17: 76625986-76625986 on Build GRCh38
rs372411661 Chr.17: 76625907-76625907 on Build GRCh38
rs556241177 Chr.17: 76625731-76625731 on Build GRCh38
rs964161553 Chr.17: 76625755-76625755 on Build GRCh38
rs763681599 Chr.17: 76626144-76626144 on Build GRCh38
rs142980817 Chr.17: 76626015-76626015 on Build GRCh38
rs750259799 Chr.17: 76625786-76625786 on Build GRCh38
rs748162481 Chr.17: 76626072-76626072 on Build GRCh38
rs376555341 Chr.17: 76626073-76626073 on Build GRCh38
rs569353015 Chr.17: 76626037-76626037 on Build GRCh38
rs1021687684 Chr.17: 76626103-76626103 on Build GRCh38
rs748723563 Chr.17: 76625853-76625853 on Build GRCh38
rs762959525 Chr.17: 76625850-76625868 on Build GRCh38
rs765491975 Chr.17: 76625892-76625892 on Build GRCh38
rs772409657 Chr.17: 76625841-76625841 on Build GRCh38
rs960905570 Chr.17: 76626143-76626143 on Build GRCh38
rs767260774 Chr.17: 76626084-76626084 on Build GRCh38
rs777590330 Chr.17: 76625951-76625951 on Build GRCh38
rs756929387 Chr.17: 76626050-76626050 on Build GRCh38
rs760353807 Chr.17: 76626014-76626014 on Build GRCh38
rs751452974 Chr.17: 76625946-76625971 on Build GRCh38
rs890938444 Chr.17: 76625730-76625730 on Build GRCh38
rs370498566 Chr.17: 76625982-76625982 on Build GRCh38
rs545202208 Chr.17: 76625838-76625838 on Build GRCh38
rs967390171 Chr.17: 76625789-76625789 on Build GRCh38
rs769192071 Chr.17: 76625969-76625969 on Build GRCh38
rs761992560 Chr.17: 76625890-76625890 on Build GRCh38
rs375799951 Chr.17: 76625927-76625927 on Build GRCh38
rs759423551 Chr.17: 76626108-76626108 on Build GRCh38
rs990522047 Chr.17: 76625776-76625776 on Build GRCh38
rs747548501 Chr.17: 76625963-76625963 on Build GRCh38
rs371043875 Chr.17: 76625871-76625871 on Build GRCh38

View Full Product Details