Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 41955364-41955901 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM33
Entrez Gene ID
55161
Gene Name
transmembrane protein 33
Gene Aliases
1600019D15Rik, SHINC-3, SHINC3
UniGene
Hs.31082
Gene Chromosome Location
Chr.4: 41935120-41960807 on Build GRCh38
Transcripts
NM_018126.2 XM_005248116.3 XM_005248117.2

SNP Detail

SNP ID Chromosome Location
rs867707512 Chr.4: 41955622-41955622 on Build GRCh38
rs922203058 Chr.4: 41955645-41955645 on Build GRCh38
rs148216994 Chr.4: 41955420-41955420 on Build GRCh38
rs1014345009 Chr.4: 41955553-41955553 on Build GRCh38
rs974943698 Chr.4: 41955642-41955642 on Build GRCh38
rs915245078 Chr.4: 41955488-41955488 on Build GRCh38
rs1001382082 Chr.4: 41955819-41955819 on Build GRCh38
rs1004785415 Chr.4: 41955721-41955721 on Build GRCh38
rs374450065 Chr.4: 41955590-41955590 on Build GRCh38
rs551762104 Chr.4: 41955408-41955408 on Build GRCh38
rs190805767 Chr.4: 41955768-41955768 on Build GRCh38
rs990825698 Chr.4: 41955470-41955470 on Build GRCh38
rs781393765 Chr.4: 41955454-41955454 on Build GRCh38
rs536462633 Chr.4: 41955492-41955492 on Build GRCh38
rs187225506 Chr.4: 41955453-41955453 on Build GRCh38
rs556691026 Chr.4: 41955526-41955526 on Build GRCh38
rs757299705 Chr.4: 41955422-41955422 on Build GRCh38
rs978516573 Chr.4: 41955594-41955594 on Build GRCh38
rs572500515 Chr.4: 41955842-41955842 on Build GRCh38
rs898974898 Chr.4: 41955817-41955817 on Build GRCh38
rs1051830369 Chr.4: 41955413-41955413 on Build GRCh38
rs967530388 Chr.4: 41955593-41955593 on Build GRCh38
rs554274071 Chr.4: 41955444-41955444 on Build GRCh38
rs1048753374 Chr.4: 41955667-41955667 on Build GRCh38
rs959704463 Chr.4: 41955538-41955538 on Build GRCh38
rs769767041 Chr.4: 41955670-41955670 on Build GRCh38
rs1003120466 Chr.4: 41955795-41955795 on Build GRCh38
rs900258226 Chr.4: 41955644-41955644 on Build GRCh38
rs933294350 Chr.4: 41955656-41955656 on Build GRCh38
rs928055490 Chr.4: 41955447-41955447 on Build GRCh38
rs527245265 Chr.4: 41955864-41955864 on Build GRCh38
rs1054382 Chr.4: 41955758-41955758 on Build GRCh38
rs1019993276 Chr.4: 41955577-41955577 on Build GRCh38
rs907902616 Chr.4: 41955700-41955700 on Build GRCh38
rs1014732811 Chr.4: 41955419-41955419 on Build GRCh38
rs1042651485 Chr.4: 41955525-41955525 on Build GRCh38
rs959443588 Chr.4: 41955461-41955461 on Build GRCh38
rs141180492 Chr.4: 41955640-41955640 on Build GRCh38
rs1052194233 Chr.4: 41955677-41955677 on Build GRCh38
rs199768495 Chr.4: 41955734-41955734 on Build GRCh38
rs779771336 Chr.4: 41955701-41955701 on Build GRCh38
rs925442503 Chr.4: 41955599-41955599 on Build GRCh38
rs11934922 Chr.4: 41955414-41955414 on Build GRCh38
rs940757533 Chr.4: 41955712-41955712 on Build GRCh38
rs1055685640 Chr.4: 41955853-41955853 on Build GRCh38
rs557781425 Chr.4: 41955754-41955754 on Build GRCh38
rs548330270 Chr.4: 41955521-41955521 on Build GRCh38
rs971313620 Chr.4: 41955762-41955762 on Build GRCh38
rs951998064 Chr.4: 41955388-41955388 on Build GRCh38
rs545676976 Chr.4: 41955715-41955715 on Build GRCh38

View Full Product Details