Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 247867362-247867907 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRIM58
Entrez Gene ID
25893
Gene Name
tripartite motif containing 58
Gene Aliases
BIA2
UniGene
Hs.269151
Gene Chromosome Location
Chr.1: 247857199-247880138 on Build GRCh38
Transcripts
NM_015431.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5911545 c.748-8C>T p.? Chr.1: 247867837-247867837 on Build GRCh38
3977435 c.770+9G>A p.? Chr.1: 247867876-247867876 on Build GRCh38
5934447 c.757G>A p.G253R Chr.1: 247867854-247867854 on Build GRCh38
6062876 c.762_763CC>AA p.L255M Chr.1: 247867859-247867860 on Build GRCh38
680579 c.748G>T p.G250C Chr.1: 247867845-247867845 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs977438920 Chr.1: 247867726-247867726 on Build GRCh38
rs12121036 Chr.1: 247867547-247867547 on Build GRCh38
rs769868779 Chr.1: 247867870-247867870 on Build GRCh38
rs997652151 Chr.1: 247867667-247867667 on Build GRCh38
rs906291942 Chr.1: 247867625-247867625 on Build GRCh38
rs968703406 Chr.1: 247867491-247867491 on Build GRCh38
rs1048946111 Chr.1: 247867452-247867452 on Build GRCh38
rs889988671 Chr.1: 247867717-247867717 on Build GRCh38
rs573641377 Chr.1: 247867445-247867445 on Build GRCh38
rs3762324 Chr.1: 247867597-247867597 on Build GRCh38
rs747506379 Chr.1: 247867828-247867828 on Build GRCh38
rs1033492524 Chr.1: 247867426-247867426 on Build GRCh38
rs1030215537 Chr.1: 247867547-247867548 on Build GRCh38
rs562876779 Chr.1: 247867610-247867610 on Build GRCh38
rs781188527 Chr.1: 247867866-247867866 on Build GRCh38
rs1056316525 Chr.1: 247867611-247867611 on Build GRCh38
rs1003788615 Chr.1: 247867412-247867412 on Build GRCh38
rs1012093859 Chr.1: 247867431-247867431 on Build GRCh38
rs924055623 Chr.1: 247867582-247867582 on Build GRCh38
rs1033957281 Chr.1: 247867635-247867635 on Build GRCh38
rs756912918 Chr.1: 247867798-247867798 on Build GRCh38
rs998372710 Chr.1: 247867522-247867522 on Build GRCh38
rs966583010 Chr.1: 247867682-247867682 on Build GRCh38
rs189142192 Chr.1: 247867555-247867555 on Build GRCh38
rs546179289 Chr.1: 247867665-247867665 on Build GRCh38
rs181636728 Chr.1: 247867566-247867566 on Build GRCh38
rs748395145 Chr.1: 247867868-247867868 on Build GRCh38
rs984221417 Chr.1: 247867872-247867872 on Build GRCh38
rs1004507418 Chr.1: 247867644-247867644 on Build GRCh38
rs551865366 Chr.1: 247867636-247867636 on Build GRCh38
rs147541167 Chr.1: 247867801-247867801 on Build GRCh38
rs573009814 Chr.1: 247867565-247867565 on Build GRCh38
rs138725505 Chr.1: 247867506-247867506 on Build GRCh38
rs1031643711 Chr.1: 247867759-247867759 on Build GRCh38
rs573587477 Chr.1: 247867574-247867574 on Build GRCh38
rs539498663 Chr.1: 247867525-247867525 on Build GRCh38
rs543988426 Chr.1: 247867620-247867620 on Build GRCh38
rs758767994 Chr.1: 247867821-247867821 on Build GRCh38
rs989380579 Chr.1: 247867480-247867480 on Build GRCh38
rs201016573 Chr.1: 247867823-247867823 on Build GRCh38
rs914600058 Chr.1: 247867483-247867483 on Build GRCh38
rs1022595771 Chr.1: 247867753-247867753 on Build GRCh38
rs1010866902 Chr.1: 247867650-247867650 on Build GRCh38
rs878881445 Chr.1: 247867699-247867699 on Build GRCh38
rs576339515 Chr.1: 247867794-247867794 on Build GRCh38
rs959563472 Chr.1: 247867471-247867471 on Build GRCh38
rs944114294 Chr.1: 247867532-247867532 on Build GRCh38
rs924697023 Chr.1: 247867507-247867507 on Build GRCh38
rs113791151 Chr.1: 247867690-247867691 on Build GRCh38
rs575294052 Chr.1: 247867509-247867509 on Build GRCh38
rs535454445 Chr.1: 247867465-247867465 on Build GRCh38
rs932815884 Chr.1: 247867864-247867864 on Build GRCh38
rs376726654 Chr.1: 247867820-247867820 on Build GRCh38
rs1007057585 Chr.1: 247867724-247867724 on Build GRCh38
rs184783795 Chr.1: 247867573-247867573 on Build GRCh38
rs372133393 Chr.1: 247867884-247867884 on Build GRCh38
rs768724055 Chr.1: 247867859-247867859 on Build GRCh38
rs1041623011 Chr.1: 247867544-247867544 on Build GRCh38
rs760007883 Chr.1: 247867626-247867626 on Build GRCh38
rs377193344 Chr.1: 247867663-247867663 on Build GRCh38
rs1022212566 Chr.1: 247867677-247867677 on Build GRCh38
rs888148500 Chr.1: 247867571-247867571 on Build GRCh38
rs901606932 Chr.1: 247867652-247867652 on Build GRCh38
rs916437652 Chr.1: 247867617-247867617 on Build GRCh38

View Full Product Details