Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 51290001-51290546 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TTC39A
Entrez Gene ID
22996
Gene Name
tetratricopeptide repeat domain 39A
Gene Aliases
C1orf34, DEME-6
UniGene
Hs.112949
Gene Chromosome Location
Chr.1: 51287258-51345116 on Build GRCh38
Transcripts
NM_001080494.3 NM_001144832.2 NM_001297663.1 NM_001297664.1 NM_001297665.1 NM_001297666.1 NM_001297667.1 XM_005270644.2 XM_006710471.2 XM_011541048.2 XM_011541049.2 XM_011541050.2 XM_011541051.2 XM_011541052.2 XM_011541053.2 XM_011541054.2 XM_017000715.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1237961 c.1534G>A p.G512S Chr.1: 51290072-51290072 on Build GRCh38
1492068 c.1298-1G>T p.? Chr.1: 51290120-51290120 on Build GRCh38
1492067 c.1487-1G>T p.? Chr.1: 51290120-51290120 on Build GRCh38
1237962 c.1345G>A p.G449S Chr.1: 51290072-51290072 on Build GRCh38
4008714 c.1493A>G p.E498G Chr.1: 51290113-51290113 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1055028522 Chr.1: 51290203-51290203 on Build GRCh38
rs764916725 Chr.1: 51290093-51290093 on Build GRCh38
rs965980130 Chr.1: 51290352-51290352 on Build GRCh38
rs74080518 Chr.1: 51290314-51290314 on Build GRCh38
rs985069924 Chr.1: 51290332-51290332 on Build GRCh38
rs149349338 Chr.1: 51290440-51290440 on Build GRCh38
rs147276961 Chr.1: 51290245-51290245 on Build GRCh38
rs896409139 Chr.1: 51290455-51290455 on Build GRCh38
rs896970968 Chr.1: 51290476-51290476 on Build GRCh38
rs962401549 Chr.1: 51290099-51290099 on Build GRCh38
rs185532830 Chr.1: 51290134-51290134 on Build GRCh38
rs1036941678 Chr.1: 51290328-51290328 on Build GRCh38
rs752716688 Chr.1: 51290097-51290097 on Build GRCh38
rs749735901 Chr.1: 51290045-51290045 on Build GRCh38
rs140853407 Chr.1: 51290370-51290370 on Build GRCh38
rs757556113 Chr.1: 51290123-51290123 on Build GRCh38
rs776124288 Chr.1: 51290044-51290044 on Build GRCh38
rs764290856 Chr.1: 51290114-51290114 on Build GRCh38
rs1013433154 Chr.1: 51290236-51290236 on Build GRCh38
rs989358616 Chr.1: 51290106-51290106 on Build GRCh38
rs769658596 Chr.1: 51290521-51290521 on Build GRCh38
rs750600095 Chr.1: 51290145-51290145 on Build GRCh38
rs756860100 Chr.1: 51290329-51290329 on Build GRCh38
rs949285746 Chr.1: 51290460-51290460 on Build GRCh38
rs746453366 Chr.1: 51290490-51290490 on Build GRCh38
rs548679765 Chr.1: 51290150-51290150 on Build GRCh38
rs528205306 Chr.1: 51290267-51290267 on Build GRCh38
rs927300973 Chr.1: 51290436-51290436 on Build GRCh38
rs760330308 Chr.1: 51290167-51290167 on Build GRCh38
rs373393772 Chr.1: 51290495-51290495 on Build GRCh38
rs373716832 Chr.1: 51290330-51290330 on Build GRCh38
rs759430309 Chr.1: 51290091-51290091 on Build GRCh38
rs947529860 Chr.1: 51290385-51290385 on Build GRCh38
rs41287286 Chr.1: 51290278-51290278 on Build GRCh38
rs995212815 Chr.1: 51290307-51290307 on Build GRCh38
rs978827134 Chr.1: 51290415-51290415 on Build GRCh38
rs759076527 Chr.1: 51290509-51290509 on Build GRCh38
rs200700558 Chr.1: 51290473-51290473 on Build GRCh38
rs777280684 Chr.1: 51290477-51290477 on Build GRCh38
rs138832140 Chr.1: 51290034-51290034 on Build GRCh38
rs937939319 Chr.1: 51290185-51290185 on Build GRCh38
rs775098140 Chr.1: 51290522-51290522 on Build GRCh38
rs956231744 Chr.1: 51290171-51290171 on Build GRCh38
rs1019880360 Chr.1: 51290343-51290343 on Build GRCh38
rs746175223 Chr.1: 51290033-51290033 on Build GRCh38
rs545533136 Chr.1: 51290109-51290109 on Build GRCh38
rs770410982 Chr.1: 51290494-51290494 on Build GRCh38
rs749836362 Chr.1: 51290472-51290472 on Build GRCh38
rs776297151 Chr.1: 51290085-51290085 on Build GRCh38
rs545023480 Chr.1: 51290057-51290057 on Build GRCh38
rs988997242 Chr.1: 51290364-51290364 on Build GRCh38
rs191134345 Chr.1: 51290234-51290234 on Build GRCh38
rs780251200 Chr.1: 51290465-51290465 on Build GRCh38
rs143899586 Chr.1: 51290048-51290048 on Build GRCh38
rs575144254 Chr.1: 51290229-51290229 on Build GRCh38
rs1057266564 Chr.1: 51290274-51290274 on Build GRCh38
rs905855066 Chr.1: 51290179-51290179 on Build GRCh38
rs369258870 Chr.1: 51290153-51290153 on Build GRCh38
rs1026567036 Chr.1: 51290037-51290037 on Build GRCh38
rs374421872 Chr.1: 51290115-51290115 on Build GRCh38
rs368894146 Chr.1: 51290047-51290047 on Build GRCh38
rs749926888 Chr.1: 51290451-51290451 on Build GRCh38
rs764607869 Chr.1: 51290187-51290187 on Build GRCh38
rs995456540 Chr.1: 51290336-51290336 on Build GRCh38
rs995613849 Chr.1: 51290112-51290112 on Build GRCh38
rs938719943 Chr.1: 51290251-51290251 on Build GRCh38
rs867191270 Chr.1: 51290036-51290036 on Build GRCh38

View Full Product Details