Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 75983462-75984005 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UVRAG
Entrez Gene ID
7405
Gene Name
UV radiation resistance associated
Gene Aliases
DHTX, VPS38, p63
UniGene
Hs.202470
Gene Chromosome Location
Chr.11: 75814852-76144238 on Build GRCh38
Transcripts
NM_003369.3 XM_011545226.2 XM_011545227.2 XM_011545228.2 XM_017018224.1 XM_017018225.1 XM_017018226.1 XM_017018227.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4036760 c.787G>A p.A263T Chr.11: 75983474-75983474 on Build GRCh38
4036759 c.778C>T p.R260W Chr.11: 75983465-75983465 on Build GRCh38
1270138 c.792A>G p.L264L Chr.11: 75983479-75983479 on Build GRCh38
7250922 c.778C>G p.R260G Chr.11: 75983465-75983465 on Build GRCh38
5999581 c.826G>A p.G276R Chr.11: 75983513-75983513 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757216549 Chr.11: 75983527-75983527 on Build GRCh38
rs976786538 Chr.11: 75983774-75983774 on Build GRCh38
rs551789386 Chr.11: 75983708-75983708 on Build GRCh38
rs1025915860 Chr.11: 75983595-75983596 on Build GRCh38
rs564005947 Chr.11: 75983575-75983575 on Build GRCh38
rs993257292 Chr.11: 75983590-75983590 on Build GRCh38
rs1035840250 Chr.11: 75983639-75983639 on Build GRCh38
rs888991217 Chr.11: 75983839-75983839 on Build GRCh38
rs745314997 Chr.11: 75983532-75983532 on Build GRCh38
rs781295105 Chr.11: 75983530-75983530 on Build GRCh38
rs189400179 Chr.11: 75983588-75983588 on Build GRCh38
rs571868298 Chr.11: 75983804-75983804 on Build GRCh38
rs897510976 Chr.11: 75983964-75983964 on Build GRCh38
rs758196665 Chr.11: 75983484-75983484 on Build GRCh38
rs560018304 Chr.11: 75983712-75983712 on Build GRCh38
rs759670313 Chr.11: 75983550-75983551 on Build GRCh38
rs934717493 Chr.11: 75983764-75983764 on Build GRCh38
rs535393253 Chr.11: 75983931-75983931 on Build GRCh38
rs747757552 Chr.11: 75983800-75983800 on Build GRCh38
rs111304511 Chr.11: 75983593-75983593 on Build GRCh38
rs773406795 Chr.11: 75983549-75983549 on Build GRCh38
rs984471655 Chr.11: 75983650-75983650 on Build GRCh38
rs924623755 Chr.11: 75983733-75983733 on Build GRCh38
rs748943576 Chr.11: 75983545-75983545 on Build GRCh38
rs991702476 Chr.11: 75983669-75983669 on Build GRCh38
rs905301305 Chr.11: 75983831-75983831 on Build GRCh38
rs868101159 Chr.11: 75983611-75983611 on Build GRCh38
rs769495942 Chr.11: 75983538-75983538 on Build GRCh38
rs933191268 Chr.11: 75983488-75983488 on Build GRCh38
rs551863526 Chr.11: 75983728-75983728 on Build GRCh38
rs1056674448 Chr.11: 75983855-75983855 on Build GRCh38
rs993336233 Chr.11: 75983874-75983874 on Build GRCh38
rs772449673 Chr.11: 75983548-75983548 on Build GRCh38
rs61749241 Chr.11: 75983505-75983505 on Build GRCh38
rs1052687653 Chr.11: 75983768-75983768 on Build GRCh38
rs528720330 Chr.11: 75983714-75983714 on Build GRCh38
rs79648481 Chr.11: 75983799-75983799 on Build GRCh38
rs767721495 Chr.11: 75983559-75983561 on Build GRCh38
rs570926032 Chr.11: 75983967-75983967 on Build GRCh38
rs754497816 Chr.11: 75983612-75983612 on Build GRCh38
rs143426525 Chr.11: 75983486-75983486 on Build GRCh38
rs752954555 Chr.11: 75983560-75983560 on Build GRCh38
rs369799282 Chr.11: 75983540-75983540 on Build GRCh38
rs1026197642 Chr.11: 75983875-75983875 on Build GRCh38
rs569923829 Chr.11: 75983826-75983826 on Build GRCh38
rs571658082 Chr.11: 75983731-75983731 on Build GRCh38
rs780707413 Chr.11: 75983743-75983743 on Build GRCh38
rs1023632799 Chr.11: 75983681-75983681 on Build GRCh38
rs1043829799 Chr.11: 75983806-75983806 on Build GRCh38
rs550742454 Chr.11: 75983810-75983810 on Build GRCh38
rs755766288 Chr.11: 75983937-75983938 on Build GRCh38
rs1037339813 Chr.11: 75983958-75983958 on Build GRCh38
rs761050220 Chr.11: 75983551-75983551 on Build GRCh38
rs1002033126 Chr.11: 75983853-75983853 on Build GRCh38
rs563856092 Chr.11: 75983482-75983482 on Build GRCh38
rs896340392 Chr.11: 75983863-75983863 on Build GRCh38
rs757392953 Chr.11: 75983567-75983569 on Build GRCh38
rs549586435 Chr.11: 75983662-75983662 on Build GRCh38
rs755768347 Chr.11: 75983928-75983928 on Build GRCh38
rs868252569 Chr.11: 75983739-75983739 on Build GRCh38
rs887437548 Chr.11: 75983930-75983930 on Build GRCh38

View Full Product Details