Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 58033890-58034383 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZSCAN1
Entrez Gene ID
284312
Gene Name
zinc finger and SCAN domain containing 1
Gene Aliases
MZF-1, ZNF915
UniGene
Hs.643437
Gene Chromosome Location
Chr.19: 58033989-58058907 on Build GRCh38
Transcripts
NM_182572.3 XM_017026609.1 XM_017026610.1

SNP Detail

SNP ID Chromosome Location
rs893461026 Chr.19: 58034131-58034172 on Build GRCh38
rs1020802758 Chr.19: 58034308-58034308 on Build GRCh38
rs1056383401 Chr.19: 58034288-58034288 on Build GRCh38
rs1036108856 Chr.19: 58033949-58033949 on Build GRCh38
rs113348493 Chr.19: 58033991-58033991 on Build GRCh38
rs1003447595 Chr.19: 58034284-58034284 on Build GRCh38
rs922197828 Chr.19: 58034129-58034129 on Build GRCh38
rs905013114 Chr.19: 58034185-58034185 on Build GRCh38
rs905945964 Chr.19: 58033912-58033912 on Build GRCh38
rs1002454362 Chr.19: 58033919-58033919 on Build GRCh38
rs940829367 Chr.19: 58034134-58034134 on Build GRCh38
rs940465269 Chr.19: 58033932-58033932 on Build GRCh38
rs898827216 Chr.19: 58033959-58033959 on Build GRCh38
rs1020330308 Chr.19: 58034078-58034078 on Build GRCh38
rs545904592 Chr.19: 58034044-58034044 on Build GRCh38
rs1054936975 Chr.19: 58033922-58033922 on Build GRCh38
rs1044731187 Chr.19: 58034277-58034277 on Build GRCh38
rs1051910888 Chr.19: 58033929-58033929 on Build GRCh38
rs1002257968 Chr.19: 58034292-58034292 on Build GRCh38
rs1010777779 Chr.19: 58034060-58034060 on Build GRCh38
rs191217883 Chr.19: 58033962-58033962 on Build GRCh38
rs532453563 Chr.19: 58034015-58034015 on Build GRCh38
rs920328856 Chr.19: 58034215-58034215 on Build GRCh38
rs570597129 Chr.19: 58034243-58034243 on Build GRCh38
rs495839 Chr.19: 58034132-58034132 on Build GRCh38
rs969133376 Chr.19: 58034093-58034093 on Build GRCh38
rs904854320 Chr.19: 58034280-58034280 on Build GRCh38
rs548002064 Chr.19: 58034183-58034183 on Build GRCh38
rs78468068 Chr.19: 58033911-58033911 on Build GRCh38
rs1003395043 Chr.19: 58034000-58034000 on Build GRCh38
rs620297 Chr.19: 58034065-58034065 on Build GRCh38
rs144900819 Chr.19: 58034045-58034046 on Build GRCh38
rs531925689 Chr.19: 58034066-58034066 on Build GRCh38
rs968808910 Chr.19: 58034312-58034312 on Build GRCh38
rs897736513 Chr.19: 58033917-58033917 on Build GRCh38
rs912047173 Chr.19: 58033931-58033931 on Build GRCh38
rs1034843028 Chr.19: 58034014-58034014 on Build GRCh38
rs568676860 Chr.19: 58034088-58034088 on Build GRCh38
rs1000263351 Chr.19: 58034352-58034352 on Build GRCh38
rs1044973833 Chr.19: 58033965-58033965 on Build GRCh38
rs912080878 Chr.19: 58034133-58034133 on Build GRCh38
rs775815486 Chr.19: 58034040-58034040 on Build GRCh38
rs892298836 Chr.19: 58034294-58034294 on Build GRCh38
rs548826191 Chr.19: 58034082-58034082 on Build GRCh38
rs539498494 Chr.19: 58034289-58034289 on Build GRCh38
rs1009371026 Chr.19: 58034294-58034295 on Build GRCh38
rs978732014 Chr.19: 58034127-58034127 on Build GRCh38

View Full Product Details