Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 164769750-164770249 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC100505795
Entrez Gene ID
100505795
Gene Name
uncharacterized LOC100505795
Gene Aliases
-
UniGene
Hs.661039
Gene Chromosome Location
Chr.1: 164769116-164774641 on Build GRCh38
Transcripts
NR_038072.1 NM_001204961.1 NM_001204963.1 NM_002585.3 XM_005245228.3 XM_005245229.3 XM_011509590.1 XM_011509591.2 XM_011509592.1 XM_017001395.1 XM_017001396.1

Gene Symbol
PBX1
Entrez Gene ID
5087
Gene Name
PBX homeobox 1
Gene Aliases
-
UniGene
Hs.557097
Gene Chromosome Location
Chr.1: 164559360-164886047 on Build GRCh38
Transcripts
NR_038072.1 NM_001204961.1 NM_001204963.1 NM_002585.3 XM_005245228.3 XM_005245229.3 XM_011509590.1 XM_011509591.2 XM_011509592.1 XM_017001395.1 XM_017001396.1

SNP Detail

SNP ID Chromosome Location
rs764468552 Chr.1: 164770222-164770222 on Build GRCh38
rs923857822 Chr.1: 164770122-164770122 on Build GRCh38
rs1026559455 Chr.1: 164769966-164769966 on Build GRCh38
rs559027849 Chr.1: 164770036-164770036 on Build GRCh38
rs137961040 Chr.1: 164770007-164770007 on Build GRCh38
rs562419111 Chr.1: 164769800-164769800 on Build GRCh38
rs1014066685 Chr.1: 164769912-164769912 on Build GRCh38
rs976389330 Chr.1: 164770073-164770073 on Build GRCh38
rs568202414 Chr.1: 164769891-164769891 on Build GRCh38
rs1049492506 Chr.1: 164769842-164769842 on Build GRCh38
rs539013121 Chr.1: 164770008-164770008 on Build GRCh38
rs1024573307 Chr.1: 164769916-164769916 on Build GRCh38
rs950983787 Chr.1: 164769968-164769968 on Build GRCh38
rs913458913 Chr.1: 164770019-164770019 on Build GRCh38
rs985042398 Chr.1: 164769969-164769969 on Build GRCh38
rs932389386 Chr.1: 164769821-164769821 on Build GRCh38
rs771809651 Chr.1: 164770079-164770079 on Build GRCh38
rs12732181 Chr.1: 164770177-164770177 on Build GRCh38
rs564046832 Chr.1: 164769894-164769894 on Build GRCh38
rs990954027 Chr.1: 164770021-164770021 on Build GRCh38
rs909645581 Chr.1: 164770190-164770190 on Build GRCh38
rs774977632 Chr.1: 164770127-164770127 on Build GRCh38
rs773979615 Chr.1: 164769939-164769939 on Build GRCh38
rs1033128101 Chr.1: 164769985-164769985 on Build GRCh38
rs185141949 Chr.1: 164770044-164770044 on Build GRCh38
rs1053718262 Chr.1: 164770136-164770136 on Build GRCh38
rs538583846 Chr.1: 164770088-164770088 on Build GRCh38
rs536643122 Chr.1: 164769788-164769788 on Build GRCh38
rs547979220 Chr.1: 164769872-164769872 on Build GRCh38
rs917712826 Chr.1: 164769784-164769784 on Build GRCh38
rs554965976 Chr.1: 164770101-164770101 on Build GRCh38
rs1008023626 Chr.1: 164769875-164769875 on Build GRCh38
rs529944046 Chr.1: 164770161-164770161 on Build GRCh38
rs1054904130 Chr.1: 164769871-164769871 on Build GRCh38
rs1040957961 Chr.1: 164769783-164769783 on Build GRCh38
rs547039790 Chr.1: 164769981-164769981 on Build GRCh38
rs949211389 Chr.1: 164770074-164770074 on Build GRCh38
rs143502812 Chr.1: 164769788-164769788 on Build GRCh38
rs914529857 Chr.1: 164770145-164770145 on Build GRCh38
rs897042506 Chr.1: 164770225-164770225 on Build GRCh38
rs181094763 Chr.1: 164769853-164769853 on Build GRCh38
rs957324243 Chr.1: 164769999-164769999 on Build GRCh38
rs1039515232 Chr.1: 164770215-164770215 on Build GRCh38
rs934655984 Chr.1: 164769869-164769869 on Build GRCh38
rs189300026 Chr.1: 164770223-164770223 on Build GRCh38
rs6670655 Chr.1: 164769934-164769934 on Build GRCh38

View Full Product Details