Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 75360808-75361353 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF503
Entrez Gene ID
84858
Gene Name
zinc finger protein 503
Gene Aliases
NOLZ-1, NOLZ1, Nlz2
UniGene
Hs.195710
Gene Chromosome Location
Chr.10: 75279726-75401894 on Build GRCh38
Transcripts
NR_120651.1 NR_038223.1 NR_038224.1 NR_038225.1 NR_038226.1

Gene Symbol
ZNF503-AS1
Entrez Gene ID
253264
Gene Name
ZNF503 antisense RNA 1
Gene Aliases
-
UniGene
Hs.662111
Gene Chromosome Location
Chr.10: 75296383-75361678 on Build GRCh38
Transcripts
NR_120651.1 NR_038223.1 NR_038224.1 NR_038225.1 NR_038226.1

SNP Detail

SNP ID Chromosome Location
rs553860792 Chr.10: 75361276-75361276 on Build GRCh38
rs769627507 Chr.10: 75361185-75361185 on Build GRCh38
rs1000232259 Chr.10: 75361308-75361308 on Build GRCh38
rs926071504 Chr.10: 75360859-75360859 on Build GRCh38
rs775217159 Chr.10: 75361302-75361302 on Build GRCh38
rs998069799 Chr.10: 75361265-75361265 on Build GRCh38
rs150047026 Chr.10: 75360847-75360848 on Build GRCh38
rs568643562 Chr.10: 75361257-75361257 on Build GRCh38
rs911062163 Chr.10: 75361224-75361224 on Build GRCh38
rs998166993 Chr.10: 75360831-75360831 on Build GRCh38
rs574494366 Chr.10: 75361130-75361130 on Build GRCh38
rs138017641 Chr.10: 75360858-75360858 on Build GRCh38
rs1004034971 Chr.10: 75361174-75361174 on Build GRCh38
rs1048127585 Chr.10: 75361161-75361161 on Build GRCh38
rs937681902 Chr.10: 75361105-75361105 on Build GRCh38
rs972788600 Chr.10: 75361119-75361119 on Build GRCh38
rs950324150 Chr.10: 75360971-75360971 on Build GRCh38
rs748537318 Chr.10: 75361311-75361311 on Build GRCh38
rs41280440 Chr.10: 75361171-75361171 on Build GRCh38
rs923735969 Chr.10: 75361310-75361310 on Build GRCh38
rs35207267 Chr.10: 75361309-75361310 on Build GRCh38
rs573476909 Chr.10: 75361301-75361301 on Build GRCh38
rs956224921 Chr.10: 75360860-75360860 on Build GRCh38
rs1035672672 Chr.10: 75361069-75361069 on Build GRCh38
rs537475417 Chr.10: 75361075-75361075 on Build GRCh38
rs1011751359 Chr.10: 75360893-75360893 on Build GRCh38
rs182679118 Chr.10: 75360888-75360888 on Build GRCh38
rs556099804 Chr.10: 75361079-75361079 on Build GRCh38
rs989414224 Chr.10: 75360908-75360908 on Build GRCh38
rs530133522 Chr.10: 75360855-75360855 on Build GRCh38
rs570689784 Chr.10: 75361032-75361032 on Build GRCh38
rs917490495 Chr.10: 75360931-75360931 on Build GRCh38
rs1044640779 Chr.10: 75360990-75360990 on Build GRCh38
rs1002972282 Chr.10: 75360997-75360997 on Build GRCh38
rs954036656 Chr.10: 75360854-75360854 on Build GRCh38
rs185543436 Chr.10: 75360938-75360938 on Build GRCh38
rs943758750 Chr.10: 75361275-75361275 on Build GRCh38
rs764185404 Chr.10: 75361289-75361289 on Build GRCh38
rs1023144890 Chr.10: 75360953-75360953 on Build GRCh38
rs552784448 Chr.10: 75360961-75360961 on Build GRCh38
rs970098337 Chr.10: 75360977-75360977 on Build GRCh38
rs1262398 Chr.10: 75361324-75361324 on Build GRCh38
rs919828926 Chr.10: 75361164-75361164 on Build GRCh38
rs533694709 Chr.10: 75360935-75360935 on Build GRCh38

View Full Product Details