Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 2767276-2767776 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC283440
Entrez Gene ID
283440
Gene Name
uncharacterized LOC283440
Gene Aliases
-
UniGene
Hs.658536
Gene Chromosome Location
Chr.12: 2761200-2771669 on Build GRCh38
Transcripts
NR_033958.1

SNP Detail

SNP ID Chromosome Location
rs973529275 Chr.12: 2767578-2767578 on Build GRCh38
rs1013479966 Chr.12: 2767378-2767378 on Build GRCh38
rs1054892837 Chr.12: 2767569-2767569 on Build GRCh38
rs919812771 Chr.12: 2767736-2767736 on Build GRCh38
rs779789716 Chr.12: 2767366-2767366 on Build GRCh38
rs556835259 Chr.12: 2767631-2767631 on Build GRCh38
rs781196488 Chr.12: 2767408-2767408 on Build GRCh38
rs571135214 Chr.12: 2767430-2767430 on Build GRCh38
rs768562381 Chr.12: 2767625-2767625 on Build GRCh38
rs971607504 Chr.12: 2767405-2767405 on Build GRCh38
rs912487527 Chr.12: 2767477-2767477 on Build GRCh38
rs571049883 Chr.12: 2767727-2767727 on Build GRCh38
rs755640192 Chr.12: 2767543-2767543 on Build GRCh38
rs779215509 Chr.12: 2767700-2767700 on Build GRCh38
rs146792120 Chr.12: 2767406-2767408 on Build GRCh38
rs769370103 Chr.12: 2767442-2767442 on Build GRCh38
rs116543541 Chr.12: 2767439-2767439 on Build GRCh38
rs754909995 Chr.12: 2767383-2767383 on Build GRCh38
rs958887094 Chr.12: 2767374-2767374 on Build GRCh38
rs948082967 Chr.12: 2767413-2767413 on Build GRCh38
rs913790115 Chr.12: 2767712-2767712 on Build GRCh38
rs561837856 Chr.12: 2767678-2767678 on Build GRCh38
rs971681063 Chr.12: 2767632-2767632 on Build GRCh38
rs375565553 Chr.12: 2767557-2767557 on Build GRCh38
rs921320122 Chr.12: 2767447-2767447 on Build GRCh38
rs770757640 Chr.12: 2767499-2767499 on Build GRCh38
rs894849903 Chr.12: 2767580-2767580 on Build GRCh38
rs4313632 Chr.12: 2767470-2767470 on Build GRCh38
rs576177971 Chr.12: 2767668-2767668 on Build GRCh38
rs977455376 Chr.12: 2767426-2767426 on Build GRCh38
rs551032465 Chr.12: 2767367-2767367 on Build GRCh38
rs534746526 Chr.12: 2767500-2767500 on Build GRCh38
rs568306307 Chr.12: 2767532-2767532 on Build GRCh38
rs927940734 Chr.12: 2767679-2767679 on Build GRCh38
rs141769347 Chr.12: 2767459-2767459 on Build GRCh38
rs572885793 Chr.12: 2767667-2767667 on Build GRCh38
rs749267267 Chr.12: 2767609-2767609 on Build GRCh38
rs939961522 Chr.12: 2767713-2767713 on Build GRCh38
rs1036693498 Chr.12: 2767506-2767506 on Build GRCh38
rs1010659316 Chr.12: 2767445-2767445 on Build GRCh38
rs545749599 Chr.12: 2767641-2767641 on Build GRCh38
rs906486284 Chr.12: 2767430-2767431 on Build GRCh38
rs758146026 Chr.12: 2767330-2767330 on Build GRCh38
rs544795020 Chr.12: 2767694-2767694 on Build GRCh38
rs986685382 Chr.12: 2767693-2767693 on Build GRCh38
rs1036894022 Chr.12: 2767729-2767729 on Build GRCh38
rs530873879 Chr.12: 2767683-2767683 on Build GRCh38
rs368282650 Chr.12: 2767642-2767642 on Build GRCh38
rs537230331 Chr.12: 2767547-2767547 on Build GRCh38
rs892320775 Chr.12: 2767433-2767433 on Build GRCh38
rs146251233 Chr.12: 2767636-2767636 on Build GRCh38
rs750347788 Chr.12: 2767339-2767341 on Build GRCh38
rs954235765 Chr.12: 2767685-2767685 on Build GRCh38
rs957465071 Chr.12: 2767431-2767431 on Build GRCh38
rs1024564631 Chr.12: 2767390-2767390 on Build GRCh38
rs1052319226 Chr.12: 2767697-2767697 on Build GRCh38
rs930998635 Chr.12: 2767740-2767740 on Build GRCh38
rs779494876 Chr.12: 2767572-2767572 on Build GRCh38
rs554686261 Chr.12: 2767528-2767528 on Build GRCh38
rs1013094341 Chr.12: 2767581-2767581 on Build GRCh38
rs148477631 Chr.12: 2767752-2767752 on Build GRCh38
rs955006180 Chr.12: 2767501-2767501 on Build GRCh38
rs1015155957 Chr.12: 2767535-2767535 on Build GRCh38
rs771699065 Chr.12: 2767738-2767738 on Build GRCh38
rs77826753 Chr.12: 2767375-2767375 on Build GRCh38
rs894243455 Chr.12: 2767648-2767648 on Build GRCh38

View Full Product Details