Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 45341192-45341727 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TPT1
Entrez Gene ID
7178
Gene Name
tumor protein, translationally-controlled 1
Gene Aliases
HRF, TCTP, p02, p23
UniGene
Hs.374596
Gene Chromosome Location
Chr.13: 45333471-45341284 on Build GRCh38
Transcripts
NM_001286272.1 NM_001286273.1 NM_003295.3 NR_024458.1

Gene Symbol
TPT1-AS1
Entrez Gene ID
100190939
Gene Name
TPT1 antisense RNA 1
Gene Aliases
-
UniGene
Hs.375766
Gene Chromosome Location
Chr.13: 45341345-45391483 on Build GRCh38
Transcripts
NM_001286272.1 NM_001286273.1 NM_003295.3 NR_024458.1

SNP Detail

SNP ID Chromosome Location
rs1021630727 Chr.13: 45341223-45341223 on Build GRCh38
rs186141920 Chr.13: 45341665-45341665 on Build GRCh38
rs545702894 Chr.13: 45341572-45341572 on Build GRCh38
rs12872567 Chr.13: 45341365-45341365 on Build GRCh38
rs557431057 Chr.13: 45341514-45341514 on Build GRCh38
rs533567063 Chr.13: 45341597-45341597 on Build GRCh38
rs1041252461 Chr.13: 45341356-45341356 on Build GRCh38
rs778101473 Chr.13: 45341669-45341669 on Build GRCh38
rs908721703 Chr.13: 45341456-45341456 on Build GRCh38
rs947670863 Chr.13: 45341297-45341297 on Build GRCh38
rs996831982 Chr.13: 45341483-45341483 on Build GRCh38
rs998964856 Chr.13: 45341503-45341503 on Build GRCh38
rs759727702 Chr.13: 45341279-45341279 on Build GRCh38
rs963988661 Chr.13: 45341221-45341221 on Build GRCh38
rs751323856 Chr.13: 45341598-45341598 on Build GRCh38
rs894049582 Chr.13: 45341615-45341615 on Build GRCh38
rs906281099 Chr.13: 45341582-45341582 on Build GRCh38
rs770946079 Chr.13: 45341276-45341276 on Build GRCh38
rs1046156588 Chr.13: 45341580-45341580 on Build GRCh38
rs990070940 Chr.13: 45341447-45341447 on Build GRCh38
rs904533619 Chr.13: 45341640-45341640 on Build GRCh38
rs1019068037 Chr.13: 45341679-45341679 on Build GRCh38
rs969278841 Chr.13: 45341531-45341531 on Build GRCh38
rs919697881 Chr.13: 45341445-45341445 on Build GRCh38
rs940665163 Chr.13: 45341314-45341314 on Build GRCh38
rs1029121994 Chr.13: 45341452-45341452 on Build GRCh38
rs112020474 Chr.13: 45341366-45341366 on Build GRCh38
rs544550814 Chr.13: 45341674-45341674 on Build GRCh38
rs944244820 Chr.13: 45341591-45341591 on Build GRCh38
rs958284857 Chr.13: 45341440-45341440 on Build GRCh38
rs1001130814 Chr.13: 45341378-45341378 on Build GRCh38
rs900086803 Chr.13: 45341464-45341464 on Build GRCh38
rs953611235 Chr.13: 45341673-45341673 on Build GRCh38
rs931207416 Chr.13: 45341521-45341521 on Build GRCh38
rs1024793959 Chr.13: 45341677-45341677 on Build GRCh38
rs897055343 Chr.13: 45341336-45341336 on Build GRCh38
rs1048711574 Chr.13: 45341570-45341570 on Build GRCh38
rs572800859 Chr.13: 45341218-45341218 on Build GRCh38
rs532241014 Chr.13: 45341457-45341457 on Build GRCh38
rs999129960 Chr.13: 45341422-45341422 on Build GRCh38
rs989649159 Chr.13: 45341280-45341280 on Build GRCh38
rs779780909 Chr.13: 45341455-45341455 on Build GRCh38
rs562699784 Chr.13: 45341327-45341327 on Build GRCh38
rs918518114 Chr.13: 45341520-45341520 on Build GRCh38
rs931505601 Chr.13: 45341318-45341318 on Build GRCh38
rs917802661 Chr.13: 45341387-45341387 on Build GRCh38
rs980868998 Chr.13: 45341547-45341547 on Build GRCh38
rs1041283507 Chr.13: 45341460-45341460 on Build GRCh38
rs56949992 Chr.13: 45341571-45341571 on Build GRCh38
rs931057536 Chr.13: 45341454-45341454 on Build GRCh38
rs1031211483 Chr.13: 45341273-45341273 on Build GRCh38
rs970964310 Chr.13: 45341245-45341245 on Build GRCh38
rs560284337 Chr.13: 45341228-45341228 on Build GRCh38
rs968326038 Chr.13: 45341259-45341259 on Build GRCh38
rs922021939 Chr.13: 45341286-45341286 on Build GRCh38
rs571551847 Chr.13: 45341458-45341458 on Build GRCh38
rs1024557436 Chr.13: 45341222-45341222 on Build GRCh38
rs561842358 Chr.13: 45341277-45341277 on Build GRCh38
rs535000224 Chr.13: 45341290-45341291 on Build GRCh38
rs563914506 Chr.13: 45341309-45341309 on Build GRCh38
rs894867193 Chr.13: 45341214-45341214 on Build GRCh38
rs12872310 Chr.13: 45341346-45341346 on Build GRCh38
rs972031102 Chr.13: 45341519-45341519 on Build GRCh38
rs887379272 Chr.13: 45341337-45341337 on Build GRCh38
rs999159879 Chr.13: 45341584-45341584 on Build GRCh38
rs540176925 Chr.13: 45341272-45341272 on Build GRCh38
rs1031241481 Chr.13: 45341429-45341429 on Build GRCh38
rs1011966792 Chr.13: 45341388-45341388 on Build GRCh38
rs538560183 Chr.13: 45341400-45341400 on Build GRCh38
rs180684458 Chr.13: 45341511-45341511 on Build GRCh38
rs940694867 Chr.13: 45341515-45341515 on Build GRCh38
rs904499882 Chr.13: 45341502-45341502 on Build GRCh38
rs773132754 Chr.13: 45341228-45341228 on Build GRCh38
rs1043673951 Chr.13: 45341638-45341638 on Build GRCh38
rs987877712 Chr.13: 45341311-45341311 on Build GRCh38
rs550047611 Chr.13: 45341401-45341401 on Build GRCh38
rs76972495 Chr.13: 45341619-45341619 on Build GRCh38
rs762971028 Chr.13: 45341266-45341266 on Build GRCh38
rs113210477 Chr.13: 45341377-45341377 on Build GRCh38
rs992086507 Chr.13: 45341381-45341381 on Build GRCh38
rs779346973 Chr.13: 45341567-45341567 on Build GRCh38
rs1043045629 Chr.13: 45341498-45341498 on Build GRCh38
rs551945656 Chr.13: 45341359-45341359 on Build GRCh38
rs894891017 Chr.13: 45341380-45341380 on Build GRCh38
rs917604543 Chr.13: 45341281-45341281 on Build GRCh38
rs758804765 Chr.13: 45341579-45341579 on Build GRCh38
rs779689826 Chr.13: 45341577-45341577 on Build GRCh38
rs934619952 Chr.13: 45341652-45341652 on Build GRCh38
rs906249779 Chr.13: 45341420-45341420 on Build GRCh38
rs142653056 Chr.13: 45341322-45341322 on Build GRCh38
rs572851621 Chr.13: 45341586-45341586 on Build GRCh38
rs1024928054 Chr.13: 45341530-45341530 on Build GRCh38
rs754331759 Chr.13: 45341508-45341508 on Build GRCh38
rs911861758 Chr.13: 45341312-45341312 on Build GRCh38
rs905002087 Chr.13: 45341704-45341705 on Build GRCh38
rs144963003 Chr.13: 45341379-45341379 on Build GRCh38
rs921544043 Chr.13: 45341602-45341602 on Build GRCh38
rs760634892 Chr.13: 45341303-45341303 on Build GRCh38

View Full Product Details