Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 51649161-51649700 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FRMD6
Entrez Gene ID
122786
Gene Name
FERM domain containing 6
Gene Aliases
C14orf31, EX1, Willin, c14_5320
UniGene
Hs.434914
Gene Chromosome Location
Chr.14: 51489100-51730727 on Build GRCh38
Transcripts
NM_001042481.2 XM_011536423.1 XM_011536424.1 NR_037676.1

Gene Symbol
FRMD6-AS1
Entrez Gene ID
145438
Gene Name
FRMD6 antisense RNA 1
Gene Aliases
C14orf82
UniGene
Hs.645410
Gene Chromosome Location
Chr.14: 51649516-51651744 on Build GRCh38
Transcripts
NM_001042481.2 XM_011536423.1 XM_011536424.1 NR_037676.1

SNP Detail

SNP ID Chromosome Location
rs995759399 Chr.14: 51649234-51649234 on Build GRCh38
rs1007254541 Chr.14: 51649574-51649574 on Build GRCh38
rs975974988 Chr.14: 51649333-51649333 on Build GRCh38
rs1013957600 Chr.14: 51649299-51649299 on Build GRCh38
rs988242221 Chr.14: 51649368-51649368 on Build GRCh38
rs186137774 Chr.14: 51649651-51649651 on Build GRCh38
rs918044713 Chr.14: 51649370-51649371 on Build GRCh38
rs866180040 Chr.14: 51649513-51649513 on Build GRCh38
rs1027287932 Chr.14: 51649270-51649270 on Build GRCh38
rs576087833 Chr.14: 51649340-51649340 on Build GRCh38
rs145608392 Chr.14: 51649345-51649345 on Build GRCh38
rs1020716682 Chr.14: 51649637-51649637 on Build GRCh38
rs905532163 Chr.14: 51649652-51649652 on Build GRCh38
rs541894673 Chr.14: 51649343-51649343 on Build GRCh38
rs563482029 Chr.14: 51649575-51649575 on Build GRCh38
rs991302175 Chr.14: 51649362-51649362 on Build GRCh38
rs146354955 Chr.14: 51649557-51649557 on Build GRCh38
rs1017377699 Chr.14: 51649314-51649314 on Build GRCh38
rs949778468 Chr.14: 51649649-51649649 on Build GRCh38
rs1020823346 Chr.14: 51649336-51649336 on Build GRCh38
rs190630913 Chr.14: 51649468-51649468 on Build GRCh38
rs1021026010 Chr.14: 51649385-51649385 on Build GRCh38
rs748510476 Chr.14: 51649334-51649334 on Build GRCh38
rs887453517 Chr.14: 51649286-51649286 on Build GRCh38
rs1037160437 Chr.14: 51649415-51649415 on Build GRCh38
rs192038204 Chr.14: 51649315-51649315 on Build GRCh38
rs1007696177 Chr.14: 51649288-51649288 on Build GRCh38
rs1037280863 Chr.14: 51649190-51649192 on Build GRCh38
rs147903936 Chr.14: 51649639-51649639 on Build GRCh38
rs184853955 Chr.14: 51649395-51649395 on Build GRCh38
rs540080101 Chr.14: 51649221-51649221 on Build GRCh38
rs577573384 Chr.14: 51649199-51649199 on Build GRCh38
rs997684627 Chr.14: 51649629-51649629 on Build GRCh38
rs11627169 Chr.14: 51649350-51649350 on Build GRCh38
rs1047514975 Chr.14: 51649237-51649237 on Build GRCh38
rs555535332 Chr.14: 51649582-51649582 on Build GRCh38
rs958200581 Chr.14: 51649358-51649358 on Build GRCh38
rs895539294 Chr.14: 51649272-51649272 on Build GRCh38
rs781701521 Chr.14: 51649225-51649225 on Build GRCh38
rs181933076 Chr.14: 51649536-51649536 on Build GRCh38
rs938049415 Chr.14: 51649359-51649359 on Build GRCh38
rs887421714 Chr.14: 51649573-51649573 on Build GRCh38
rs142369309 Chr.14: 51649440-51649440 on Build GRCh38
rs752968315 Chr.14: 51649191-51649191 on Build GRCh38
rs113604416 Chr.14: 51649534-51649534 on Build GRCh38
rs949582402 Chr.14: 51649380-51649380 on Build GRCh38

View Full Product Details