Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 25064913-25065412 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SNORD116-6
Entrez Gene ID
100033418
Gene Name
small nucleolar RNA, C/D box 116-6
Gene Aliases
HBII-85-6
UniGene
Gene Chromosome Location
Chr.15: 25065025-25065122 on Build GRCh38
Transcripts
NR_003321.1

SNP Detail

SNP ID Chromosome Location
rs568196108 Chr.15: 25065105-25065105 on Build GRCh38
rs28572178 Chr.15: 25065286-25065286 on Build GRCh38
rs546468936 Chr.15: 25065055-25065055 on Build GRCh38
rs375759144 Chr.15: 25065103-25065103 on Build GRCh38
rs777338634 Chr.15: 25064992-25064992 on Build GRCh38
rs559163448 Chr.15: 25065281-25065281 on Build GRCh38
rs746413229 Chr.15: 25065042-25065042 on Build GRCh38
rs927967992 Chr.15: 25065182-25065182 on Build GRCh38
rs574909565 Chr.15: 25065295-25065295 on Build GRCh38
rs138438418 Chr.15: 25065337-25065338 on Build GRCh38
rs973223528 Chr.15: 25065254-25065254 on Build GRCh38
rs149620745 Chr.15: 25065290-25065290 on Build GRCh38
rs961713951 Chr.15: 25064995-25064995 on Build GRCh38
rs753591911 Chr.15: 25065097-25065097 on Build GRCh38
rs767629600 Chr.15: 25065090-25065090 on Build GRCh38
rs879508347 Chr.15: 25065151-25065151 on Build GRCh38
rs564446468 Chr.15: 25065389-25065389 on Build GRCh38
rs748389144 Chr.15: 25065134-25065134 on Build GRCh38
rs758052457 Chr.15: 25065034-25065034 on Build GRCh38
rs150901171 Chr.15: 25064975-25064975 on Build GRCh38
rs766269105 Chr.15: 25065096-25065096 on Build GRCh38
rs770287118 Chr.15: 25065047-25065047 on Build GRCh38
rs991684735 Chr.15: 25065301-25065301 on Build GRCh38
rs959341776 Chr.15: 25065185-25065185 on Build GRCh38
rs528248952 Chr.15: 25065027-25065027 on Build GRCh38
rs961911259 Chr.15: 25065247-25065247 on Build GRCh38
rs779074771 Chr.15: 25065046-25065046 on Build GRCh38
rs542311453 Chr.15: 25065358-25065358 on Build GRCh38
rs746489547 Chr.15: 25065108-25065108 on Build GRCh38
rs935507836 Chr.15: 25065050-25065050 on Build GRCh38
rs528372920 Chr.15: 25065390-25065390 on Build GRCh38
rs770098360 Chr.15: 25065000-25065000 on Build GRCh38
rs563696852 Chr.15: 25065378-25065378 on Build GRCh38
rs774672159 Chr.15: 25065131-25065131 on Build GRCh38
rs745917119 Chr.15: 25065045-25065046 on Build GRCh38
rs1053936667 Chr.15: 25065082-25065082 on Build GRCh38
rs200330102 Chr.15: 25065106-25065106 on Build GRCh38
rs777388321 Chr.15: 25065035-25065035 on Build GRCh38
rs116168980 Chr.15: 25065006-25065006 on Build GRCh38
rs372095469 Chr.15: 25065099-25065099 on Build GRCh38
rs761990540 Chr.15: 25065060-25065060 on Build GRCh38
rs771583276 Chr.15: 25064989-25064989 on Build GRCh38
rs774620911 Chr.15: 25065058-25065058 on Build GRCh38
rs559836024 Chr.15: 25064960-25064960 on Build GRCh38
rs578202828 Chr.15: 25065002-25065002 on Build GRCh38
rs577456799 Chr.15: 25065289-25065289 on Build GRCh38
rs984639144 Chr.15: 25064957-25064958 on Build GRCh38
rs186873007 Chr.15: 25065237-25065237 on Build GRCh38
rs766360065 Chr.15: 25065161-25065161 on Build GRCh38
rs778313895 Chr.15: 25064983-25064983 on Build GRCh38
rs749608348 Chr.15: 25065053-25065053 on Build GRCh38
rs754986307 Chr.15: 25065159-25065161 on Build GRCh38
rs772238050 Chr.15: 25065138-25065138 on Build GRCh38
rs768301372 Chr.15: 25064980-25064980 on Build GRCh38
rs916995926 Chr.15: 25065303-25065303 on Build GRCh38
rs537351982 Chr.15: 25065270-25065270 on Build GRCh38
rs915100850 Chr.15: 25065021-25065021 on Build GRCh38
rs773166850 Chr.15: 25065139-25065139 on Build GRCh38
rs191313369 Chr.15: 25065386-25065386 on Build GRCh38
rs971497530 Chr.15: 25065329-25065329 on Build GRCh38
rs749361591 Chr.15: 25065030-25065030 on Build GRCh38
rs763107862 Chr.15: 25065003-25065003 on Build GRCh38
rs760557119 Chr.15: 25065091-25065091 on Build GRCh38
rs944486579 Chr.15: 25065379-25065379 on Build GRCh38
rs977094812 Chr.15: 25065337-25065337 on Build GRCh38
rs756054171 Chr.15: 25065012-25065012 on Build GRCh38
rs780668537 Chr.15: 25065114-25065114 on Build GRCh38
rs760119168 Chr.15: 25064993-25064993 on Build GRCh38
rs752044242 Chr.15: 25064951-25064951 on Build GRCh38
rs935542140 Chr.15: 25065359-25065359 on Build GRCh38
rs923736850 Chr.15: 25065327-25065327 on Build GRCh38
rs6576402 Chr.15: 25065293-25065293 on Build GRCh38
rs760753551 Chr.15: 25065154-25065154 on Build GRCh38
rs28505623 Chr.15: 25065126-25065126 on Build GRCh38
rs3863395 Chr.15: 25065008-25065008 on Build GRCh38
rs202138878 Chr.15: 25065024-25065024 on Build GRCh38
rs759363477 Chr.15: 25065167-25065167 on Build GRCh38
rs775750471 Chr.15: 25065052-25065052 on Build GRCh38
rs28396421 Chr.15: 25065388-25065388 on Build GRCh38
rs756590960 Chr.15: 25065109-25065109 on Build GRCh38
rs747599006 Chr.15: 25064985-25064985 on Build GRCh38
rs765174034 Chr.15: 25065171-25065171 on Build GRCh38
rs181655783 Chr.15: 25065010-25065010 on Build GRCh38
rs778730892 Chr.15: 25065166-25065166 on Build GRCh38
rs542270404 Chr.15: 25065004-25065004 on Build GRCh38
rs757384292 Chr.15: 25064982-25064986 on Build GRCh38
rs573931161 Chr.15: 25065198-25065198 on Build GRCh38
rs141858980 Chr.15: 25065254-25065254 on Build GRCh38
rs766106412 Chr.15: 25065023-25065023 on Build GRCh38

View Full Product Details