Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 25185525-25186023 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SNORD115-9
Entrez Gene ID
100033446
Gene Name
small nucleolar RNA, C/D box 115-9
Gene Aliases
HBII-52-9
UniGene
Gene Chromosome Location
Chr.15: 25185631-25185712 on Build GRCh38
Transcripts
NR_003301.1

SNP Detail

SNP ID Chromosome Location
rs555548458 Chr.15: 25185752-25185752 on Build GRCh38
rs770867890 Chr.15: 25185822-25185822 on Build GRCh38
rs977770774 Chr.15: 25185582-25185582 on Build GRCh38
rs760162189 Chr.15: 25185687-25185687 on Build GRCh38
rs954453039 Chr.15: 25185988-25185988 on Build GRCh38
rs183959995 Chr.15: 25185682-25185682 on Build GRCh38
rs763161059 Chr.15: 25185691-25185691 on Build GRCh38
rs985605632 Chr.15: 25185557-25185557 on Build GRCh38
rs753769093 Chr.15: 25185808-25185808 on Build GRCh38
rs954284005 Chr.15: 25185819-25185819 on Build GRCh38
rs901901610 Chr.15: 25185949-25185949 on Build GRCh38
rs957690504 Chr.15: 25185794-25185794 on Build GRCh38
rs987055767 Chr.15: 25185990-25185990 on Build GRCh38
rs570216647 Chr.15: 25185842-25185842 on Build GRCh38
rs761174076 Chr.15: 25185613-25185613 on Build GRCh38
rs767086418 Chr.15: 25185675-25185675 on Build GRCh38
rs898487774 Chr.15: 25185696-25185696 on Build GRCh38
rs111899633 Chr.15: 25185937-25185937 on Build GRCh38
rs35974003 Chr.15: 25185614-25185615 on Build GRCh38
rs779912124 Chr.15: 25185712-25185712 on Build GRCh38
rs768372767 Chr.15: 25185669-25185669 on Build GRCh38
rs750517211 Chr.15: 25185758-25185758 on Build GRCh38
rs940082177 Chr.15: 25185674-25185674 on Build GRCh38
rs756104902 Chr.15: 25185710-25185710 on Build GRCh38
rs912638942 Chr.15: 25185661-25185661 on Build GRCh38
rs781125054 Chr.15: 25185640-25185640 on Build GRCh38
rs1029003562 Chr.15: 25185798-25185798 on Build GRCh38
rs556285574 Chr.15: 25185800-25185800 on Build GRCh38
rs35997926 Chr.15: 25185750-25185750 on Build GRCh38
rs545010225 Chr.15: 25185910-25185910 on Build GRCh38
rs905728209 Chr.15: 25185944-25185944 on Build GRCh38
rs763394182 Chr.15: 25185731-25185731 on Build GRCh38
rs974770204 Chr.15: 25185768-25185768 on Build GRCh38
rs745926091 Chr.15: 25185579-25185579 on Build GRCh38
rs771820573 Chr.15: 25185719-25185719 on Build GRCh38
rs754357369 Chr.15: 25185616-25185616 on Build GRCh38
rs751918221 Chr.15: 25185745-25185745 on Build GRCh38
rs569856580 Chr.15: 25185642-25185642 on Build GRCh38
rs919965530 Chr.15: 25185880-25185880 on Build GRCh38
rs180886631 Chr.15: 25185627-25185627 on Build GRCh38
rs1025136045 Chr.15: 25185567-25185567 on Build GRCh38
rs766917180 Chr.15: 25185614-25185614 on Build GRCh38
rs893529160 Chr.15: 25185981-25185981 on Build GRCh38
rs747804703 Chr.15: 25185718-25185718 on Build GRCh38
rs757240325 Chr.15: 25185639-25185639 on Build GRCh38
rs772913288 Chr.15: 25185724-25185724 on Build GRCh38
rs940136393 Chr.15: 25185855-25185855 on Build GRCh38
rs112940749 Chr.15: 25185982-25185982 on Build GRCh38
rs768789978 Chr.15: 25185604-25185605 on Build GRCh38
rs553128785 Chr.15: 25185654-25185654 on Build GRCh38
rs750494624 Chr.15: 25185706-25185706 on Build GRCh38
rs529434369 Chr.15: 25185976-25185976 on Build GRCh38
rs1014069432 Chr.15: 25185695-25185695 on Build GRCh38
rs370424025 Chr.15: 25185598-25185598 on Build GRCh38
rs777799811 Chr.15: 25185633-25185633 on Build GRCh38
rs753117388 Chr.15: 25185690-25185690 on Build GRCh38
rs1019763232 Chr.15: 25185998-25185998 on Build GRCh38
rs200037448 Chr.15: 25185622-25185622 on Build GRCh38
rs764410644 Chr.15: 25185736-25185736 on Build GRCh38
rs773811599 Chr.15: 25185670-25185670 on Build GRCh38
rs113354812 Chr.15: 25185772-25185772 on Build GRCh38
rs760203014 Chr.15: 25185725-25185725 on Build GRCh38
rs1028632822 Chr.15: 25185984-25185984 on Build GRCh38
rs912698747 Chr.15: 25185844-25185844 on Build GRCh38
rs749117377 Chr.15: 25185714-25185714 on Build GRCh38
rs200831581 Chr.15: 25185636-25185636 on Build GRCh38
rs990452041 Chr.15: 25185795-25185795 on Build GRCh38
rs955474246 Chr.15: 25185742-25185742 on Build GRCh38
rs188638335 Chr.15: 25185702-25185702 on Build GRCh38
rs1054809346 Chr.15: 25185973-25185973 on Build GRCh38
rs562623248 Chr.15: 25185991-25185991 on Build GRCh38
rs533814336 Chr.15: 25185716-25185716 on Build GRCh38
rs540522769 Chr.15: 25185945-25185945 on Build GRCh38
rs1037337320 Chr.15: 25185612-25185612 on Build GRCh38
rs561990027 Chr.15: 25185974-25185974 on Build GRCh38
rs535117184 Chr.15: 25185968-25185969 on Build GRCh38
rs201480811 Chr.15: 25185730-25185730 on Build GRCh38
rs775332931 Chr.15: 25185655-25185655 on Build GRCh38
rs895057659 Chr.15: 25185919-25185919 on Build GRCh38
rs537986590 Chr.15: 25185784-25185784 on Build GRCh38
rs745852864 Chr.15: 25185648-25185648 on Build GRCh38
rs972771838 Chr.15: 25185870-25185870 on Build GRCh38
rs752800890 Chr.15: 25185619-25185619 on Build GRCh38
rs568949189 Chr.15: 25185668-25185668 on Build GRCh38
rs770428640 Chr.15: 25185728-25185728 on Build GRCh38
rs567422675 Chr.15: 25185762-25185762 on Build GRCh38
rs773971560 Chr.15: 25185606-25185606 on Build GRCh38
rs916980879 Chr.15: 25185893-25185893 on Build GRCh38
rs566783533 Chr.15: 25185703-25185703 on Build GRCh38
rs931304897 Chr.15: 25185883-25185883 on Build GRCh38
rs111273383 Chr.15: 25185936-25185936 on Build GRCh38
rs1047537901 Chr.15: 25185689-25185689 on Build GRCh38
rs577741691 Chr.15: 25185901-25185901 on Build GRCh38
rs749025458 Chr.15: 25185665-25185665 on Build GRCh38
rs966411145 Chr.15: 25185576-25185576 on Build GRCh38
rs765825374 Chr.15: 25185688-25185688 on Build GRCh38
rs371309873 Chr.15: 25185761-25185761 on Build GRCh38
rs1055099384 Chr.15: 25185906-25185906 on Build GRCh38
rs539807791 Chr.15: 25185672-25185672 on Build GRCh38
rs776951543 Chr.15: 25185968-25185968 on Build GRCh38
rs28408782 Chr.15: 25185588-25185588 on Build GRCh38
rs905744113 Chr.15: 25185705-25185705 on Build GRCh38
rs1032035257 Chr.15: 25185778-25185778 on Build GRCh38
rs772828665 Chr.15: 25185676-25185676 on Build GRCh38
rs367575748 Chr.15: 25185934-25185934 on Build GRCh38
rs778760123 Chr.15: 25185926-25185926 on Build GRCh38
rs756215856 Chr.15: 25185693-25185693 on Build GRCh38

View Full Product Details