Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 81179115-81179608 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AATK-AS1
Entrez Gene ID
388428
Gene Name
AATK antisense RNA 1
Gene Aliases
-
UniGene
Hs.591202
Gene Chromosome Location
Chr.17: 81165507-81183164 on Build GRCh38
Transcripts
NR_027255.1

SNP Detail

SNP ID Chromosome Location
rs1014913932 Chr.17: 81179587-81179587 on Build GRCh38
rs528739531 Chr.17: 81179480-81179480 on Build GRCh38
rs1006710619 Chr.17: 81179384-81179384 on Build GRCh38
rs760411121 Chr.17: 81179357-81179357 on Build GRCh38
rs542851135 Chr.17: 81179459-81179459 on Build GRCh38
rs369456061 Chr.17: 81179172-81179172 on Build GRCh38
rs891204366 Chr.17: 81179519-81179519 on Build GRCh38
rs540944480 Chr.17: 81179169-81179169 on Build GRCh38
rs776456348 Chr.17: 81179170-81179170 on Build GRCh38
rs1051204618 Chr.17: 81179487-81179487 on Build GRCh38
rs768047088 Chr.17: 81179194-81179194 on Build GRCh38
rs975954497 Chr.17: 81179415-81179415 on Build GRCh38
rs756996795 Chr.17: 81179473-81179473 on Build GRCh38
rs1029746115 Chr.17: 81179364-81179364 on Build GRCh38
rs979209542 Chr.17: 81179445-81179445 on Build GRCh38
rs967109017 Chr.17: 81179571-81179571 on Build GRCh38
rs139748079 Chr.17: 81179446-81179446 on Build GRCh38
rs1018096992 Chr.17: 81179556-81179556 on Build GRCh38
rs905994469 Chr.17: 81179359-81179359 on Build GRCh38
rs929966165 Chr.17: 81179481-81179481 on Build GRCh38
rs536834800 Chr.17: 81179341-81179341 on Build GRCh38
rs903528528 Chr.17: 81179293-81179293 on Build GRCh38
rs955231453 Chr.17: 81179379-81179379 on Build GRCh38
rs1005981512 Chr.17: 81179279-81179279 on Build GRCh38
rs538715071 Chr.17: 81179212-81179212 on Build GRCh38
rs1043819997 Chr.17: 81179276-81179276 on Build GRCh38
rs981246918 Chr.17: 81179418-81179418 on Build GRCh38
rs754060051 Chr.17: 81179429-81179429 on Build GRCh38
rs569545087 Chr.17: 81179329-81179329 on Build GRCh38
rs1014854301 Chr.17: 81179411-81179411 on Build GRCh38
rs939543278 Chr.17: 81179438-81179438 on Build GRCh38
rs975882799 Chr.17: 81179239-81179239 on Build GRCh38
rs8076007 Chr.17: 81179414-81179414 on Build GRCh38
rs376204437 Chr.17: 81179358-81179358 on Build GRCh38
rs911140221 Chr.17: 81179141-81179141 on Build GRCh38
rs950300358 Chr.17: 81179510-81179510 on Build GRCh38
rs762385501 Chr.17: 81179191-81179191 on Build GRCh38
rs866081943 Chr.17: 81179559-81179559 on Build GRCh38
rs569693949 Chr.17: 81179452-81179452 on Build GRCh38
rs895381561 Chr.17: 81179274-81179274 on Build GRCh38
rs765235066 Chr.17: 81179565-81179565 on Build GRCh38
rs957050435 Chr.17: 81179150-81179150 on Build GRCh38
rs994231333 Chr.17: 81179363-81179363 on Build GRCh38
rs921706905 Chr.17: 81179249-81179249 on Build GRCh38
rs143461333 Chr.17: 81179406-81179407 on Build GRCh38
rs565108877 Chr.17: 81179563-81179563 on Build GRCh38
rs934952813 Chr.17: 81179332-81179332 on Build GRCh38
rs775815986 Chr.17: 81179197-81179197 on Build GRCh38
rs117897794 Chr.17: 81179203-81179203 on Build GRCh38
rs146378992 Chr.17: 81179413-81179413 on Build GRCh38
rs1045910136 Chr.17: 81179538-81179538 on Build GRCh38
rs747824832 Chr.17: 81179171-81179171 on Build GRCh38
rs534612135 Chr.17: 81179397-81179397 on Build GRCh38
rs1001619187 Chr.17: 81179386-81179386 on Build GRCh38
rs373952909 Chr.17: 81179190-81179190 on Build GRCh38
rs117293007 Chr.17: 81179183-81179183 on Build GRCh38
rs892020294 Chr.17: 81179350-81179350 on Build GRCh38
rs111464682 Chr.17: 81179551-81179551 on Build GRCh38
rs6565543 Chr.17: 81179401-81179401 on Build GRCh38
rs538067869 Chr.17: 81179586-81179586 on Build GRCh38
rs370251767 Chr.17: 81179389-81179389 on Build GRCh38
rs183033350 Chr.17: 81179458-81179458 on Build GRCh38
rs113394320 Chr.17: 81179259-81179259 on Build GRCh38
rs7223661 Chr.17: 81179464-81179464 on Build GRCh38
rs567526112 Chr.17: 81179416-81179416 on Build GRCh38

View Full Product Details