Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 64213996-64214503 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LINC01538
Entrez Gene ID
400654
Gene Name
long intergenic non-protein coding RNA 1538
Gene Aliases
-
UniGene
Hs.434235
Gene Chromosome Location
Chr.18: 64213082-64260055 on Build GRCh38
Transcripts
NR_033983.1 NR_033881.1

Gene Symbol
LOC284294
Entrez Gene ID
284294
Gene Name
uncharacterized LOC284294
Gene Aliases
-
UniGene
Hs.385642
Gene Chromosome Location
Chr.18: 64104091-64423601 on Build GRCh38
Transcripts
NR_033983.1 NR_033881.1

SNP Detail

SNP ID Chromosome Location
rs555142189 Chr.18: 64214182-64214182 on Build GRCh38
rs979713879 Chr.18: 64214118-64214118 on Build GRCh38
rs866532424 Chr.18: 64214146-64214146 on Build GRCh38
rs377396113 Chr.18: 64214155-64214158 on Build GRCh38
rs993133891 Chr.18: 64214179-64214179 on Build GRCh38
rs931323271 Chr.18: 64214226-64214226 on Build GRCh38
rs569193614 Chr.18: 64214096-64214096 on Build GRCh38
rs936519985 Chr.18: 64214284-64214284 on Build GRCh38
rs542886539 Chr.18: 64214312-64214312 on Build GRCh38
rs772835624 Chr.18: 64214168-64214168 on Build GRCh38
rs76958775 Chr.18: 64214121-64214121 on Build GRCh38
rs1021785573 Chr.18: 64214101-64214101 on Build GRCh38
rs775971611 Chr.18: 64214152-64214152 on Build GRCh38
rs992791320 Chr.18: 64214480-64214480 on Build GRCh38
rs1010781010 Chr.18: 64214320-64214320 on Build GRCh38
rs907739832 Chr.18: 64214185-64214185 on Build GRCh38
rs770653009 Chr.18: 64214450-64214450 on Build GRCh38
rs891998040 Chr.18: 64214318-64214318 on Build GRCh38
rs968409020 Chr.18: 64214394-64214394 on Build GRCh38
rs1021392228 Chr.18: 64214342-64214342 on Build GRCh38
rs182223440 Chr.18: 64214061-64214061 on Build GRCh38
rs749798163 Chr.18: 64214108-64214108 on Build GRCh38
rs972224518 Chr.18: 64214459-64214459 on Build GRCh38
rs992846595 Chr.18: 64214161-64214161 on Build GRCh38
rs938628892 Chr.18: 64214141-64214141 on Build GRCh38
rs1037125059 Chr.18: 64214218-64214218 on Build GRCh38
rs919356826 Chr.18: 64214147-64214147 on Build GRCh38
rs140080183 Chr.18: 64214288-64214288 on Build GRCh38
rs915864316 Chr.18: 64214192-64214192 on Build GRCh38
rs190130061 Chr.18: 64214445-64214445 on Build GRCh38
rs898537704 Chr.18: 64214225-64214225 on Build GRCh38
rs892216407 Chr.18: 64214310-64214310 on Build GRCh38
rs185611120 Chr.18: 64214257-64214257 on Build GRCh38
rs764675833 Chr.18: 64214156-64214156 on Build GRCh38
rs151177277 Chr.18: 64214234-64214234 on Build GRCh38
rs1045045975 Chr.18: 64214244-64214244 on Build GRCh38
rs903756923 Chr.18: 64214262-64214262 on Build GRCh38
rs142114118 Chr.18: 64214196-64214196 on Build GRCh38
rs953659823 Chr.18: 64214091-64214091 on Build GRCh38
rs1049730758 Chr.18: 64214229-64214229 on Build GRCh38
rs531631711 Chr.18: 64214397-64214397 on Build GRCh38
rs888818120 Chr.18: 64214391-64214391 on Build GRCh38
rs562797035 Chr.18: 64214339-64214339 on Build GRCh38
rs545797769 Chr.18: 64214334-64214334 on Build GRCh38
rs75793171 Chr.18: 64214477-64214477 on Build GRCh38

View Full Product Details