Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 2462840-2463343 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SNORD119
Entrez Gene ID
100113378
Gene Name
small nucleolar RNA, C/D box 119
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.20: 2462952-2463047 on Build GRCh38
Transcripts
NR_003684.1 NM_003091.3 NM_198216.1

Gene Symbol
SNRPB
Entrez Gene ID
6628
Gene Name
small nuclear ribonucleoprotein polypeptides B and B1
Gene Aliases
CCMS, COD, SNRPB1, Sm-B/B', SmB/B', SmB/SmB', snRNP-B
UniGene
Hs.83753
Gene Chromosome Location
Chr.20: 2461635-2470853 on Build GRCh38
Transcripts
NR_003684.1 NM_003091.3 NM_198216.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1227013 c.457G>A p.A153T Chr.20: 2463191-2463191 on Build GRCh38
5109366 c.525T>A p.P175P Chr.20: 2463123-2463123 on Build GRCh38
3404983 c.515G>A p.R172H Chr.20: 2463133-2463133 on Build GRCh38
6408577 c.541C>T p.R181* Chr.20: 2463107-2463107 on Build GRCh38
7199248 c.457G>C p.A153P Chr.20: 2463191-2463191 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770865345 Chr.20: 2463200-2463200 on Build GRCh38
rs953532073 Chr.20: 2462959-2462959 on Build GRCh38
rs1054767222 Chr.20: 2463300-2463300 on Build GRCh38
rs759310762 Chr.20: 2463235-2463235 on Build GRCh38
rs747470682 Chr.20: 2463059-2463059 on Build GRCh38
rs373871174 Chr.20: 2463122-2463122 on Build GRCh38
rs770099491 Chr.20: 2463081-2463081 on Build GRCh38
rs903609200 Chr.20: 2463185-2463185 on Build GRCh38
rs941188391 Chr.20: 2463039-2463039 on Build GRCh38
rs201974960 Chr.20: 2463043-2463043 on Build GRCh38
rs763535486 Chr.20: 2463253-2463253 on Build GRCh38
rs758950973 Chr.20: 2463045-2463045 on Build GRCh38
rs751038700 Chr.20: 2463261-2463261 on Build GRCh38
rs11545674 Chr.20: 2463132-2463132 on Build GRCh38
rs781713014 Chr.20: 2463162-2463162 on Build GRCh38
rs772106010 Chr.20: 2463095-2463095 on Build GRCh38
rs776456665 Chr.20: 2463225-2463225 on Build GRCh38
rs956944780 Chr.20: 2462910-2462910 on Build GRCh38
rs896137389 Chr.20: 2463084-2463084 on Build GRCh38
rs761749996 Chr.20: 2463240-2463242 on Build GRCh38
rs540627358 Chr.20: 2462917-2462917 on Build GRCh38
rs749358211 Chr.20: 2463094-2463094 on Build GRCh38
rs147230511 Chr.20: 2463098-2463098 on Build GRCh38
rs759220904 Chr.20: 2463118-2463118 on Build GRCh38
rs201164290 Chr.20: 2463237-2463237 on Build GRCh38
rs997754086 Chr.20: 2463317-2463317 on Build GRCh38
rs775753327 Chr.20: 2463092-2463092 on Build GRCh38
rs750787684 Chr.20: 2463176-2463176 on Build GRCh38
rs769498639 Chr.20: 2463242-2463242 on Build GRCh38
rs752259594 Chr.20: 2463121-2463121 on Build GRCh38
rs752021176 Chr.20: 2463135-2463135 on Build GRCh38
rs376318628 Chr.20: 2463191-2463191 on Build GRCh38
rs73571231 Chr.20: 2462956-2462956 on Build GRCh38
rs774021768 Chr.20: 2463183-2463184 on Build GRCh38
rs756455021 Chr.20: 2463184-2463184 on Build GRCh38
rs746112809 Chr.20: 2463077-2463077 on Build GRCh38
rs529593266 Chr.20: 2462958-2462958 on Build GRCh38
rs746944646 Chr.20: 2463196-2463196 on Build GRCh38
rs1031226295 Chr.20: 2462866-2462866 on Build GRCh38
rs34216892 Chr.20: 2463122-2463123 on Build GRCh38
rs773019859 Chr.20: 2463099-2463099 on Build GRCh38
rs1031192223 Chr.20: 2463201-2463201 on Build GRCh38
rs140604228 Chr.20: 2463192-2463192 on Build GRCh38
rs778125656 Chr.20: 2463049-2463049 on Build GRCh38
rs760597171 Chr.20: 2463102-2463102 on Build GRCh38
rs909324612 Chr.20: 2463006-2463006 on Build GRCh38
rs755337256 Chr.20: 2462989-2462989 on Build GRCh38
rs1030411450 Chr.20: 2462972-2462972 on Build GRCh38
rs776364274 Chr.20: 2463116-2463116 on Build GRCh38
rs770650165 Chr.20: 2463113-2463113 on Build GRCh38
rs781561378 Chr.20: 2463065-2463065 on Build GRCh38
rs767377277 Chr.20: 2463243-2463246 on Build GRCh38
rs376998028 Chr.20: 2463133-2463133 on Build GRCh38
rs892693077 Chr.20: 2463311-2463311 on Build GRCh38
rs775123917 Chr.20: 2463249-2463249 on Build GRCh38
rs762265524 Chr.20: 2463273-2463273 on Build GRCh38
rs141971905 Chr.20: 2463044-2463044 on Build GRCh38
rs762607869 Chr.20: 2463251-2463251 on Build GRCh38
rs986212609 Chr.20: 2463003-2463003 on Build GRCh38
rs929910057 Chr.20: 2463079-2463079 on Build GRCh38
rs780410112 Chr.20: 2463187-2463187 on Build GRCh38
rs113467048 Chr.20: 2463134-2463134 on Build GRCh38
rs370050698 Chr.20: 2463161-2463161 on Build GRCh38
rs569470508 Chr.20: 2463193-2463193 on Build GRCh38
rs757566588 Chr.20: 2463060-2463060 on Build GRCh38

View Full Product Details