Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 44518888-44519380 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TSPEAR
Entrez Gene ID
54084
Gene Name
thrombospondin type laminin G domain and EAR repeats
Gene Aliases
C21orf29, DFNB98, TSP-EAR
UniGene
Hs.660703
Gene Chromosome Location
Chr.21: 44497892-44711580 on Build GRCh38
Transcripts
NM_001272037.1 NM_144991.2 NR_026547.1 NR_026548.1

Gene Symbol
TSPEAR-AS2
Entrez Gene ID
114043
Gene Name
TSPEAR antisense RNA 2
Gene Aliases
C21orf90
UniGene
Hs.592163
Gene Chromosome Location
Chr.21: 44517215-44518977 on Build GRCh38
Transcripts
NM_001272037.1 NM_144991.2 NR_026547.1 NR_026548.1

SNP Detail

SNP ID Chromosome Location
rs587740038 Chr.21: 44518998-44518998 on Build GRCh38
rs893753391 Chr.21: 44519351-44519351 on Build GRCh38
rs139395652 Chr.21: 44519090-44519090 on Build GRCh38
rs937617101 Chr.21: 44519122-44519122 on Build GRCh38
rs1036134638 Chr.21: 44519148-44519148 on Build GRCh38
rs1036970150 Chr.21: 44519287-44519287 on Build GRCh38
rs941734615 Chr.21: 44519145-44519145 on Build GRCh38
rs957416437 Chr.21: 44519069-44519069 on Build GRCh38
rs587605320 Chr.21: 44519089-44519089 on Build GRCh38
rs1030233627 Chr.21: 44519027-44519027 on Build GRCh38
rs1005978978 Chr.21: 44518997-44518997 on Build GRCh38
rs183523169 Chr.21: 44519247-44519247 on Build GRCh38
rs981464977 Chr.21: 44519241-44519241 on Build GRCh38
rs587649020 Chr.21: 44519195-44519196 on Build GRCh38
rs1032137330 Chr.21: 44519068-44519068 on Build GRCh38
rs369483279 Chr.21: 44519082-44519082 on Build GRCh38
rs116840338 Chr.21: 44519360-44519360 on Build GRCh38
rs1007407849 Chr.21: 44519344-44519344 on Build GRCh38
rs981879325 Chr.21: 44519098-44519098 on Build GRCh38
rs782664092 Chr.21: 44518978-44518978 on Build GRCh38
rs967901772 Chr.21: 44519235-44519235 on Build GRCh38
rs142083045 Chr.21: 44519273-44519273 on Build GRCh38
rs587733112 Chr.21: 44519083-44519083 on Build GRCh38
rs997613214 Chr.21: 44519023-44519023 on Build GRCh38
rs957884438 Chr.21: 44519195-44519195 on Build GRCh38
rs1009395963 Chr.21: 44519358-44519358 on Build GRCh38
rs782802701 Chr.21: 44519330-44519330 on Build GRCh38
rs61574846 Chr.21: 44519243-44519243 on Build GRCh38
rs146368710 Chr.21: 44518991-44518991 on Build GRCh38
rs782172815 Chr.21: 44518933-44518933 on Build GRCh38
rs8132443 Chr.21: 44519185-44519185 on Build GRCh38
rs587613629 Chr.21: 44519173-44519173 on Build GRCh38
rs587667472 Chr.21: 44519290-44519290 on Build GRCh38
rs587643135 Chr.21: 44519186-44519186 on Build GRCh38
rs587604796 Chr.21: 44519323-44519323 on Build GRCh38
rs1055935240 Chr.21: 44519129-44519129 on Build GRCh38
rs587705033 Chr.21: 44519189-44519189 on Build GRCh38
rs190947881 Chr.21: 44519115-44519115 on Build GRCh38
rs1007343999 Chr.21: 44519035-44519035 on Build GRCh38
rs1021247534 Chr.21: 44519359-44519359 on Build GRCh38
rs8128835 Chr.21: 44519222-44519222 on Build GRCh38
rs150062512 Chr.21: 44519166-44519166 on Build GRCh38
rs587610705 Chr.21: 44519284-44519284 on Build GRCh38
rs782543820 Chr.21: 44519075-44519075 on Build GRCh38
rs587705738 Chr.21: 44519274-44519274 on Build GRCh38
rs587668088 Chr.21: 44519060-44519060 on Build GRCh38
rs997181731 Chr.21: 44519164-44519164 on Build GRCh38
rs587622442 Chr.21: 44519092-44519092 on Build GRCh38
rs1046000204 Chr.21: 44518925-44518925 on Build GRCh38
rs186183912 Chr.21: 44519091-44519091 on Build GRCh38
rs147598035 Chr.21: 44519264-44519264 on Build GRCh38
rs60390607 Chr.21: 44519174-44519174 on Build GRCh38
rs897596250 Chr.21: 44519154-44519154 on Build GRCh38
rs587598870 Chr.21: 44519008-44519008 on Build GRCh38
rs909389547 Chr.21: 44519130-44519130 on Build GRCh38
rs953327936 Chr.21: 44519030-44519030 on Build GRCh38
rs143833189 Chr.21: 44518950-44518950 on Build GRCh38
rs1003378572 Chr.21: 44519361-44519361 on Build GRCh38
rs587623594 Chr.21: 44518957-44518957 on Build GRCh38
rs8132614 Chr.21: 44519311-44519311 on Build GRCh38
rs1009838953 Chr.21: 44519223-44519223 on Build GRCh38
rs990262760 Chr.21: 44519085-44519085 on Build GRCh38
rs386819160 Chr.21: 44519185-44519187 on Build GRCh38
rs933066422 Chr.21: 44519313-44519313 on Build GRCh38
rs1056008706 Chr.21: 44518990-44518990 on Build GRCh38
rs782075054 Chr.21: 44519233-44519233 on Build GRCh38
rs1053612558 Chr.21: 44519349-44519349 on Build GRCh38

View Full Product Details