Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 45830354-45830848 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC100129027
Entrez Gene ID
100129027
Gene Name
uncharacterized LOC100129027
Gene Aliases
-
UniGene
Hs.689591
Gene Chromosome Location
Chr.21: 45827841-45836419 on Build GRCh38
Transcripts
NR_038876.1

SNP Detail

SNP ID Chromosome Location
rs984677204 Chr.21: 45830407-45830407 on Build GRCh38
rs1002302808 Chr.21: 45830578-45830578 on Build GRCh38
rs934504864 Chr.21: 45830429-45830429 on Build GRCh38
rs574910311 Chr.21: 45830675-45830675 on Build GRCh38
rs950672827 Chr.21: 45830677-45830677 on Build GRCh38
rs755076576 Chr.21: 45830434-45830436 on Build GRCh38
rs777879135 Chr.21: 45830773-45830774 on Build GRCh38
rs531345602 Chr.21: 45830481-45830481 on Build GRCh38
rs929214364 Chr.21: 45830406-45830406 on Build GRCh38
rs945650659 Chr.21: 45830819-45830819 on Build GRCh38
rs376541086 Chr.21: 45830510-45830510 on Build GRCh38
rs1044346046 Chr.21: 45830499-45830499 on Build GRCh38
rs79344857 Chr.21: 45830737-45830737 on Build GRCh38
rs556577595 Chr.21: 45830659-45830659 on Build GRCh38
rs386819366 Chr.21: 45830675-45830676 on Build GRCh38
rs369621910 Chr.21: 45830680-45830680 on Build GRCh38
rs376934348 Chr.21: 45830552-45830552 on Build GRCh38
rs576066055 Chr.21: 45830786-45830786 on Build GRCh38
rs1028101936 Chr.21: 45830603-45830603 on Build GRCh38
rs557715155 Chr.21: 45830620-45830620 on Build GRCh38
rs960763669 Chr.21: 45830671-45830671 on Build GRCh38
rs549350794 Chr.21: 45830483-45830483 on Build GRCh38
rs943175975 Chr.21: 45830697-45830697 on Build GRCh38
rs190637800 Chr.21: 45830474-45830474 on Build GRCh38
rs962427500 Chr.21: 45830556-45830556 on Build GRCh38
rs1026563060 Chr.21: 45830523-45830523 on Build GRCh38
rs1006358300 Chr.21: 45830531-45830531 on Build GRCh38
rs115793986 Chr.21: 45830380-45830380 on Build GRCh38
rs1054225369 Chr.21: 45830463-45830463 on Build GRCh38
rs570917735 Chr.21: 45830640-45830640 on Build GRCh38
rs11700736 Chr.21: 45830529-45830529 on Build GRCh38
rs909066878 Chr.21: 45830693-45830693 on Build GRCh38
rs989917949 Chr.21: 45830767-45830767 on Build GRCh38
rs527893693 Chr.21: 45830409-45830409 on Build GRCh38
rs545530811 Chr.21: 45830467-45830467 on Build GRCh38
rs564980133 Chr.21: 45830501-45830501 on Build GRCh38
rs1033793049 Chr.21: 45830664-45830664 on Build GRCh38
rs564558793 Chr.21: 45830765-45830765 on Build GRCh38
rs572693554 Chr.21: 45830736-45830736 on Build GRCh38
rs771222236 Chr.21: 45830408-45830408 on Build GRCh38
rs1052822024 Chr.21: 45830742-45830742 on Build GRCh38
rs538379413 Chr.21: 45830658-45830658 on Build GRCh38
rs542499345 Chr.21: 45830676-45830676 on Build GRCh38
rs575250527 Chr.21: 45830749-45830749 on Build GRCh38
rs770322363 Chr.21: 45830573-45830573 on Build GRCh38
rs919205528 Chr.21: 45830383-45830383 on Build GRCh38
rs567745264 Chr.21: 45830500-45830500 on Build GRCh38
rs756313403 Chr.21: 45830604-45830605 on Build GRCh38

View Full Product Details