Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 45836175-45836678 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC100129027
Entrez Gene ID
100129027
Gene Name
uncharacterized LOC100129027
Gene Aliases
-
UniGene
Hs.689591
Gene Chromosome Location
Chr.21: 45827841-45836419 on Build GRCh38
Transcripts
NR_038876.1

SNP Detail

SNP ID Chromosome Location
rs1030106110 Chr.21: 45836203-45836203 on Build GRCh38
rs150760819 Chr.21: 45836233-45836233 on Build GRCh38
rs73380636 Chr.21: 45836546-45836546 on Build GRCh38
rs13047964 Chr.21: 45836458-45836458 on Build GRCh38
rs188604337 Chr.21: 45836273-45836273 on Build GRCh38
rs963735230 Chr.21: 45836341-45836341 on Build GRCh38
rs13050816 Chr.21: 45836501-45836501 on Build GRCh38
rs1029205639 Chr.21: 45836638-45836638 on Build GRCh38
rs555309649 Chr.21: 45836456-45836456 on Build GRCh38
rs968401985 Chr.21: 45836469-45836469 on Build GRCh38
rs545200005 Chr.21: 45836578-45836578 on Build GRCh38
rs1029694671 Chr.21: 45836379-45836379 on Build GRCh38
rs1043390404 Chr.21: 45836264-45836264 on Build GRCh38
rs73380634 Chr.21: 45836401-45836401 on Build GRCh38
rs13047976 Chr.21: 45836486-45836486 on Build GRCh38
rs895680752 Chr.21: 45836548-45836548 on Build GRCh38
rs899158082 Chr.21: 45836564-45836564 on Build GRCh38
rs949439778 Chr.21: 45836345-45836345 on Build GRCh38
rs1047725576 Chr.21: 45836358-45836358 on Build GRCh38
rs777063129 Chr.21: 45836327-45836327 on Build GRCh38
rs904268642 Chr.21: 45836643-45836643 on Build GRCh38
rs748374203 Chr.21: 45836448-45836448 on Build GRCh38
rs768755847 Chr.21: 45836637-45836637 on Build GRCh38
rs773295427 Chr.21: 45836507-45836507 on Build GRCh38
rs931858967 Chr.21: 45836595-45836595 on Build GRCh38
rs905782165 Chr.21: 45836528-45836528 on Build GRCh38
rs114444961 Chr.21: 45836559-45836559 on Build GRCh38
rs1042970428 Chr.21: 45836527-45836527 on Build GRCh38
rs573565968 Chr.21: 45836513-45836513 on Build GRCh38
rs1025061130 Chr.21: 45836514-45836514 on Build GRCh38
rs539635392 Chr.21: 45836329-45836329 on Build GRCh38
rs987600808 Chr.21: 45836415-45836415 on Build GRCh38
rs1037654223 Chr.21: 45836418-45836418 on Build GRCh38
rs907954691 Chr.21: 45836529-45836529 on Build GRCh38
rs1011479110 Chr.21: 45836236-45836236 on Build GRCh38
rs559386068 Chr.21: 45836557-45836557 on Build GRCh38
rs747166566 Chr.21: 45836633-45836633 on Build GRCh38
rs557964202 Chr.21: 45836396-45836396 on Build GRCh38
rs572896232 Chr.21: 45836490-45836490 on Build GRCh38
rs543135630 Chr.21: 45836453-45836453 on Build GRCh38
rs371338899 Chr.21: 45836508-45836508 on Build GRCh38
rs775702469 Chr.21: 45836267-45836267 on Build GRCh38
rs890263879 Chr.21: 45836626-45836626 on Build GRCh38
rs900399381 Chr.21: 45836419-45836419 on Build GRCh38
rs921906870 Chr.21: 45836594-45836594 on Build GRCh38
rs1051621965 Chr.21: 45836613-45836613 on Build GRCh38
rs76120661 Chr.21: 45836210-45836210 on Build GRCh38
rs912287772 Chr.21: 45836449-45836449 on Build GRCh38
rs376240899 Chr.21: 45836549-45836549 on Build GRCh38
rs937135866 Chr.21: 45836530-45836530 on Build GRCh38
rs1004858629 Chr.21: 45836323-45836323 on Build GRCh38
rs769815531 Chr.21: 45836506-45836506 on Build GRCh38
rs907859530 Chr.21: 45836368-45836368 on Build GRCh38
rs895922916 Chr.21: 45836314-45836314 on Build GRCh38
rs566408342 Chr.21: 45836287-45836287 on Build GRCh38
rs1001401943 Chr.21: 45836274-45836274 on Build GRCh38
rs979678676 Chr.21: 45836320-45836320 on Build GRCh38
rs995255842 Chr.21: 45836598-45836598 on Build GRCh38
rs973680670 Chr.21: 45836355-45836355 on Build GRCh38
rs991390866 Chr.21: 45836340-45836341 on Build GRCh38
rs563505934 Chr.21: 45836640-45836640 on Build GRCh38

View Full Product Details