Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 21850306-21850849 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNIP4
Entrez Gene ID
80333
Gene Name
potassium voltage-gated channel interacting protein 4
Gene Aliases
CALP, KCHIP4
UniGene
Hs.655705
Gene Chromosome Location
Chr.4: 20728520-21948801 on Build GRCh38
Transcripts
NM_025221.5 NM_147181.3 NM_147182.3 XM_011513882.2 XM_011513886.2 NR_002813.1

Gene Symbol
KCNIP4-IT1
Entrez Gene ID
359822
Gene Name
KCNIP4 intronic transcript 1
Gene Aliases
NCRNA00099, UM9(5), UM9-5
UniGene
Hs.354058
Gene Chromosome Location
Chr.4: 21843341-21853188 on Build GRCh38
Transcripts
NM_025221.5 NM_147181.3 NM_147182.3 XM_011513882.2 XM_011513886.2 NR_002813.1

SNP Detail

SNP ID Chromosome Location
rs934598749 Chr.4: 21850354-21850354 on Build GRCh38
rs892009678 Chr.4: 21850622-21850622 on Build GRCh38
rs948591711 Chr.4: 21850542-21850542 on Build GRCh38
rs531695392 Chr.4: 21850429-21850429 on Build GRCh38
rs948539759 Chr.4: 21850779-21850779 on Build GRCh38
rs541457398 Chr.4: 21850512-21850512 on Build GRCh38
rs111793805 Chr.4: 21850523-21850523 on Build GRCh38
rs964893593 Chr.4: 21850662-21850662 on Build GRCh38
rs762827743 Chr.4: 21850641-21850641 on Build GRCh38
rs966354682 Chr.4: 21850803-21850803 on Build GRCh38
rs770799636 Chr.4: 21850496-21850496 on Build GRCh38
rs146892936 Chr.4: 21850796-21850796 on Build GRCh38
rs922087597 Chr.4: 21850543-21850543 on Build GRCh38
rs2175779 Chr.4: 21850806-21850806 on Build GRCh38
rs142460042 Chr.4: 21850793-21850793 on Build GRCh38
rs150003165 Chr.4: 21850402-21850402 on Build GRCh38
rs956333624 Chr.4: 21850399-21850399 on Build GRCh38
rs548650568 Chr.4: 21850780-21850780 on Build GRCh38
rs144348745 Chr.4: 21850489-21850489 on Build GRCh38
rs550403711 Chr.4: 21850406-21850406 on Build GRCh38
rs1002806378 Chr.4: 21850575-21850575 on Build GRCh38
rs933624756 Chr.4: 21850464-21850464 on Build GRCh38
rs907322321 Chr.4: 21850568-21850568 on Build GRCh38
rs1029190232 Chr.4: 21850807-21850807 on Build GRCh38
rs919770137 Chr.4: 21850522-21850522 on Build GRCh38
rs114064407 Chr.4: 21850750-21850750 on Build GRCh38
rs1022328292 Chr.4: 21850645-21850645 on Build GRCh38
rs1004846550 Chr.4: 21850781-21850781 on Build GRCh38
rs549683814 Chr.4: 21850709-21850709 on Build GRCh38
rs1053463635 Chr.4: 21850704-21850704 on Build GRCh38
rs749884341 Chr.4: 21850629-21850629 on Build GRCh38
rs558956215 Chr.4: 21850495-21850495 on Build GRCh38
rs895654008 Chr.4: 21850392-21850392 on Build GRCh38
rs1028051308 Chr.4: 21850697-21850697 on Build GRCh38
rs536486899 Chr.4: 21850371-21850371 on Build GRCh38
rs938776575 Chr.4: 21850801-21850801 on Build GRCh38
rs530097470 Chr.4: 21850759-21850759 on Build GRCh38
rs572384500 Chr.4: 21850669-21850669 on Build GRCh38
rs545945142 Chr.4: 21850663-21850663 on Build GRCh38
rs13129819 Chr.4: 21850411-21850411 on Build GRCh38
rs148760632 Chr.4: 21850691-21850691 on Build GRCh38
rs773875448 Chr.4: 21850758-21850758 on Build GRCh38
rs13105122 Chr.4: 21850428-21850428 on Build GRCh38
rs961376121 Chr.4: 21850749-21850749 on Build GRCh38
rs1053062280 Chr.4: 21850584-21850584 on Build GRCh38
rs908203189 Chr.4: 21850739-21850739 on Build GRCh38
rs796692225 Chr.4: 21850762-21850762 on Build GRCh38
rs904188753 Chr.4: 21850352-21850352 on Build GRCh38
rs925639760 Chr.4: 21850680-21850680 on Build GRCh38
rs532612807 Chr.4: 21850707-21850707 on Build GRCh38
rs567200416 Chr.4: 21850376-21850376 on Build GRCh38
rs115317911 Chr.4: 21850545-21850545 on Build GRCh38
rs905701825 Chr.4: 21850755-21850755 on Build GRCh38
rs984136745 Chr.4: 21850485-21850485 on Build GRCh38
rs949929458 Chr.4: 21850736-21850736 on Build GRCh38
rs974905294 Chr.4: 21850518-21850518 on Build GRCh38
rs1011829130 Chr.4: 21850636-21850636 on Build GRCh38
rs572051498 Chr.4: 21850565-21850565 on Build GRCh38
rs556093637 Chr.4: 21850459-21850459 on Build GRCh38
rs561475179 Chr.4: 21850716-21850716 on Build GRCh38
rs537546979 Chr.4: 21850434-21850434 on Build GRCh38

View Full Product Details