Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 112647842-112648356 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LARP7
Entrez Gene ID
51574
Gene Name
La ribonucleoprotein domain family member 7
Gene Aliases
ALAZS, HDCMA18P, PIP7S
UniGene
Hs.713663
Gene Chromosome Location
Chr.4: 112636964-112657592 on Build GRCh38
Transcripts
NM_001267039.1 NM_015454.2 NM_016648.3 NR_049768.1 NR_029835.1 NR_029859.1 NR_029860.1

Gene Symbol
MIR302A
Entrez Gene ID
407028
Gene Name
microRNA 302a
Gene Aliases
MIRN302, MIRN302A, hsa-mir-302, mir-302a
UniGene
Gene Chromosome Location
Chr.4: 112648183-112648251 on Build GRCh38
Transcripts
NM_001267039.1 NM_015454.2 NM_016648.3 NR_049768.1 NR_029835.1 NR_029859.1 NR_029860.1

Gene Symbol
MIR302D
Entrez Gene ID
442896
Gene Name
microRNA 302d
Gene Aliases
MIRN302D, mir-302d
UniGene
Gene Chromosome Location
Chr.4: 112648004-112648071 on Build GRCh38
Transcripts
NM_001267039.1 NM_015454.2 NM_016648.3 NR_049768.1 NR_029835.1 NR_029859.1 NR_029860.1

Gene Symbol
MIR367
Entrez Gene ID
442912
Gene Name
microRNA 367
Gene Aliases
MIRN367, hsa-mir-367
UniGene
Gene Chromosome Location
Chr.4: 112647874-112647941 on Build GRCh38
Transcripts
NM_001267039.1 NM_015454.2 NM_016648.3 NR_049768.1 NR_029835.1 NR_029859.1 NR_029860.1

SNP Detail

SNP ID Chromosome Location
rs756793908 Chr.4: 112648228-112648228 on Build GRCh38
rs766032176 Chr.4: 112647868-112647870 on Build GRCh38
rs748964754 Chr.4: 112648069-112648069 on Build GRCh38
rs770044270 Chr.4: 112648183-112648183 on Build GRCh38
rs896333735 Chr.4: 112647887-112647887 on Build GRCh38
rs775257471 Chr.4: 112648081-112648081 on Build GRCh38
rs774464901 Chr.4: 112647949-112647949 on Build GRCh38
rs768129180 Chr.4: 112648226-112648226 on Build GRCh38
rs550687674 Chr.4: 112648328-112648328 on Build GRCh38
rs763883824 Chr.4: 112648245-112648247 on Build GRCh38
rs753398130 Chr.4: 112648118-112648118 on Build GRCh38
rs752031608 Chr.4: 112648114-112648114 on Build GRCh38
rs41279309 Chr.4: 112647961-112647961 on Build GRCh38
rs774234053 Chr.4: 112648010-112648010 on Build GRCh38
rs369489772 Chr.4: 112647906-112647906 on Build GRCh38
rs778682764 Chr.4: 112648149-112648149 on Build GRCh38
rs770645069 Chr.4: 112648074-112648074 on Build GRCh38
rs138673224 Chr.4: 112648051-112648051 on Build GRCh38
rs375594223 Chr.4: 112648173-112648173 on Build GRCh38
rs746561600 Chr.4: 112647918-112647918 on Build GRCh38
rs779847870 Chr.4: 112648165-112648165 on Build GRCh38
rs537751343 Chr.4: 112648323-112648323 on Build GRCh38
rs368344055 Chr.4: 112648247-112648247 on Build GRCh38
rs762822201 Chr.4: 112647938-112647938 on Build GRCh38
rs200951552 Chr.4: 112648326-112648326 on Build GRCh38
rs115879346 Chr.4: 112648075-112648075 on Build GRCh38
rs765440386 Chr.4: 112648091-112648091 on Build GRCh38
rs759501204 Chr.4: 112647950-112647950 on Build GRCh38
rs747375422 Chr.4: 112648104-112648104 on Build GRCh38
rs780422206 Chr.4: 112648060-112648061 on Build GRCh38
rs764851038 Chr.4: 112648318-112648318 on Build GRCh38
rs760260709 Chr.4: 112648234-112648234 on Build GRCh38
rs773454900 Chr.4: 112648195-112648195 on Build GRCh38
rs751041937 Chr.4: 112648267-112648268 on Build GRCh38
rs1001109052 Chr.4: 112648125-112648125 on Build GRCh38
rs574329211 Chr.4: 112648055-112648055 on Build GRCh38
rs368089282 Chr.4: 112648314-112648314 on Build GRCh38
rs768639028 Chr.4: 112648085-112648085 on Build GRCh38
rs375688232 Chr.4: 112648029-112648029 on Build GRCh38
rs757024872 Chr.4: 112648141-112648141 on Build GRCh38
rs1000958560 Chr.4: 112648113-112648113 on Build GRCh38
rs754518688 Chr.4: 112648046-112648046 on Build GRCh38
rs750063201 Chr.4: 112648232-112648232 on Build GRCh38
rs373558217 Chr.4: 112647937-112647937 on Build GRCh38
rs971111539 Chr.4: 112647967-112647967 on Build GRCh38
rs764414260 Chr.4: 112648034-112648034 on Build GRCh38
rs766226829 Chr.4: 112648137-112648138 on Build GRCh38
rs780618243 Chr.4: 112648318-112648319 on Build GRCh38
rs1014873419 Chr.4: 112648227-112648227 on Build GRCh38
rs772711592 Chr.4: 112647982-112647982 on Build GRCh38
rs757800447 Chr.4: 112648011-112648012 on Build GRCh38
rs758806646 Chr.4: 112648317-112648319 on Build GRCh38
rs557059739 Chr.4: 112648157-112648157 on Build GRCh38
rs756422508 Chr.4: 112647965-112647965 on Build GRCh38
rs750835506 Chr.4: 112648092-112648092 on Build GRCh38
rs1028787993 Chr.4: 112648274-112648275 on Build GRCh38
rs755458358 Chr.4: 112648115-112648115 on Build GRCh38
rs761859575 Chr.4: 112648176-112648176 on Build GRCh38
rs975568610 Chr.4: 112648001-112648001 on Build GRCh38
rs1020254777 Chr.4: 112647988-112647988 on Build GRCh38
rs373885472 Chr.4: 112648277-112648277 on Build GRCh38
rs564323934 Chr.4: 112648238-112648238 on Build GRCh38
rs754478462 Chr.4: 112647959-112647960 on Build GRCh38
rs757634396 Chr.4: 112648217-112648238 on Build GRCh38
rs150161032 Chr.4: 112647932-112647932 on Build GRCh38
rs769885618 Chr.4: 112648251-112648251 on Build GRCh38
rs537289389 Chr.4: 112647966-112647966 on Build GRCh38
rs774640335 Chr.4: 112648276-112648276 on Build GRCh38
rs777429513 Chr.4: 112648117-112648117 on Build GRCh38
rs772933029 Chr.4: 112648112-112648115 on Build GRCh38
rs140979090 Chr.4: 112648181-112648182 on Build GRCh38
rs768183847 Chr.4: 112647921-112647921 on Build GRCh38
rs372925614 Chr.4: 112647993-112647993 on Build GRCh38
rs760031880 Chr.4: 112648223-112648223 on Build GRCh38
rs751887605 Chr.4: 112648213-112648213 on Build GRCh38
rs752656934 Chr.4: 112647963-112647963 on Build GRCh38
rs771483662 Chr.4: 112647905-112647905 on Build GRCh38
rs769776104 Chr.4: 112648112-112648113 on Build GRCh38
rs760619273 Chr.4: 112648083-112648083 on Build GRCh38
rs763197825 Chr.4: 112648095-112648095 on Build GRCh38
rs1032493659 Chr.4: 112647945-112647945 on Build GRCh38
rs961760351 Chr.4: 112648288-112648288 on Build GRCh38
rs749880723 Chr.4: 112648319-112648319 on Build GRCh38
rs747735950 Chr.4: 112648054-112648054 on Build GRCh38
rs756236916 Chr.4: 112647870-112647870 on Build GRCh38
rs13136737 Chr.4: 112647979-112647979 on Build GRCh38
rs377427380 Chr.4: 112648053-112648053 on Build GRCh38
rs746394438 Chr.4: 112647973-112647973 on Build GRCh38
rs1006161088 Chr.4: 112648307-112648307 on Build GRCh38
rs557219694 Chr.4: 112648237-112648237 on Build GRCh38
rs372040364 Chr.4: 112648259-112648259 on Build GRCh38
rs754275028 Chr.4: 112647976-112647980 on Build GRCh38
rs759670726 Chr.4: 112648298-112648298 on Build GRCh38
rs949248695 Chr.4: 112648082-112648082 on Build GRCh38
rs535597757 Chr.4: 112647898-112647898 on Build GRCh38
rs770876012 Chr.4: 112648006-112648006 on Build GRCh38
rs376629554 Chr.4: 112647929-112647929 on Build GRCh38
rs745640158 Chr.4: 112648077-112648077 on Build GRCh38
rs892479152 Chr.4: 112648166-112648166 on Build GRCh38
rs201912794 Chr.4: 112648033-112648033 on Build GRCh38
rs753607927 Chr.4: 112648280-112648280 on Build GRCh38
rs561936580 Chr.4: 112648271-112648271 on Build GRCh38
rs751269136 Chr.4: 112647885-112647886 on Build GRCh38
rs773961442 Chr.4: 112648150-112648150 on Build GRCh38
rs776797112 Chr.4: 112648096-112648097 on Build GRCh38
rs769536153 Chr.4: 112647981-112647981 on Build GRCh38
rs757092597 Chr.4: 112647878-112647878 on Build GRCh38
rs778523465 Chr.4: 112648229-112648229 on Build GRCh38
rs370548538 Chr.4: 112648037-112648037 on Build GRCh38
rs746814799 Chr.4: 112648239-112648239 on Build GRCh38
rs780791325 Chr.4: 112647972-112647973 on Build GRCh38
rs569377692 Chr.4: 112648330-112648330 on Build GRCh38
rs781077657 Chr.4: 112648248-112648248 on Build GRCh38
rs987252205 Chr.4: 112648296-112648296 on Build GRCh38
rs1019896774 Chr.4: 112648169-112648169 on Build GRCh38
rs113912016 Chr.4: 112647963-112647964 on Build GRCh38
rs779103063 Chr.4: 112648026-112648034 on Build GRCh38
rs369906379 Chr.4: 112647933-112647933 on Build GRCh38
rs1025449508 Chr.4: 112648180-112648180 on Build GRCh38
rs754088075 Chr.4: 112648039-112648039 on Build GRCh38
rs747791358 Chr.4: 112648204-112648207 on Build GRCh38
rs1013348949 Chr.4: 112647888-112647888 on Build GRCh38
rs766669262 Chr.4: 112648212-112648212 on Build GRCh38
rs1000855041 Chr.4: 112647944-112647944 on Build GRCh38
rs777290469 Chr.4: 112648062-112648062 on Build GRCh38
rs767348693 Chr.4: 112648024-112648024 on Build GRCh38
rs748870691 Chr.4: 112648016-112648016 on Build GRCh38
rs565608094 Chr.4: 112648250-112648250 on Build GRCh38
rs755262015 Chr.4: 112648324-112648338 on Build GRCh38
rs546014795 Chr.4: 112648167-112648167 on Build GRCh38
rs754247159 Chr.4: 112647971-112647971 on Build GRCh38
rs762025461 Chr.4: 112648086-112648086 on Build GRCh38
rs566921391 Chr.4: 112648023-112648023 on Build GRCh38
rs1036300321 Chr.4: 112648073-112648073 on Build GRCh38
rs746252165 Chr.4: 112648038-112648046 on Build GRCh38
rs759279233 Chr.4: 112648157-112648157 on Build GRCh38
rs758053064 Chr.4: 112648325-112648325 on Build GRCh38
rs773287194 Chr.4: 112648256-112648256 on Build GRCh38
rs761615635 Chr.4: 112648102-112648108 on Build GRCh38
rs768598159 Chr.4: 112648091-112648097 on Build GRCh38
rs758844548 Chr.4: 112647978-112647978 on Build GRCh38
rs575245475 Chr.4: 112648160-112648160 on Build GRCh38
rs759554748 Chr.4: 112648020-112648020 on Build GRCh38
rs779207004 Chr.4: 112647972-112647972 on Build GRCh38
rs762723524 Chr.4: 112648113-112648114 on Build GRCh38
rs763188281 Chr.4: 112648209-112648209 on Build GRCh38
rs766460796 Chr.4: 112647943-112647943 on Build GRCh38
rs963244160 Chr.4: 112648049-112648049 on Build GRCh38
rs772091925 Chr.4: 112648078-112648078 on Build GRCh38
rs1054243777 Chr.4: 112648134-112648134 on Build GRCh38
rs747278241 Chr.4: 112648070-112648071 on Build GRCh38
rs772626627 Chr.4: 112648049-112648055 on Build GRCh38

View Full Product Details