Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 93767141-93767680 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TSG1
Entrez Gene ID
643432
Gene Name
tumor suppressor TSG1
Gene Aliases
-
UniGene
Hs.509936
Gene Chromosome Location
Chr.6: 93707083-93776481 on Build GRCh38
Transcripts
NR_015362.1

SNP Detail

SNP ID Chromosome Location
rs191637665 Chr.6: 93767252-93767252 on Build GRCh38
rs775070418 Chr.6: 93767526-93767526 on Build GRCh38
rs769799267 Chr.6: 93767585-93767585 on Build GRCh38
rs1049175771 Chr.6: 93767562-93767562 on Build GRCh38
rs956783087 Chr.6: 93767290-93767290 on Build GRCh38
rs910147467 Chr.6: 93767564-93767564 on Build GRCh38
rs182152200 Chr.6: 93767567-93767567 on Build GRCh38
rs906833515 Chr.6: 93767261-93767261 on Build GRCh38
rs926344004 Chr.6: 93767366-93767366 on Build GRCh38
rs966721398 Chr.6: 93767496-93767496 on Build GRCh38
rs930740546 Chr.6: 93767544-93767544 on Build GRCh38
rs368565846 Chr.6: 93767598-93767598 on Build GRCh38
rs762601132 Chr.6: 93767613-93767613 on Build GRCh38
rs114159393 Chr.6: 93767374-93767374 on Build GRCh38
rs967674680 Chr.6: 93767347-93767347 on Build GRCh38
rs953556895 Chr.6: 93767387-93767387 on Build GRCh38
rs1012799702 Chr.6: 93767249-93767249 on Build GRCh38
rs186876132 Chr.6: 93767351-93767351 on Build GRCh38
rs899859110 Chr.6: 93767640-93767641 on Build GRCh38
rs919282905 Chr.6: 93767537-93767537 on Build GRCh38
rs899931052 Chr.6: 93767248-93767248 on Build GRCh38
rs972074570 Chr.6: 93767530-93767530 on Build GRCh38
rs894351007 Chr.6: 93767236-93767236 on Build GRCh38
rs930145693 Chr.6: 93767447-93767447 on Build GRCh38
rs1039688951 Chr.6: 93767617-93767617 on Build GRCh38
rs1009484858 Chr.6: 93767307-93767307 on Build GRCh38
rs78361730 Chr.6: 93767323-93767323 on Build GRCh38
rs934082978 Chr.6: 93767650-93767650 on Build GRCh38
rs367593316 Chr.6: 93767490-93767490 on Build GRCh38
rs1048915817 Chr.6: 93767210-93767210 on Build GRCh38
rs1047267749 Chr.6: 93767543-93767543 on Build GRCh38
rs1031783338 Chr.6: 93767310-93767310 on Build GRCh38
rs182558688 Chr.6: 93767321-93767321 on Build GRCh38
rs998550881 Chr.6: 93767286-93767286 on Build GRCh38
rs529850508 Chr.6: 93767648-93767648 on Build GRCh38
rs960101653 Chr.6: 93767368-93767368 on Build GRCh38
rs1029485056 Chr.6: 93767276-93767276 on Build GRCh38
rs189794345 Chr.6: 93767406-93767406 on Build GRCh38

View Full Product Details