Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 163338591-163339095 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DKFZp451B082
Entrez Gene ID
401282
Gene Name
uncharacterized LOC401282
Gene Aliases
-
UniGene
Hs.444817
Gene Chromosome Location
Chr.6: 163338342-163347033 on Build GRCh38
Transcripts
NR_033862.1

SNP Detail

SNP ID Chromosome Location
rs183083745 Chr.6: 163338882-163338882 on Build GRCh38
rs1056519341 Chr.6: 163338836-163338836 on Build GRCh38
rs7754281 Chr.6: 163338724-163338724 on Build GRCh38
rs553481123 Chr.6: 163338959-163338959 on Build GRCh38
rs1001012659 Chr.6: 163338924-163338924 on Build GRCh38
rs987177854 Chr.6: 163338761-163338761 on Build GRCh38
rs978088322 Chr.6: 163339058-163339058 on Build GRCh38
rs746934853 Chr.6: 163338809-163338809 on Build GRCh38
rs530899985 Chr.6: 163338679-163338679 on Build GRCh38
rs993845400 Chr.6: 163338938-163338938 on Build GRCh38
rs370634535 Chr.6: 163338613-163338613 on Build GRCh38
rs565462843 Chr.6: 163338642-163338642 on Build GRCh38
rs1008061369 Chr.6: 163338994-163338994 on Build GRCh38
rs753856034 Chr.6: 163338625-163338625 on Build GRCh38
rs1044869667 Chr.6: 163338907-163338907 on Build GRCh38
rs373977723 Chr.6: 163338755-163338755 on Build GRCh38
rs575852805 Chr.6: 163339038-163339038 on Build GRCh38
rs758318871 Chr.6: 163338743-163338743 on Build GRCh38
rs1056598521 Chr.6: 163338892-163338892 on Build GRCh38
rs62430074 Chr.6: 163338899-163338899 on Build GRCh38
rs62430073 Chr.6: 163338816-163338816 on Build GRCh38
rs772874502 Chr.6: 163338913-163338913 on Build GRCh38
rs564730657 Chr.6: 163338838-163338838 on Build GRCh38
rs12525070 Chr.6: 163338762-163338762 on Build GRCh38
rs1052631285 Chr.6: 163338955-163338955 on Build GRCh38
rs930561220 Chr.6: 163338616-163338616 on Build GRCh38
rs940628265 Chr.6: 163338833-163338833 on Build GRCh38
rs1012324274 Chr.6: 163338615-163338615 on Build GRCh38
rs921762104 Chr.6: 163338808-163338808 on Build GRCh38
rs551004962 Chr.6: 163338701-163338701 on Build GRCh38
rs561317817 Chr.6: 163338707-163338707 on Build GRCh38
rs566654539 Chr.6: 163338746-163338746 on Build GRCh38
rs780865980 Chr.6: 163338821-163338821 on Build GRCh38
rs12525069 Chr.6: 163338756-163338756 on Build GRCh38
rs75140423 Chr.6: 163338881-163338881 on Build GRCh38
rs545288495 Chr.6: 163338981-163338981 on Build GRCh38
rs898135673 Chr.6: 163338931-163338931 on Build GRCh38
rs865924215 Chr.6: 163338795-163338795 on Build GRCh38
rs968064500 Chr.6: 163339040-163339040 on Build GRCh38
rs892697930 Chr.6: 163338975-163338975 on Build GRCh38
rs1026986587 Chr.6: 163339034-163339034 on Build GRCh38
rs775702596 Chr.6: 163338794-163338794 on Build GRCh38
rs964080934 Chr.6: 163338771-163338771 on Build GRCh38
rs56195563 Chr.6: 163339033-163339033 on Build GRCh38
rs886364063 Chr.6: 163338652-163338652 on Build GRCh38
rs537805172 Chr.6: 163338842-163338842 on Build GRCh38
rs964049198 Chr.6: 163339013-163339013 on Build GRCh38
rs147669446 Chr.6: 163339063-163339063 on Build GRCh38
rs532173153 Chr.6: 163338875-163338875 on Build GRCh38
rs939504332 Chr.6: 163338884-163338884 on Build GRCh38
rs1020033060 Chr.6: 163338988-163338988 on Build GRCh38
rs62430072 Chr.6: 163338745-163338745 on Build GRCh38
rs778677463 Chr.6: 163338637-163338637 on Build GRCh38
rs187745585 Chr.6: 163338976-163338976 on Build GRCh38
rs746269817 Chr.6: 163338972-163338972 on Build GRCh38
rs1049003558 Chr.6: 163338621-163338621 on Build GRCh38
rs904059534 Chr.6: 163338744-163338744 on Build GRCh38
rs964032949 Chr.6: 163338798-163338798 on Build GRCh38
rs975510896 Chr.6: 163338774-163338774 on Build GRCh38

View Full Product Details