Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 39406232-39406743 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
POU6F2
Entrez Gene ID
11281
Gene Name
POU class 6 homeobox 2
Gene Aliases
RPF-1, WT5, WTSL
UniGene
Hs.137106
Gene Chromosome Location
Chr.7: 38977878-39468601 on Build GRCh38
Transcripts
NM_001166018.1 NM_007252.3 XM_011515113.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4844180 c.910C>T p.Q304* Chr.7: 39406624-39406624 on Build GRCh38
1089507 c.965G>C p.G322A Chr.7: 39406679-39406679 on Build GRCh38
6498835 c.936G>A p.M312I Chr.7: 39406650-39406650 on Build GRCh38
1089508 c.971C>A p.A324D Chr.7: 39406685-39406685 on Build GRCh38
6420116 c.1025A>C p.Q342P Chr.7: 39406739-39406739 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs577609665 Chr.7: 39406512-39406512 on Build GRCh38
rs747368655 Chr.7: 39406693-39406693 on Build GRCh38
rs894526539 Chr.7: 39406493-39406493 on Build GRCh38
rs11354354 Chr.7: 39406415-39406415 on Build GRCh38
rs769103198 Chr.7: 39406568-39406568 on Build GRCh38
rs1033101394 Chr.7: 39406657-39406657 on Build GRCh38
rs766591913 Chr.7: 39406599-39406599 on Build GRCh38
rs760205418 Chr.7: 39406625-39406626 on Build GRCh38
rs748605828 Chr.7: 39406667-39406667 on Build GRCh38
rs148810830 Chr.7: 39406407-39406407 on Build GRCh38
rs141249466 Chr.7: 39406650-39406650 on Build GRCh38
rs765097323 Chr.7: 39406633-39406633 on Build GRCh38
rs998839813 Chr.7: 39406409-39406409 on Build GRCh38
rs555806902 Chr.7: 39406369-39406369 on Build GRCh38
rs769282596 Chr.7: 39406658-39406658 on Build GRCh38
rs771329494 Chr.7: 39406650-39406651 on Build GRCh38
rs1034406108 Chr.7: 39406489-39406489 on Build GRCh38
rs779320120 Chr.7: 39406661-39406661 on Build GRCh38
rs774864814 Chr.7: 39406578-39406578 on Build GRCh38
rs34498132 Chr.7: 39406447-39406448 on Build GRCh38
rs941679212 Chr.7: 39406315-39406315 on Build GRCh38
rs778523090 Chr.7: 39406720-39406720 on Build GRCh38
rs202118321 Chr.7: 39406416-39406416 on Build GRCh38
rs536031914 Chr.7: 39406308-39406308 on Build GRCh38
rs386712393 Chr.7: 39406415-39406417 on Build GRCh38
rs767456859 Chr.7: 39406414-39406415 on Build GRCh38
rs1036736262 Chr.7: 39406577-39406577 on Build GRCh38
rs771114500 Chr.7: 39406697-39406697 on Build GRCh38
rs999551323 Chr.7: 39406515-39406515 on Build GRCh38
rs751372756 Chr.7: 39406640-39406640 on Build GRCh38
rs1037347868 Chr.7: 39406376-39406376 on Build GRCh38
rs181595446 Chr.7: 39406461-39406461 on Build GRCh38
rs201463016 Chr.7: 39406583-39406583 on Build GRCh38
rs906400025 Chr.7: 39406473-39406473 on Build GRCh38
rs965658738 Chr.7: 39406395-39406395 on Build GRCh38
rs757039637 Chr.7: 39406642-39406642 on Build GRCh38
rs955052623 Chr.7: 39406530-39406530 on Build GRCh38
rs184581147 Chr.7: 39406272-39406272 on Build GRCh38
rs1032968466 Chr.7: 39406307-39406320 on Build GRCh38
rs772938783 Chr.7: 39406587-39406588 on Build GRCh38
rs939709924 Chr.7: 39406565-39406565 on Build GRCh38
rs926057709 Chr.7: 39406433-39406433 on Build GRCh38
rs549436954 Chr.7: 39406430-39406430 on Build GRCh38
rs866917384 Chr.7: 39406519-39406519 on Build GRCh38
rs773444754 Chr.7: 39406585-39406585 on Build GRCh38
rs773606054 Chr.7: 39406678-39406678 on Build GRCh38
rs376549423 Chr.7: 39406590-39406590 on Build GRCh38
rs967384474 Chr.7: 39406503-39406503 on Build GRCh38
rs760322303 Chr.7: 39406548-39406548 on Build GRCh38
rs985910505 Chr.7: 39406284-39406284 on Build GRCh38
rs201195691 Chr.7: 39406415-39406415 on Build GRCh38
rs988774869 Chr.7: 39406454-39406454 on Build GRCh38
rs1030573933 Chr.7: 39406523-39406523 on Build GRCh38
rs150386704 Chr.7: 39406704-39406704 on Build GRCh38
rs759645219 Chr.7: 39406623-39406623 on Build GRCh38
rs750674145 Chr.7: 39406521-39406521 on Build GRCh38
rs34812541 Chr.7: 39406417-39406417 on Build GRCh38
rs1022071290 Chr.7: 39406495-39406495 on Build GRCh38
rs1001986338 Chr.7: 39406483-39406483 on Build GRCh38
rs753930640 Chr.7: 39406606-39406606 on Build GRCh38
rs202231692 Chr.7: 39406417-39406417 on Build GRCh38
rs772545735 Chr.7: 39406673-39406673 on Build GRCh38
rs1011553909 Chr.7: 39406494-39406494 on Build GRCh38
rs1046293736 Chr.7: 39406470-39406470 on Build GRCh38
rs148254024 Chr.7: 39406635-39406635 on Build GRCh38
rs752611585 Chr.7: 39406634-39406634 on Build GRCh38
rs770178428 Chr.7: 39406576-39406576 on Build GRCh38
rs937863156 Chr.7: 39406505-39406505 on Build GRCh38
rs376654925 Chr.7: 39406648-39406648 on Build GRCh38
rs745382420 Chr.7: 39406566-39406566 on Build GRCh38
rs878893496 Chr.7: 39406571-39406571 on Build GRCh38

View Full Product Details