Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 158532614-158533114 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MIR595
Entrez Gene ID
693180
Gene Name
microRNA 595
Gene Aliases
MIRN595, hsa-mir-595
UniGene
Gene Chromosome Location
Chr.7: 158532718-158532813 on Build GRCh38
Transcripts
NR_030325.1 NM_001308267.1 NM_001308268.1 NM_002847.4 NM_130842.3 NM_130843.3 XM_011516446.1 XM_011516447.2 XM_011516448.2 XM_011516449.2 XM_017012476.1

Gene Symbol
PTPRN2
Entrez Gene ID
5799
Gene Name
protein tyrosine phosphatase, receptor type N2
Gene Aliases
IA-2beta, IAR, ICAAR, PTPRP, R-PTP-N2
UniGene
Hs.490789
Gene Chromosome Location
Chr.7: 157539052-158587802 on Build GRCh38
Transcripts
NR_030325.1 NM_001308267.1 NM_001308268.1 NM_002847.4 NM_130842.3 NM_130843.3 XM_011516446.1 XM_011516447.2 XM_011516448.2 XM_011516449.2 XM_017012476.1

SNP Detail

SNP ID Chromosome Location
rs553527634 Chr.7: 158532862-158532862 on Build GRCh38
rs894966134 Chr.7: 158533087-158533087 on Build GRCh38
rs117344178 Chr.7: 158532719-158532719 on Build GRCh38
rs112159390 Chr.7: 158532895-158532895 on Build GRCh38
rs139230958 Chr.7: 158532869-158532869 on Build GRCh38
rs374577536 Chr.7: 158532807-158532807 on Build GRCh38
rs565878297 Chr.7: 158532647-158532647 on Build GRCh38
rs377680720 Chr.7: 158532785-158532785 on Build GRCh38
rs773873816 Chr.7: 158532776-158532776 on Build GRCh38
rs4909238 Chr.7: 158532880-158532880 on Build GRCh38
rs911350703 Chr.7: 158532914-158532914 on Build GRCh38
rs749190661 Chr.7: 158532858-158532858 on Build GRCh38
rs767128325 Chr.7: 158532782-158532782 on Build GRCh38
rs751208072 Chr.7: 158532669-158532669 on Build GRCh38
rs12670231 Chr.7: 158532646-158532646 on Build GRCh38
rs1029507022 Chr.7: 158532801-158532801 on Build GRCh38
rs370327205 Chr.7: 158532894-158532894 on Build GRCh38
rs761267002 Chr.7: 158533036-158533036 on Build GRCh38
rs528394938 Chr.7: 158532693-158532693 on Build GRCh38
rs769625770 Chr.7: 158532851-158532851 on Build GRCh38
rs370078657 Chr.7: 158532846-158532846 on Build GRCh38
rs1054522048 Chr.7: 158533040-158533040 on Build GRCh38
rs113721796 Chr.7: 158532987-158532987 on Build GRCh38
rs941062116 Chr.7: 158532646-158532646 on Build GRCh38
rs771765680 Chr.7: 158533047-158533047 on Build GRCh38
rs1037466418 Chr.7: 158533068-158533068 on Build GRCh38
rs201352992 Chr.7: 158532812-158532812 on Build GRCh38
rs188826916 Chr.7: 158532710-158532710 on Build GRCh38
rs374140691 Chr.7: 158532775-158532775 on Build GRCh38
rs369465307 Chr.7: 158532743-158532743 on Build GRCh38
rs775484225 Chr.7: 158532765-158532765 on Build GRCh38
rs375146018 Chr.7: 158532737-158532737 on Build GRCh38
rs756907469 Chr.7: 158532681-158532681 on Build GRCh38
rs202194191 Chr.7: 158532800-158532800 on Build GRCh38
rs1012547276 Chr.7: 158532856-158532856 on Build GRCh38
rs977398994 Chr.7: 158532965-158532965 on Build GRCh38
rs757960484 Chr.7: 158532931-158532931 on Build GRCh38
rs4909237 Chr.7: 158532811-158532811 on Build GRCh38
rs762864463 Chr.7: 158532774-158532774 on Build GRCh38
rs761415486 Chr.7: 158532779-158532779 on Build GRCh38
rs765589180 Chr.7: 158532799-158532799 on Build GRCh38
rs932968479 Chr.7: 158533015-158533015 on Build GRCh38
rs942890160 Chr.7: 158532951-158532951 on Build GRCh38
rs779630994 Chr.7: 158533042-158533042 on Build GRCh38
rs929554670 Chr.7: 158532694-158532694 on Build GRCh38
rs929603704 Chr.7: 158532868-158532868 on Build GRCh38
rs918261059 Chr.7: 158532866-158532866 on Build GRCh38
rs563198461 Chr.7: 158532900-158532900 on Build GRCh38
rs781540130 Chr.7: 158532839-158532839 on Build GRCh38
rs745641777 Chr.7: 158532747-158532747 on Build GRCh38
rs758846227 Chr.7: 158532802-158532802 on Build GRCh38
rs754232990 Chr.7: 158532697-158532697 on Build GRCh38
rs114151270 Chr.7: 158532723-158532723 on Build GRCh38
rs750126925 Chr.7: 158532783-158532783 on Build GRCh38
rs769523925 Chr.7: 158532758-158532758 on Build GRCh38
rs779717492 Chr.7: 158532852-158532852 on Build GRCh38
rs113547124 Chr.7: 158532883-158532883 on Build GRCh38
rs1024927471 Chr.7: 158532867-158532867 on Build GRCh38
rs747174159 Chr.7: 158532724-158532724 on Build GRCh38
rs899450966 Chr.7: 158532673-158532673 on Build GRCh38
rs956004924 Chr.7: 158532791-158532791 on Build GRCh38
rs1047400995 Chr.7: 158533023-158533023 on Build GRCh38

View Full Product Details