Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 20973852-20974345 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC286114
Entrez Gene ID
286114
Gene Name
uncharacterized LOC286114
Gene Aliases
-
UniGene
Hs.651884
Gene Chromosome Location
Chr.8: 20973986-20995119 on Build GRCh38
Transcripts
NR_033894.1

SNP Detail

SNP ID Chromosome Location
rs1040149416 Chr.8: 20974172-20974172 on Build GRCh38
rs373674661 Chr.8: 20973891-20973891 on Build GRCh38
rs368657388 Chr.8: 20973969-20973969 on Build GRCh38
rs953491791 Chr.8: 20974013-20974013 on Build GRCh38
rs533322833 Chr.8: 20973975-20973975 on Build GRCh38
rs773632502 Chr.8: 20974236-20974236 on Build GRCh38
rs78575314 Chr.8: 20973990-20973990 on Build GRCh38
rs948408593 Chr.8: 20973882-20973882 on Build GRCh38
rs182643781 Chr.8: 20974230-20974230 on Build GRCh38
rs748401841 Chr.8: 20974190-20974190 on Build GRCh38
rs547634772 Chr.8: 20974271-20974271 on Build GRCh38
rs932065609 Chr.8: 20973904-20973904 on Build GRCh38
rs920228110 Chr.8: 20974078-20974078 on Build GRCh38
rs897498515 Chr.8: 20973955-20973955 on Build GRCh38
rs555301334 Chr.8: 20973977-20973977 on Build GRCh38
rs1048845663 Chr.8: 20973963-20973963 on Build GRCh38
rs529156591 Chr.8: 20974259-20974259 on Build GRCh38
rs960941232 Chr.8: 20974249-20974249 on Build GRCh38
rs919143149 Chr.8: 20974062-20974062 on Build GRCh38
rs541233636 Chr.8: 20974068-20974068 on Build GRCh38
rs915284104 Chr.8: 20973899-20973899 on Build GRCh38
rs561025842 Chr.8: 20974159-20974159 on Build GRCh38
rs1036259687 Chr.8: 20973943-20973943 on Build GRCh38
rs1048795632 Chr.8: 20974093-20974093 on Build GRCh38
rs780272886 Chr.8: 20974018-20974018 on Build GRCh38
rs568494 Chr.8: 20974178-20974178 on Build GRCh38
rs1044444064 Chr.8: 20973886-20973886 on Build GRCh38
rs900598743 Chr.8: 20974174-20974174 on Build GRCh38
rs568373699 Chr.8: 20974255-20974255 on Build GRCh38
rs551930315 Chr.8: 20974114-20974115 on Build GRCh38
rs1011101017 Chr.8: 20974187-20974187 on Build GRCh38
rs62499586 Chr.8: 20974130-20974130 on Build GRCh38
rs575855857 Chr.8: 20974188-20974188 on Build GRCh38
rs1019009478 Chr.8: 20973956-20973956 on Build GRCh38
rs137962772 Chr.8: 20974006-20974006 on Build GRCh38
rs370557194 Chr.8: 20973889-20973889 on Build GRCh38
rs866564472 Chr.8: 20974009-20974009 on Build GRCh38
rs745529730 Chr.8: 20974202-20974202 on Build GRCh38
rs999517427 Chr.8: 20974219-20974219 on Build GRCh38
rs906701031 Chr.8: 20974152-20974152 on Build GRCh38
rs150744729 Chr.8: 20973981-20973981 on Build GRCh38
rs371317538 Chr.8: 20974043-20974043 on Build GRCh38
rs566181431 Chr.8: 20973970-20973970 on Build GRCh38
rs1031217026 Chr.8: 20973989-20973989 on Build GRCh38
rs915234802 Chr.8: 20974011-20974011 on Build GRCh38
rs535689523 Chr.8: 20973920-20973920 on Build GRCh38
rs919079743 Chr.8: 20974273-20974273 on Build GRCh38
rs17093448 Chr.8: 20974226-20974226 on Build GRCh38
rs996200860 Chr.8: 20973973-20973973 on Build GRCh38
rs1045454734 Chr.8: 20974141-20974141 on Build GRCh38
rs944527651 Chr.8: 20974038-20974038 on Build GRCh38
rs758930422 Chr.8: 20974081-20974081 on Build GRCh38
rs894134991 Chr.8: 20974122-20974122 on Build GRCh38
rs989478652 Chr.8: 20973998-20973998 on Build GRCh38
rs994472344 Chr.8: 20973958-20973958 on Build GRCh38
rs911733173 Chr.8: 20974021-20974021 on Build GRCh38
rs189964772 Chr.8: 20974044-20974044 on Build GRCh38
rs930588400 Chr.8: 20974126-20974126 on Build GRCh38
rs752395341 Chr.8: 20973915-20973915 on Build GRCh38
rs141278137 Chr.8: 20973935-20973935 on Build GRCh38
rs1004214173 Chr.8: 20973982-20973982 on Build GRCh38
rs956985348 Chr.8: 20974186-20974186 on Build GRCh38
rs763435962 Chr.8: 20974269-20974269 on Build GRCh38
rs989530475 Chr.8: 20973898-20973898 on Build GRCh38
rs992333711 Chr.8: 20974305-20974305 on Build GRCh38
rs185207617 Chr.8: 20973912-20973912 on Build GRCh38
rs944643902 Chr.8: 20973941-20973941 on Build GRCh38
rs751997817 Chr.8: 20974124-20974124 on Build GRCh38
rs550078837 Chr.8: 20973888-20973888 on Build GRCh38
rs986633963 Chr.8: 20974213-20974213 on Build GRCh38
rs966274714 Chr.8: 20974235-20974235 on Build GRCh38
rs1008723284 Chr.8: 20974171-20974171 on Build GRCh38

View Full Product Details