Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 21995032-21995528 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDKN2A
Entrez Gene ID
1029
Gene Name
cyclin dependent kinase inhibitor 2A
Gene Aliases
ARF, CDK4I, CDKN2, CMM2, INK4, INK4A, MLM, MTS-1, MTS1, P14, P14ARF, P16, P16-INK4A, P16INK4, P16INK4A, P19, P19ARF, TP16
UniGene
Hs.512599
Gene Chromosome Location
Chr.9: 21967752-21995043 on Build GRCh38
Transcripts
XM_011517679.1 NR_003529.3 NR_047532.1 NR_047533.1 NR_047534.1 NR_047535.1 NR_047536.1 NR_047537.1 NR_047538.1 NR_047539.1 NR_047540.1 NR_047541.1 NR_047542.1 NR_047543.1 NR_120536.1

Gene Symbol
CDKN2B-AS1
Entrez Gene ID
100048912
Gene Name
CDKN2B antisense RNA 1
Gene Aliases
ANRIL, CDKN2B-AS, CDKN2BAS, NCRNA00089, PCAT12, p15AS
UniGene
Hs.493614
Gene Chromosome Location
Chr.9: 21994791-22121097 on Build GRCh38
Transcripts
XM_011517679.1 NR_003529.3 NR_047532.1 NR_047533.1 NR_047534.1 NR_047535.1 NR_047536.1 NR_047537.1 NR_047538.1 NR_047539.1 NR_047540.1 NR_047541.1 NR_047542.1 NR_047543.1 NR_120536.1

SNP Detail

SNP ID Chromosome Location
rs540953023 Chr.9: 21995468-21995468 on Build GRCh38
rs771270265 Chr.9: 21995156-21995156 on Build GRCh38
rs184930832 Chr.9: 21995087-21995087 on Build GRCh38
rs180959346 Chr.9: 21995469-21995469 on Build GRCh38
rs940346959 Chr.9: 21995282-21995282 on Build GRCh38
rs1010427763 Chr.9: 21995476-21995476 on Build GRCh38
rs1041446597 Chr.9: 21995491-21995491 on Build GRCh38
rs980248308 Chr.9: 21995187-21995187 on Build GRCh38
rs1057396968 Chr.9: 21995456-21995456 on Build GRCh38
rs964860460 Chr.9: 21995359-21995359 on Build GRCh38
rs923320414 Chr.9: 21995155-21995155 on Build GRCh38
rs111839442 Chr.9: 21995136-21995136 on Build GRCh38
rs943519892 Chr.9: 21995139-21995139 on Build GRCh38
rs1050191774 Chr.9: 21995355-21995355 on Build GRCh38
rs1001094833 Chr.9: 21995327-21995327 on Build GRCh38
rs1037605139 Chr.9: 21995189-21995189 on Build GRCh38
rs979644416 Chr.9: 21995120-21995120 on Build GRCh38
rs974814898 Chr.9: 21995153-21995153 on Build GRCh38
rs578242143 Chr.9: 21995311-21995311 on Build GRCh38
rs1037636294 Chr.9: 21995432-21995432 on Build GRCh38
rs1040157860 Chr.9: 21995309-21995309 on Build GRCh38
rs927548729 Chr.9: 21995210-21995210 on Build GRCh38
rs988850485 Chr.9: 21995109-21995109 on Build GRCh38
rs1020406800 Chr.9: 21995307-21995307 on Build GRCh38
rs941987522 Chr.9: 21995420-21995420 on Build GRCh38
rs562748036 Chr.9: 21995183-21995183 on Build GRCh38
rs954998637 Chr.9: 21995401-21995401 on Build GRCh38
rs903791253 Chr.9: 21995362-21995362 on Build GRCh38
rs1032530491 Chr.9: 21995353-21995353 on Build GRCh38
rs542564280 Chr.9: 21995113-21995113 on Build GRCh38
rs976133113 Chr.9: 21995488-21995488 on Build GRCh38
rs905714166 Chr.9: 21995191-21995191 on Build GRCh38
rs576847829 Chr.9: 21995081-21995081 on Build GRCh38
rs548699520 Chr.9: 21995370-21995370 on Build GRCh38
rs931838144 Chr.9: 21995344-21995344 on Build GRCh38
rs562374859 Chr.9: 21995239-21995239 on Build GRCh38
rs544572683 Chr.9: 21995276-21995276 on Build GRCh38
rs1028894184 Chr.9: 21995117-21995117 on Build GRCh38
rs1000170568 Chr.9: 21995129-21995129 on Build GRCh38
rs996382328 Chr.9: 21995506-21995506 on Build GRCh38
rs1032563092 Chr.9: 21995500-21995500 on Build GRCh38
rs189042431 Chr.9: 21995278-21995278 on Build GRCh38
rs774601528 Chr.9: 21995402-21995402 on Build GRCh38
rs556810622 Chr.9: 21995058-21995058 on Build GRCh38
rs968396890 Chr.9: 21995312-21995312 on Build GRCh38
rs372430243 Chr.9: 21995325-21995325 on Build GRCh38
rs886439156 Chr.9: 21995181-21995181 on Build GRCh38
rs551685870 Chr.9: 21995504-21995509 on Build GRCh38
rs553593710 Chr.9: 21995104-21995104 on Build GRCh38
rs907890833 Chr.9: 21995404-21995404 on Build GRCh38
rs1035447916 Chr.9: 21995215-21995215 on Build GRCh38
rs1048767329 Chr.9: 21995260-21995260 on Build GRCh38
rs996404168 Chr.9: 21995111-21995111 on Build GRCh38
rs964891735 Chr.9: 21995503-21995503 on Build GRCh38
rs1048152644 Chr.9: 21995179-21995179 on Build GRCh38
rs925481547 Chr.9: 21995138-21995138 on Build GRCh38
rs999787793 Chr.9: 21995498-21995498 on Build GRCh38
rs891830653 Chr.9: 21995391-21995391 on Build GRCh38
rs546649896 Chr.9: 21995361-21995361 on Build GRCh38
rs1001308679 Chr.9: 21995073-21995073 on Build GRCh38
rs895518558 Chr.9: 21995227-21995227 on Build GRCh38
rs112789036 Chr.9: 21995145-21995145 on Build GRCh38
rs980280854 Chr.9: 21995447-21995447 on Build GRCh38
rs904203740 Chr.9: 21995494-21995494 on Build GRCh38
rs1001341190 Chr.9: 21995213-21995213 on Build GRCh38
rs533242911 Chr.9: 21995279-21995279 on Build GRCh38
rs927161876 Chr.9: 21995435-21995435 on Build GRCh38
rs530846411 Chr.9: 21995241-21995241 on Build GRCh38
rs988951589 Chr.9: 21995141-21995141 on Build GRCh38
rs746283593 Chr.9: 21995441-21995441 on Build GRCh38
rs528241798 Chr.9: 21995445-21995445 on Build GRCh38
rs1030875886 Chr.9: 21995375-21995375 on Build GRCh38
rs974847321 Chr.9: 21995365-21995365 on Build GRCh38
rs74582729 Chr.9: 21995110-21995110 on Build GRCh38
rs933343355 Chr.9: 21995160-21995160 on Build GRCh38

View Full Product Details