Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 27276856-27277356 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LINC00032
Entrez Gene ID
158035
Gene Name
long intergenic non-protein coding RNA 32
Gene Aliases
C9orf14, NCRNA00032
UniGene
Hs.201554
Gene Chromosome Location
Chr.9: 27245684-27282793 on Build GRCh38
Transcripts
NR_026679.1

SNP Detail

SNP ID Chromosome Location
rs1029149173 Chr.9: 27276948-27276948 on Build GRCh38
rs559133254 Chr.9: 27276925-27276925 on Build GRCh38
rs991742196 Chr.9: 27276927-27276927 on Build GRCh38
rs192111066 Chr.9: 27276939-27276939 on Build GRCh38
rs531267592 Chr.9: 27277256-27277256 on Build GRCh38
rs375397060 Chr.9: 27277253-27277253 on Build GRCh38
rs1023297628 Chr.9: 27277175-27277175 on Build GRCh38
rs529447409 Chr.9: 27277308-27277308 on Build GRCh38
rs539437544 Chr.9: 27277045-27277045 on Build GRCh38
rs903281368 Chr.9: 27277305-27277305 on Build GRCh38
rs932251769 Chr.9: 27277202-27277202 on Build GRCh38
rs542133823 Chr.9: 27277180-27277180 on Build GRCh38
rs77761279 Chr.9: 27277291-27277291 on Build GRCh38
rs538045364 Chr.9: 27277053-27277053 on Build GRCh38
rs998821511 Chr.9: 27276897-27276897 on Build GRCh38
rs548105782 Chr.9: 27277310-27277310 on Build GRCh38
rs973936737 Chr.9: 27277297-27277297 on Build GRCh38
rs183232388 Chr.9: 27277239-27277239 on Build GRCh38
rs566211108 Chr.9: 27277315-27277315 on Build GRCh38
rs932307148 Chr.9: 27277247-27277247 on Build GRCh38
rs989129828 Chr.9: 27277032-27277032 on Build GRCh38
rs761883850 Chr.9: 27277275-27277275 on Build GRCh38
rs756291946 Chr.9: 27276901-27276901 on Build GRCh38
rs558303104 Chr.9: 27277046-27277046 on Build GRCh38
rs996862200 Chr.9: 27276941-27276941 on Build GRCh38
rs927793293 Chr.9: 27277071-27277071 on Build GRCh38
rs892201513 Chr.9: 27277230-27277230 on Build GRCh38
rs570114675 Chr.9: 27277051-27277051 on Build GRCh38
rs922297807 Chr.9: 27277242-27277242 on Build GRCh38
rs547773462 Chr.9: 27276978-27276978 on Build GRCh38
rs570554179 Chr.9: 27277153-27277153 on Build GRCh38
rs940785624 Chr.9: 27277112-27277112 on Build GRCh38
rs982551439 Chr.9: 27277277-27277279 on Build GRCh38
rs959919236 Chr.9: 27276916-27276916 on Build GRCh38
rs899186635 Chr.9: 27277197-27277197 on Build GRCh38
rs568424213 Chr.9: 27276909-27276909 on Build GRCh38
rs866682464 Chr.9: 27277254-27277254 on Build GRCh38
rs1040664307 Chr.9: 27277008-27277008 on Build GRCh38
rs533364935 Chr.9: 27277321-27277321 on Build GRCh38
rs10967848 Chr.9: 27277283-27277283 on Build GRCh38
rs980412005 Chr.9: 27277243-27277243 on Build GRCh38
rs553767070 Chr.9: 27277224-27277224 on Build GRCh38
rs1012858557 Chr.9: 27277167-27277167 on Build GRCh38
rs895790858 Chr.9: 27277155-27277155 on Build GRCh38
rs952585304 Chr.9: 27277176-27277176 on Build GRCh38
rs572021777 Chr.9: 27277229-27277229 on Build GRCh38
rs1057179017 Chr.9: 27276886-27276886 on Build GRCh38
rs7038108 Chr.9: 27277027-27277027 on Build GRCh38
rs1057219458 Chr.9: 27277312-27277312 on Build GRCh38
rs923538661 Chr.9: 27277013-27277013 on Build GRCh38
rs777582381 Chr.9: 27277188-27277188 on Build GRCh38
rs556436323 Chr.9: 27277073-27277073 on Build GRCh38
rs915773849 Chr.9: 27276929-27276929 on Build GRCh38
rs375004430 Chr.9: 27277295-27277295 on Build GRCh38
rs574543158 Chr.9: 27277076-27277076 on Build GRCh38
rs542108982 Chr.9: 27276896-27276896 on Build GRCh38
rs907929839 Chr.9: 27277089-27277089 on Build GRCh38
rs1035660988 Chr.9: 27277148-27277148 on Build GRCh38
rs984323347 Chr.9: 27277217-27277217 on Build GRCh38
rs530745938 Chr.9: 27277004-27277004 on Build GRCh38
rs1043171211 Chr.9: 27277062-27277062 on Build GRCh38
rs908268327 Chr.9: 27277001-27277001 on Build GRCh38
rs933618431 Chr.9: 27277018-27277018 on Build GRCh38
rs954899071 Chr.9: 27276975-27276975 on Build GRCh38
rs998872159 Chr.9: 27277081-27277081 on Build GRCh38
rs969312571 Chr.9: 27277048-27277048 on Build GRCh38
rs1015318328 Chr.9: 27277223-27277223 on Build GRCh38
rs983922674 Chr.9: 27276998-27276998 on Build GRCh38
rs1012150598 Chr.9: 27277060-27277060 on Build GRCh38
rs1051099001 Chr.9: 27277026-27277026 on Build GRCh38
rs1008645730 Chr.9: 27276997-27276997 on Build GRCh38
rs7848478 Chr.9: 27276933-27276933 on Build GRCh38

View Full Product Details