Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 76787230-76787767 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PCA3
Entrez Gene ID
50652
Gene Name
prostate cancer associated 3 (non-protein coding)
Gene Aliases
DD3, NCRNA00019, PCAT3
UniGene
Hs.663766
Gene Chromosome Location
Chr.9: 76764436-76787569 on Build GRCh38
Transcripts
NR_015342.2 NR_132312.1 NR_132313.1 NM_001308047.1 NM_001308048.1 NM_015225.2 NR_131751.1 XM_005251745.1 XM_005251746.1 XM_005251748.1 XM_005251750.1 XM_005251751.1 XM_005251754.1 XM_006716982.1 XM_006716983.1 XM_006716984.2 XM_006716985.1 XM_006716986.1 XM_011518323.1 XM_011518326.2 XM_011518327.1 XM_011518328.2 XM_017014345.1 XM_017014346.1 XM_017014347.1 XM_017014348.1 XM_017014349.1 XM_017014350.1 XM_017014351.1 XM_017014352.1 XM_017014353.1

Gene Symbol
PRUNE2
Entrez Gene ID
158471
Gene Name
prune homolog 2
Gene Aliases
BMCC1, BNIPXL, C9orf65, KIAA0367
UniGene
Hs.262857
Gene Chromosome Location
Chr.9: 76611376-76906220 on Build GRCh38
Transcripts
NR_015342.2 NR_132312.1 NR_132313.1 NM_001308047.1 NM_001308048.1 NM_015225.2 NR_131751.1 XM_005251745.1 XM_005251746.1 XM_005251748.1 XM_005251750.1 XM_005251751.1 XM_005251754.1 XM_006716982.1 XM_006716983.1 XM_006716984.2 XM_006716985.1 XM_006716986.1 XM_011518323.1 XM_011518326.2 XM_011518327.1 XM_011518328.2 XM_017014345.1 XM_017014346.1 XM_017014347.1 XM_017014348.1 XM_017014349.1 XM_017014350.1 XM_017014351.1 XM_017014352.1 XM_017014353.1

SNP Detail

SNP ID Chromosome Location
rs938244827 Chr.9: 76787558-76787558 on Build GRCh38
rs59682576 Chr.9: 76787581-76787581 on Build GRCh38
rs5898507 Chr.9: 76787307-76787308 on Build GRCh38
rs914071054 Chr.9: 76787265-76787265 on Build GRCh38
rs922421494 Chr.9: 76787592-76787592 on Build GRCh38
rs556720928 Chr.9: 76787532-76787532 on Build GRCh38
rs768420446 Chr.9: 76787580-76787580 on Build GRCh38
rs914990753 Chr.9: 76787321-76787321 on Build GRCh38
rs146708075 Chr.9: 76787303-76787304 on Build GRCh38
rs907399829 Chr.9: 76787708-76787708 on Build GRCh38
rs768907939 Chr.9: 76787672-76787672 on Build GRCh38
rs554681591 Chr.9: 76787480-76787480 on Build GRCh38
rs756763173 Chr.9: 76787361-76787361 on Build GRCh38
rs772443891 Chr.9: 76787662-76787662 on Build GRCh38
rs137955063 Chr.9: 76787475-76787475 on Build GRCh38
rs923588885 Chr.9: 76787290-76787290 on Build GRCh38
rs941389810 Chr.9: 76787660-76787660 on Build GRCh38
rs142675841 Chr.9: 76787322-76787322 on Build GRCh38
rs917924067 Chr.9: 76787579-76787579 on Build GRCh38
rs886969950 Chr.9: 76787341-76787341 on Build GRCh38
rs989127316 Chr.9: 76787544-76787544 on Build GRCh38
rs5898508 Chr.9: 76787633-76787634 on Build GRCh38
rs908455524 Chr.9: 76787644-76787644 on Build GRCh38
rs1039543828 Chr.9: 76787345-76787345 on Build GRCh38
rs59757774 Chr.9: 76787644-76787645 on Build GRCh38
rs780941403 Chr.9: 76787384-76787384 on Build GRCh38
rs186615971 Chr.9: 76787462-76787462 on Build GRCh38
rs1005405898 Chr.9: 76787376-76787376 on Build GRCh38
rs1046325131 Chr.9: 76787698-76787698 on Build GRCh38
rs3924629 Chr.9: 76787668-76787668 on Build GRCh38
rs58399742 Chr.9: 76787278-76787278 on Build GRCh38
rs547883705 Chr.9: 76787302-76787302 on Build GRCh38
rs576665125 Chr.9: 76787545-76787545 on Build GRCh38
rs1024451694 Chr.9: 76787501-76787501 on Build GRCh38
rs776672818 Chr.9: 76787688-76787688 on Build GRCh38
rs112605710 Chr.9: 76787415-76787415 on Build GRCh38
rs530641049 Chr.9: 76787344-76787344 on Build GRCh38
rs989262761 Chr.9: 76787303-76787303 on Build GRCh38
rs4317645 Chr.9: 76787530-76787530 on Build GRCh38
rs146929015 Chr.9: 76787387-76787387 on Build GRCh38
rs12235152 Chr.9: 76787558-76787558 on Build GRCh38
rs573067390 Chr.9: 76787658-76787658 on Build GRCh38
rs1048300827 Chr.9: 76787611-76787611 on Build GRCh38
rs933910303 Chr.9: 76787601-76787601 on Build GRCh38
rs956437871 Chr.9: 76787543-76787543 on Build GRCh38
rs899736627 Chr.9: 76787687-76787687 on Build GRCh38
rs4585807 Chr.9: 76787377-76787377 on Build GRCh38
rs1047009943 Chr.9: 76787612-76787612 on Build GRCh38
rs890811225 Chr.9: 76787730-76787730 on Build GRCh38
rs4631539 Chr.9: 76787401-76787401 on Build GRCh38
rs397707010 Chr.9: 76787640-76787641 on Build GRCh38
rs138034175 Chr.9: 76787266-76787268 on Build GRCh38
rs200803230 Chr.9: 76787306-76787307 on Build GRCh38
rs756998586 Chr.9: 76787366-76787366 on Build GRCh38
rs1046864256 Chr.9: 76787330-76787330 on Build GRCh38
rs972056927 Chr.9: 76787517-76787517 on Build GRCh38
rs574562941 Chr.9: 76787520-76787520 on Build GRCh38
rs115877995 Chr.9: 76787394-76787394 on Build GRCh38
rs938026047 Chr.9: 76787291-76787291 on Build GRCh38
rs561691108 Chr.9: 76787721-76787721 on Build GRCh38
rs150908895 Chr.9: 76787663-76787663 on Build GRCh38
rs879641509 Chr.9: 76787293-76787293 on Build GRCh38
rs887082119 Chr.9: 76787389-76787389 on Build GRCh38
rs756044527 Chr.9: 76787304-76787306 on Build GRCh38
rs5003618 Chr.9: 76787300-76787300 on Build GRCh38

View Full Product Details