Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 87008872-87009395 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC440173
Entrez Gene ID
440173
Gene Name
uncharacterized LOC440173
Gene Aliases
-
UniGene
Hs.127361
Gene Chromosome Location
Chr.9: 87008451-87042126 on Build GRCh38
Transcripts
NR_027471.1

SNP Detail

SNP ID Chromosome Location
rs955398512 Chr.9: 87009184-87009185 on Build GRCh38
rs938297131 Chr.9: 87009360-87009360 on Build GRCh38
rs893778656 Chr.9: 87009129-87009129 on Build GRCh38
rs780515664 Chr.9: 87009358-87009358 on Build GRCh38
rs969818403 Chr.9: 87009303-87009303 on Build GRCh38
rs911964834 Chr.9: 87008957-87008957 on Build GRCh38
rs959766229 Chr.9: 87009340-87009340 on Build GRCh38
rs572047695 Chr.9: 87008896-87008896 on Build GRCh38
rs974230357 Chr.9: 87009230-87009230 on Build GRCh38
rs919653393 Chr.9: 87008956-87008956 on Build GRCh38
rs756520705 Chr.9: 87009161-87009161 on Build GRCh38
rs189414541 Chr.9: 87008952-87008952 on Build GRCh38
rs1008201783 Chr.9: 87009185-87009187 on Build GRCh38
rs559987421 Chr.9: 87009252-87009252 on Build GRCh38
rs1021878829 Chr.9: 87009096-87009096 on Build GRCh38
rs767399411 Chr.9: 87009157-87009166 on Build GRCh38
rs906895056 Chr.9: 87008963-87008963 on Build GRCh38
rs543378073 Chr.9: 87009077-87009077 on Build GRCh38
rs1052881665 Chr.9: 87009125-87009125 on Build GRCh38
rs1003660566 Chr.9: 87009277-87009277 on Build GRCh38
rs1052999907 Chr.9: 87008965-87008965 on Build GRCh38
rs745630522 Chr.9: 87009199-87009199 on Build GRCh38
rs1011374902 Chr.9: 87009071-87009071 on Build GRCh38
rs1044075582 Chr.9: 87008959-87008959 on Build GRCh38
rs961939964 Chr.9: 87009214-87009214 on Build GRCh38
rs1051881789 Chr.9: 87008901-87008901 on Build GRCh38
rs528957447 Chr.9: 87009098-87009098 on Build GRCh38
rs768914119 Chr.9: 87009332-87009332 on Build GRCh38
rs138630472 Chr.9: 87009165-87009165 on Build GRCh38
rs903933169 Chr.9: 87009106-87009106 on Build GRCh38
rs557762134 Chr.9: 87009009-87009009 on Build GRCh38
rs996921866 Chr.9: 87009109-87009109 on Build GRCh38
rs563240991 Chr.9: 87009087-87009087 on Build GRCh38
rs940181912 Chr.9: 87008903-87008903 on Build GRCh38
rs528824747 Chr.9: 87009307-87009307 on Build GRCh38
rs1045282415 Chr.9: 87009211-87009211 on Build GRCh38
rs938559123 Chr.9: 87008964-87008964 on Build GRCh38
rs905398196 Chr.9: 87009232-87009232 on Build GRCh38
rs935342449 Chr.9: 87008998-87008998 on Build GRCh38
rs577550153 Chr.9: 87009014-87009014 on Build GRCh38
rs551774960 Chr.9: 87009362-87009362 on Build GRCh38
rs920129687 Chr.9: 87009259-87009259 on Build GRCh38
rs891719213 Chr.9: 87009068-87009068 on Build GRCh38

View Full Product Details