Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 50008759-50009258 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLCN5
Entrez Gene ID
1184
Gene Name
chloride voltage-gated channel 5
Gene Aliases
CLC5, CLCK2, ClC-5, DENTS, NPHL1, NPHL2, XLRH, XRN, hCIC-K2
UniGene
Hs.166486
Gene Chromosome Location
Chr.X: 49922615-50099235 on Build GRCh38
Transcripts
NM_001127898.3 NM_001127899.3 NM_001272102.1 XM_017029257.1 XM_017029258.1 NR_029850.1

Gene Symbol
MIR362
Entrez Gene ID
574030
Gene Name
microRNA 362
Gene Aliases
MIRN362, hsa-mir-362, mir-362
UniGene
Gene Chromosome Location
Chr.X: 50008964-50009028 on Build GRCh38
Transcripts
NM_001127898.3 NM_001127899.3 NM_001272102.1 XM_017029257.1 XM_017029258.1 NR_029850.1

SNP Detail

SNP ID Chromosome Location
rs1048552733 Chr.X: 50009006-50009006 on Build GRCh38
rs370341989 Chr.X: 50009102-50009102 on Build GRCh38
rs374141752 Chr.X: 50008989-50008989 on Build GRCh38
rs959899814 Chr.X: 50009138-50009138 on Build GRCh38
rs368034272 Chr.X: 50009071-50009071 on Build GRCh38
rs886748399 Chr.X: 50009082-50009082 on Build GRCh38
rs781842604 Chr.X: 50008976-50008976 on Build GRCh38
rs781899067 Chr.X: 50008822-50008822 on Build GRCh38
rs782805535 Chr.X: 50008931-50008931 on Build GRCh38
rs782167423 Chr.X: 50008922-50008922 on Build GRCh38
rs1004397043 Chr.X: 50009098-50009098 on Build GRCh38
rs782452229 Chr.X: 50009226-50009226 on Build GRCh38
rs900099694 Chr.X: 50008862-50008862 on Build GRCh38
rs73490086 Chr.X: 50008959-50008959 on Build GRCh38
rs1039965086 Chr.X: 50008827-50008827 on Build GRCh38
rs781853309 Chr.X: 50009179-50009179 on Build GRCh38
rs782143793 Chr.X: 50008924-50008924 on Build GRCh38
rs782272417 Chr.X: 50009207-50009207 on Build GRCh38
rs149596 Chr.X: 50009197-50009197 on Build GRCh38
rs782518683 Chr.X: 50009182-50009182 on Build GRCh38
rs190696903 Chr.X: 50009234-50009234 on Build GRCh38
rs782438731 Chr.X: 50008856-50008856 on Build GRCh38
rs376832657 Chr.X: 50008923-50008923 on Build GRCh38
rs781978006 Chr.X: 50008946-50008946 on Build GRCh38
rs781797185 Chr.X: 50008995-50008995 on Build GRCh38
rs782748998 Chr.X: 50008975-50008975 on Build GRCh38
rs782611023 Chr.X: 50009107-50009107 on Build GRCh38

View Full Product Details