Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 73796427-73796954 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TSIX
Entrez Gene ID
9383
Gene Name
TSIX transcript, XIST antisense RNA
Gene Aliases
LINC00013, NCRNA00013, XIST-AS, XIST-AS1, XISTAS
UniGene
Hs.529901
Gene Chromosome Location
Chr.X: 73792205-73829231 on Build GRCh38
Transcripts
NR_003255.2

SNP Detail

SNP ID Chromosome Location
rs140010072 Chr.X: 73796767-73796767 on Build GRCh38
rs745579414 Chr.X: 73796535-73796535 on Build GRCh38
rs941964132 Chr.X: 73796735-73796735 on Build GRCh38
rs992175524 Chr.X: 73796853-73796853 on Build GRCh38
rs894297082 Chr.X: 73796727-73796727 on Build GRCh38
rs376719695 Chr.X: 73796837-73796837 on Build GRCh38
rs367659911 Chr.X: 73796454-73796454 on Build GRCh38
rs769070359 Chr.X: 73796657-73796657 on Build GRCh38
rs187774084 Chr.X: 73796842-73796842 on Build GRCh38
rs990398150 Chr.X: 73796541-73796541 on Build GRCh38
rs1018421544 Chr.X: 73796824-73796824 on Build GRCh38
rs946083127 Chr.X: 73796565-73796565 on Build GRCh38
rs964293954 Chr.X: 73796505-73796505 on Build GRCh38
rs908954672 Chr.X: 73796592-73796592 on Build GRCh38
rs191179897 Chr.X: 73796661-73796661 on Build GRCh38
rs1042077327 Chr.X: 73796599-73796599 on Build GRCh38
rs903928680 Chr.X: 73796795-73796795 on Build GRCh38
rs993182282 Chr.X: 73796784-73796784 on Build GRCh38
rs983292491 Chr.X: 73796580-73796580 on Build GRCh38
rs777411074 Chr.X: 73796874-73796874 on Build GRCh38
rs1055670647 Chr.X: 73796722-73796722 on Build GRCh38
rs746491877 Chr.X: 73796613-73796613 on Build GRCh38
rs1009681488 Chr.X: 73796763-73796763 on Build GRCh38
rs754681747 Chr.X: 73796804-73796804 on Build GRCh38
rs748463143 Chr.X: 73796680-73796680 on Build GRCh38
rs933016153 Chr.X: 73796662-73796662 on Build GRCh38
rs1037640845 Chr.X: 73796742-73796742 on Build GRCh38
rs977458470 Chr.X: 73796890-73796890 on Build GRCh38
rs891384775 Chr.X: 73796715-73796715 on Build GRCh38
rs766676336 Chr.X: 73796825-73796825 on Build GRCh38
rs941801158 Chr.X: 73796639-73796640 on Build GRCh38
rs749221824 Chr.X: 73796587-73796587 on Build GRCh38
rs371070864 Chr.X: 73796599-73796600 on Build GRCh38
rs1051743684 Chr.X: 73796671-73796672 on Build GRCh38
rs773696432 Chr.X: 73796816-73796816 on Build GRCh38
rs914786034 Chr.X: 73796545-73796545 on Build GRCh38
rs183342000 Chr.X: 73796820-73796820 on Build GRCh38
rs758027465 Chr.X: 73796546-73796546 on Build GRCh38
rs1038680864 Chr.X: 73796648-73796648 on Build GRCh38
rs187453338 Chr.X: 73796601-73796601 on Build GRCh38
rs1076875 Chr.X: 73796841-73796841 on Build GRCh38

View Full Product Details