Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 39513901-39514466 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAM3A
Entrez Gene ID
1587
Gene Name
ADAM metallopeptidase domain 3A (pseudogene)
Gene Aliases
ADAM3, CYRN1, tMDCI
UniGene
Hs.98848
Gene Chromosome Location
Chr.8: 39451045-39522989 on Build GRCh38
Transcripts
NR_001569.3 NR_024106.2 NR_024107.2 NR_073423.1

SNP Detail

SNP ID Chromosome Location
rs927808099 Chr.8: 39514022-39514022 on Build GRCh38
rs79143343 Chr.8: 39514210-39514210 on Build GRCh38
rs889370773 Chr.8: 39513956-39513956 on Build GRCh38
rs890631696 Chr.8: 39514429-39514429 on Build GRCh38
rs201444785 Chr.8: 39514118-39514118 on Build GRCh38
rs533342935 Chr.8: 39514322-39514322 on Build GRCh38
rs386724643 Chr.8: 39514118-39514119 on Build GRCh38
rs542682276 Chr.8: 39514442-39514442 on Build GRCh38
rs369722299 Chr.8: 39514259-39514259 on Build GRCh38
rs11280925 Chr.8: 39514209-39514210 on Build GRCh38
rs748391115 Chr.8: 39514082-39514082 on Build GRCh38
rs939159152 Chr.8: 39514042-39514042 on Build GRCh38
rs202041548 Chr.8: 39514207-39514208 on Build GRCh38
rs71237172 Chr.8: 39514115-39514116 on Build GRCh38
rs533976788 Chr.8: 39514364-39514364 on Build GRCh38
rs374707945 Chr.8: 39514387-39514387 on Build GRCh38
rs983470241 Chr.8: 39513973-39513973 on Build GRCh38
rs962217056 Chr.8: 39514285-39514285 on Build GRCh38
rs575079105 Chr.8: 39514046-39514048 on Build GRCh38
rs1009116914 Chr.8: 39514097-39514098 on Build GRCh38
rs946404351 Chr.8: 39514192-39514192 on Build GRCh38
rs372209490 Chr.8: 39514119-39514119 on Build GRCh38
rs902158048 Chr.8: 39514353-39514353 on Build GRCh38
rs7844681 Chr.8: 39514424-39514424 on Build GRCh38
rs192619272 Chr.8: 39514238-39514238 on Build GRCh38
rs374422810 Chr.8: 39514203-39514203 on Build GRCh38
rs1043394586 Chr.8: 39514236-39514236 on Build GRCh38
rs538151169 Chr.8: 39514094-39514094 on Build GRCh38
rs371996688 Chr.8: 39514441-39514441 on Build GRCh38
rs368861774 Chr.8: 39514204-39514204 on Build GRCh38
rs1019548640 Chr.8: 39514121-39514121 on Build GRCh38
rs144922900 Chr.8: 39514204-39514205 on Build GRCh38
rs75834925 Chr.8: 39514115-39514115 on Build GRCh38

View Full Product Details