Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 55452380-55452927 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SBF1P1
Entrez Gene ID
100133234
Gene Name
SET binding factor 1 pseudogene 1
Gene Aliases
-
UniGene
Hs.729852
Gene Chromosome Location
Chr.8: 55449197-55455321 on Build GRCh38
Transcripts
NR_027765.2 NM_052898.1

Gene Symbol
XKR4
Entrez Gene ID
114786
Gene Name
XK related 4
Gene Aliases
XRG4
UniGene
Hs.130197
Gene Chromosome Location
Chr.8: 55102457-55526155 on Build GRCh38
Transcripts
NR_027765.2 NM_052898.1

SNP Detail

SNP ID Chromosome Location
rs934415723 Chr.8: 55452471-55452471 on Build GRCh38
rs1047507464 Chr.8: 55452405-55452405 on Build GRCh38
rs747056581 Chr.8: 55452752-55452752 on Build GRCh38
rs552467284 Chr.8: 55452895-55452895 on Build GRCh38
rs1027384749 Chr.8: 55452897-55452897 on Build GRCh38
rs113077660 Chr.8: 55452550-55452550 on Build GRCh38
rs182820502 Chr.8: 55452577-55452577 on Build GRCh38
rs758897971 Chr.8: 55452701-55452701 on Build GRCh38
rs543050204 Chr.8: 55452726-55452726 on Build GRCh38
rs996699036 Chr.8: 55452433-55452433 on Build GRCh38
rs892838606 Chr.8: 55452472-55452472 on Build GRCh38
rs968873957 Chr.8: 55452888-55452888 on Build GRCh38
rs188340843 Chr.8: 55452587-55452587 on Build GRCh38
rs992891573 Chr.8: 55452466-55452466 on Build GRCh38
rs904596894 Chr.8: 55452806-55452806 on Build GRCh38
rs897019084 Chr.8: 55452783-55452783 on Build GRCh38
rs147358665 Chr.8: 55452569-55452569 on Build GRCh38
rs932638585 Chr.8: 55452794-55452794 on Build GRCh38
rs968629762 Chr.8: 55452451-55452451 on Build GRCh38
rs969789351 Chr.8: 55452665-55452665 on Build GRCh38
rs959769458 Chr.8: 55452458-55452458 on Build GRCh38
rs1031118027 Chr.8: 55452838-55452838 on Build GRCh38
rs148409670 Chr.8: 55452581-55452581 on Build GRCh38
rs895704309 Chr.8: 55452614-55452614 on Build GRCh38
rs537000201 Chr.8: 55452753-55452753 on Build GRCh38
rs922990810 Chr.8: 55452463-55452463 on Build GRCh38
rs867532 Chr.8: 55452551-55452551 on Build GRCh38
rs551840183 Chr.8: 55452890-55452890 on Build GRCh38
rs921224869 Chr.8: 55452793-55452793 on Build GRCh38
rs1035746087 Chr.8: 55452438-55452438 on Build GRCh38
rs574135871 Chr.8: 55452603-55452604 on Build GRCh38
rs755284929 Chr.8: 55452489-55452489 on Build GRCh38
rs998429583 Chr.8: 55452828-55452828 on Build GRCh38
rs7000644 Chr.8: 55452482-55452482 on Build GRCh38
rs528724792 Chr.8: 55452846-55452846 on Build GRCh38
rs551447903 Chr.8: 55452576-55452576 on Build GRCh38
rs564117810 Chr.8: 55452448-55452448 on Build GRCh38
rs905785462 Chr.8: 55452539-55452539 on Build GRCh38
rs73591686 Chr.8: 55452455-55452455 on Build GRCh38
rs555552637 Chr.8: 55452609-55452609 on Build GRCh38
rs577210650 Chr.8: 55452724-55452724 on Build GRCh38
rs899897398 Chr.8: 55452432-55452432 on Build GRCh38
rs540634563 Chr.8: 55452444-55452444 on Build GRCh38
rs139618671 Chr.8: 55452716-55452716 on Build GRCh38
rs562790910 Chr.8: 55452850-55452850 on Build GRCh38
rs1041164316 Chr.8: 55452425-55452425 on Build GRCh38
rs1014555462 Chr.8: 55452882-55452882 on Build GRCh38
rs1057094135 Chr.8: 55452893-55452893 on Build GRCh38
rs776875215 Chr.8: 55452879-55452879 on Build GRCh38
rs1022559242 Chr.8: 55452640-55452640 on Build GRCh38
rs1027027083 Chr.8: 55452446-55452446 on Build GRCh38
rs572127073 Chr.8: 55452423-55452423 on Build GRCh38
rs1041418281 Chr.8: 55452564-55452564 on Build GRCh38
rs1022737001 Chr.8: 55452487-55452487 on Build GRCh38
rs1018695584 Chr.8: 55452801-55452801 on Build GRCh38
rs931478323 Chr.8: 55452527-55452527 on Build GRCh38
rs899929997 Chr.8: 55452568-55452568 on Build GRCh38
rs920053743 Chr.8: 55452525-55452525 on Build GRCh38
rs914206006 Chr.8: 55452646-55452646 on Build GRCh38
rs1027350936 Chr.8: 55452694-55452694 on Build GRCh38
rs559908413 Chr.8: 55452825-55452825 on Build GRCh38
rs931537289 Chr.8: 55452404-55452404 on Build GRCh38
rs887154125 Chr.8: 55452415-55452415 on Build GRCh38
rs988552083 Chr.8: 55452623-55452623 on Build GRCh38
rs542822376 Chr.8: 55452796-55452796 on Build GRCh38
rs944257352 Chr.8: 55452759-55452759 on Build GRCh38
rs117684375 Chr.8: 55452545-55452545 on Build GRCh38
rs961459807 Chr.8: 55452441-55452441 on Build GRCh38
rs973165310 Chr.8: 55452667-55452667 on Build GRCh38
rs112581252 Chr.8: 55452585-55452585 on Build GRCh38
rs1056977940 Chr.8: 55452769-55452769 on Build GRCh38
rs979788200 Chr.8: 55452889-55452889 on Build GRCh38
rs150598763 Chr.8: 55452636-55452636 on Build GRCh38
rs1013940779 Chr.8: 55452627-55452627 on Build GRCh38
rs1042915158 Chr.8: 55452578-55452578 on Build GRCh38
rs570041480 Chr.8: 55452518-55452518 on Build GRCh38
rs950493098 Chr.8: 55452901-55452901 on Build GRCh38
rs2975982 Chr.8: 55452891-55452891 on Build GRCh38
rs186201810 Chr.8: 55452452-55452452 on Build GRCh38
rs552451421 Chr.8: 55452795-55452795 on Build GRCh38
rs994234738 Chr.8: 55452786-55452786 on Build GRCh38
rs559567301 Chr.8: 55452555-55452555 on Build GRCh38
rs535265939 Chr.8: 55452593-55452593 on Build GRCh38
rs911361507 Chr.8: 55452454-55452454 on Build GRCh38
rs2929048 Chr.8: 55452760-55452760 on Build GRCh38
rs373304428 Chr.8: 55452797-55452797 on Build GRCh38
rs534718708 Chr.8: 55452695-55452695 on Build GRCh38
rs980920238 Chr.8: 55452522-55452522 on Build GRCh38
rs773402189 Chr.8: 55452727-55452727 on Build GRCh38
rs969756778 Chr.8: 55452499-55452499 on Build GRCh38
rs563680929 Chr.8: 55452528-55452528 on Build GRCh38
rs895747905 Chr.8: 55452851-55452851 on Build GRCh38

View Full Product Details