Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.Y: 19604450-19605045 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TXLNGY
Entrez Gene ID
246126
Gene Name
taxilin gamma pseudogene, Y-linked
Gene Aliases
CYorf15A, CYorf15B, TXLNG2P
UniGene
Hs.522863
Gene Chromosome Location
Chr.Y: 19567358-19607170 on Build GRCh38
Transcripts
NR_045128.1

SNP Detail

SNP ID Chromosome Location
rs995555119 Chr.Y: 19604950-19604950 on Build GRCh38
rs757727917 Chr.Y: 19604532-19604532 on Build GRCh38
rs766591331 Chr.Y: 19604648-19604653 on Build GRCh38
rs202123330 Chr.Y: 19604638-19604638 on Build GRCh38
rs75254362 Chr.Y: 19604580-19604580 on Build GRCh38
rs36033553 Chr.Y: 19604685-19604686 on Build GRCh38
rs867852110 Chr.Y: 19604686-19604686 on Build GRCh38

View Full Product Details