Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 15889-16440 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WASH7P
Entrez Gene ID
653635
Gene Name
WAS protein family homolog 7 pseudogene
Gene Aliases
FAM39F, WASH5P
UniGene
Hs.446466
Gene Chromosome Location
Chr.1: 14362-29370 on Build GRCh38
Transcripts
NR_024540.1

SNP Detail

SNP ID Chromosome Location
rs775449456 Chr.1: 16187-16187 on Build GRCh38
rs377597122 Chr.1: 16308-16308 on Build GRCh38
rs866639523 Chr.1: 16280-16280 on Build GRCh38
rs4005864 Chr.1: 16112-16112 on Build GRCh38
rs4011288 Chr.1: 16299-16299 on Build GRCh38
rs920943684 Chr.1: 16117-16117 on Build GRCh38
rs978752438 Chr.1: 15986-15986 on Build GRCh38
rs1049085958 Chr.1: 16278-16278 on Build GRCh38
rs113141985 Chr.1: 16288-16288 on Build GRCh38
rs565634052 Chr.1: 16250-16250 on Build GRCh38
rs2385982 Chr.1: 16009-16009 on Build GRCh38
rs930763425 Chr.1: 16344-16344 on Build GRCh38
rs4011289 Chr.1: 16311-16311 on Build GRCh38
rs561144608 Chr.1: 16160-16160 on Build GRCh38
rs999184390 Chr.1: 16390-16390 on Build GRCh38
rs201230158 Chr.1: 16240-16241 on Build GRCh38
rs545118963 Chr.1: 16051-16051 on Build GRCh38
rs1010661750 Chr.1: 16376-16376 on Build GRCh38
rs1005151435 Chr.1: 16288-16288 on Build GRCh38
rs571417824 Chr.1: 16181-16181 on Build GRCh38
rs556270939 Chr.1: 16032-16032 on Build GRCh38
rs199651039 Chr.1: 15911-15911 on Build GRCh38
rs915196349 Chr.1: 16143-16143 on Build GRCh38
rs1042444118 Chr.1: 16045-16045 on Build GRCh38
rs878899929 Chr.1: 16336-16336 on Build GRCh38
rs966345447 Chr.1: 16389-16389 on Build GRCh38
rs577706668 Chr.1: 16048-16048 on Build GRCh38
rs1053598232 Chr.1: 16020-16020 on Build GRCh38
rs573463475 Chr.1: 15972-15972 on Build GRCh38
rs200783265 Chr.1: 15996-15996 on Build GRCh38
rs960522484 Chr.1: 16342-16342 on Build GRCh38
rs200402379 Chr.1: 15975-15975 on Build GRCh38
rs549993963 Chr.1: 16173-16173 on Build GRCh38
rs555553763 Chr.1: 15974-15974 on Build GRCh38
rs867839637 Chr.1: 16151-16151 on Build GRCh38
rs2747966 Chr.1: 16298-16298 on Build GRCh38
rs773430888 Chr.1: 16146-16146 on Build GRCh38
rs71224536 Chr.1: 16186-16186 on Build GRCh38
rs79687254 Chr.1: 16172-16172 on Build GRCh38
rs989800783 Chr.1: 16142-16142 on Build GRCh38
rs4005863 Chr.1: 16131-16131 on Build GRCh38
rs945312137 Chr.1: 16237-16237 on Build GRCh38
rs4024958 Chr.1: 16072-16072 on Build GRCh38
rs114451402 Chr.1: 16328-16328 on Build GRCh38
rs932552237 Chr.1: 16138-16138 on Build GRCh38
rs567583108 Chr.1: 16184-16184 on Build GRCh38
rs578034443 Chr.1: 16303-16303 on Build GRCh38
rs3877232 Chr.1: 16147-16147 on Build GRCh38
rs953842574 Chr.1: 16310-16310 on Build GRCh38
rs764852654 Chr.1: 16059-16059 on Build GRCh38
rs879223435 Chr.1: 16380-16380 on Build GRCh38
rs9645376 Chr.1: 15954-15954 on Build GRCh38
rs77838826 Chr.1: 16300-16300 on Build GRCh38
rs4005888 Chr.1: 16220-16220 on Build GRCh38
rs879271035 Chr.1: 16128-16128 on Build GRCh38
rs544021564 Chr.1: 16127-16127 on Build GRCh38
rs4011287 Chr.1: 16297-16297 on Build GRCh38
rs1049503175 Chr.1: 16346-16346 on Build GRCh38
rs372319358 Chr.1: 16068-16068 on Build GRCh38
rs111404672 Chr.1: 16415-16415 on Build GRCh38
rs907885049 Chr.1: 16349-16349 on Build GRCh38
rs906600682 Chr.1: 16170-16170 on Build GRCh38
rs9308534 Chr.1: 16188-16188 on Build GRCh38
rs959619973 Chr.1: 16085-16085 on Build GRCh38
rs370936929 Chr.1: 16086-16086 on Build GRCh38
rs9308533 Chr.1: 16110-16110 on Build GRCh38
rs953834333 Chr.1: 16209-16209 on Build GRCh38
rs982534139 Chr.1: 15969-15969 on Build GRCh38
rs77747673 Chr.1: 16337-16337 on Build GRCh38
rs375149030 Chr.1: 16347-16347 on Build GRCh38
rs541172944 Chr.1: 16071-16071 on Build GRCh38
rs1015340985 Chr.1: 16107-16107 on Build GRCh38
rs1055616909 Chr.1: 16248-16248 on Build GRCh38
rs112448831 Chr.1: 15956-15956 on Build GRCh38
rs893632644 Chr.1: 16043-16043 on Build GRCh38
rs3955605 Chr.1: 16277-16277 on Build GRCh38
rs563966321 Chr.1: 16140-16140 on Build GRCh38
rs560448997 Chr.1: 16065-16065 on Build GRCh38
rs10442898 Chr.1: 16026-16026 on Build GRCh38
rs534467406 Chr.1: 15973-15973 on Build GRCh38
rs1034888660 Chr.1: 16399-16399 on Build GRCh38
rs996729891 Chr.1: 16369-16369 on Build GRCh38
rs886667411 Chr.1: 16282-16282 on Build GRCh38
rs113743222 Chr.1: 16054-16054 on Build GRCh38
rs940884534 Chr.1: 16355-16355 on Build GRCh38
rs182023638 Chr.1: 16016-16016 on Build GRCh38
rs201325378 Chr.1: 16015-16015 on Build GRCh38
rs976507462 Chr.1: 16194-16194 on Build GRCh38
rs573865431 Chr.1: 16005-16005 on Build GRCh38
rs80224649 Chr.1: 16125-16125 on Build GRCh38
rs4005866 Chr.1: 15913-15913 on Build GRCh38
rs6660632 Chr.1: 16089-16089 on Build GRCh38
rs925439668 Chr.1: 15999-15999 on Build GRCh38
rs578123551 Chr.1: 16109-16109 on Build GRCh38
rs531570980 Chr.1: 16168-16168 on Build GRCh38
rs879072284 Chr.1: 15952-15952 on Build GRCh38
rs375964566 Chr.1: 15922-15922 on Build GRCh38
rs538298689 Chr.1: 15940-15940 on Build GRCh38
rs939397358 Chr.1: 16245-16245 on Build GRCh38
rs6701319 Chr.1: 16084-16084 on Build GRCh38
rs1003478706 Chr.1: 16066-16066 on Build GRCh38
rs78376469 Chr.1: 16103-16103 on Build GRCh38
rs548165136 Chr.1: 16142-16142 on Build GRCh38
rs757777347 Chr.1: 16375-16375 on Build GRCh38
rs866100273 Chr.1: 16098-16098 on Build GRCh38
rs994889227 Chr.1: 16180-16180 on Build GRCh38
rs9970088 Chr.1: 16262-16262 on Build GRCh38
rs74462471 Chr.1: 16319-16319 on Build GRCh38
rs76789111 Chr.1: 16111-16111 on Build GRCh38
rs527608257 Chr.1: 16079-16079 on Build GRCh38
rs915124736 Chr.1: 16213-16213 on Build GRCh38
rs777777570 Chr.1: 16385-16385 on Build GRCh38
rs17855870 Chr.1: 15912-15912 on Build GRCh38
rs950947302 Chr.1: 16093-16093 on Build GRCh38
rs930628768 Chr.1: 16276-16276 on Build GRCh38
rs553729152 Chr.1: 15950-15950 on Build GRCh38
rs148220436 Chr.1: 16378-16378 on Build GRCh38
rs4005862 Chr.1: 16140-16140 on Build GRCh38
rs564675576 Chr.1: 16091-16091 on Build GRCh38
rs989189587 Chr.1: 16212-16212 on Build GRCh38
rs966563359 Chr.1: 16330-16330 on Build GRCh38
rs976391880 Chr.1: 16110-16110 on Build GRCh38
rs572465511 Chr.1: 16042-16042 on Build GRCh38
rs796132385 Chr.1: 16320-16320 on Build GRCh38
rs1009440411 Chr.1: 16044-16044 on Build GRCh38
rs3877230 Chr.1: 16099-16099 on Build GRCh38
rs977980248 Chr.1: 16336-16336 on Build GRCh38
rs4005865 Chr.1: 16067-16067 on Build GRCh38
rs200655866 Chr.1: 16244-16244 on Build GRCh38
rs868360655 Chr.1: 16164-16164 on Build GRCh38
rs377417848 Chr.1: 15937-15937 on Build GRCh38
rs200010825 Chr.1: 16201-16201 on Build GRCh38
rs6541501 Chr.1: 16010-16010 on Build GRCh38
rs77165470 Chr.1: 16176-16176 on Build GRCh38
rs555665869 Chr.1: 16268-16268 on Build GRCh38
rs889503296 Chr.1: 16374-16374 on Build GRCh38
rs71292708 Chr.1: 16107-16107 on Build GRCh38
rs549960873 Chr.1: 16252-16252 on Build GRCh38
rs958192433 Chr.1: 16010-16013 on Build GRCh38
rs991039103 Chr.1: 16407-16407 on Build GRCh38
rs186230301 Chr.1: 16301-16301 on Build GRCh38
rs76714260 Chr.1: 16295-16295 on Build GRCh38
rs1033656814 Chr.1: 16074-16074 on Build GRCh38
rs965097611 Chr.1: 16186-16186 on Build GRCh38
rs529651976 Chr.1: 16141-16141 on Build GRCh38
rs3877231 Chr.1: 16107-16107 on Build GRCh38
rs977863129 Chr.1: 16243-16243 on Build GRCh38
rs755466349 Chr.1: 16227-16227 on Build GRCh38
rs115927992 Chr.1: 16296-16296 on Build GRCh38
rs752267461 Chr.1: 16363-16363 on Build GRCh38
rs966962310 Chr.1: 15978-15978 on Build GRCh38
rs532187655 Chr.1: 16203-16203 on Build GRCh38
rs534245681 Chr.1: 16256-16256 on Build GRCh38
rs1040762304 Chr.1: 15987-15987 on Build GRCh38
rs902125053 Chr.1: 16364-16364 on Build GRCh38
rs199889494 Chr.1: 16387-16387 on Build GRCh38
rs1022338060 Chr.1: 16388-16388 on Build GRCh38
rs921164171 Chr.1: 16001-16001 on Build GRCh38
rs191082466 Chr.1: 16402-16402 on Build GRCh38
rs11489794 Chr.1: 16257-16257 on Build GRCh38
rs3877229 Chr.1: 15964-15964 on Build GRCh38
rs561670553 Chr.1: 16365-16365 on Build GRCh38
rs75082847 Chr.1: 16014-16014 on Build GRCh38
rs139793388 Chr.1: 16017-16018 on Build GRCh38
rs916636345 Chr.1: 16343-16343 on Build GRCh38

View Full Product Details