Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 56644010-56644558 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MT1DP
Entrez Gene ID
326343
Gene Name
metallothionein 1D, pseudogene
Gene Aliases
MTM
UniGene
Hs.679600
Gene Chromosome Location
Chr.16: 56643687-56644941 on Build GRCh38
Transcripts
NR_003658.2 NR_027781.1

SNP Detail

SNP ID Chromosome Location
rs57179692 Chr.16: 56644451-56644451 on Build GRCh38
rs917227230 Chr.16: 56644273-56644273 on Build GRCh38
rs906104819 Chr.16: 56644387-56644387 on Build GRCh38
rs1009911874 Chr.16: 56644469-56644469 on Build GRCh38
rs774164720 Chr.16: 56644183-56644183 on Build GRCh38
rs573762532 Chr.16: 56644044-56644044 on Build GRCh38
rs571804424 Chr.16: 56644228-56644228 on Build GRCh38
rs952000419 Chr.16: 56644225-56644225 on Build GRCh38
rs115648467 Chr.16: 56644031-56644031 on Build GRCh38
rs550229840 Chr.16: 56644219-56644219 on Build GRCh38
rs566083316 Chr.16: 56644355-56644355 on Build GRCh38
rs573776362 Chr.16: 56644448-56644448 on Build GRCh38
rs1044584196 Chr.16: 56644095-56644095 on Build GRCh38
rs192579139 Chr.16: 56644212-56644212 on Build GRCh38
rs868821374 Chr.16: 56644143-56644143 on Build GRCh38
rs562289446 Chr.16: 56644055-56644055 on Build GRCh38
rs561525494 Chr.16: 56644214-56644214 on Build GRCh38
rs534051698 Chr.16: 56644365-56644365 on Build GRCh38
rs555414836 Chr.16: 56644381-56644381 on Build GRCh38
rs115208582 Chr.16: 56644210-56644210 on Build GRCh38
rs147894818 Chr.16: 56644483-56644483 on Build GRCh38
rs1009859449 Chr.16: 56644124-56644124 on Build GRCh38
rs531861407 Chr.16: 56644215-56644215 on Build GRCh38
rs61135921 Chr.16: 56644099-56644099 on Build GRCh38
rs189118997 Chr.16: 56644046-56644046 on Build GRCh38
rs1029359432 Chr.16: 56644508-56644508 on Build GRCh38
rs555493088 Chr.16: 56644036-56644036 on Build GRCh38
rs1037044119 Chr.16: 56644266-56644266 on Build GRCh38
rs577281355 Chr.16: 56644484-56644484 on Build GRCh38
rs375821525 Chr.16: 56644406-56644406 on Build GRCh38
rs547785845 Chr.16: 56644315-56644315 on Build GRCh38
rs1018529611 Chr.16: 56644193-56644193 on Build GRCh38
rs150353189 Chr.16: 56644243-56644243 on Build GRCh38
rs1018583274 Chr.16: 56644473-56644473 on Build GRCh38
rs760171949 Chr.16: 56644524-56644524 on Build GRCh38
rs549423182 Chr.16: 56644213-56644213 on Build GRCh38
rs113733152 Chr.16: 56644244-56644244 on Build GRCh38
rs767158058 Chr.16: 56644435-56644435 on Build GRCh38
rs770780875 Chr.16: 56644115-56644115 on Build GRCh38
rs112801948 Chr.16: 56644098-56644098 on Build GRCh38
rs950350140 Chr.16: 56644341-56644341 on Build GRCh38
rs66564161 Chr.16: 56644113-56644113 on Build GRCh38
rs1045069732 Chr.16: 56644361-56644361 on Build GRCh38

View Full Product Details