Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 129383746-129384290 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPL32P3
Entrez Gene ID
132241
Gene Name
ribosomal protein L32 pseudogene 3
Gene Aliases
-
UniGene
Hs.477537
Gene Chromosome Location
Chr.3: 129382834-129399439 on Build GRCh38
Transcripts
NR_003111.2

SNP Detail

SNP ID Chromosome Location
rs73865420 Chr.3: 129383928-129383928 on Build GRCh38
rs900701720 Chr.3: 129384073-129384073 on Build GRCh38
rs994831934 Chr.3: 129383806-129383806 on Build GRCh38
rs779448231 Chr.3: 129383795-129383795 on Build GRCh38
rs920679074 Chr.3: 129384002-129384002 on Build GRCh38
rs3796397 Chr.3: 129383811-129383811 on Build GRCh38
rs999642973 Chr.3: 129384153-129384153 on Build GRCh38
rs969241228 Chr.3: 129384218-129384218 on Build GRCh38
rs748082594 Chr.3: 129384245-129384245 on Build GRCh38
rs944991333 Chr.3: 129384061-129384061 on Build GRCh38
rs996286055 Chr.3: 129384077-129384077 on Build GRCh38
rs565194257 Chr.3: 129383969-129383969 on Build GRCh38
rs575079058 Chr.3: 129383923-129383923 on Build GRCh38
rs144751202 Chr.3: 129383939-129383939 on Build GRCh38
rs977797360 Chr.3: 129383949-129383949 on Build GRCh38
rs916378603 Chr.3: 129383768-129383768 on Build GRCh38
rs138366961 Chr.3: 129383987-129383987 on Build GRCh38
rs192063574 Chr.3: 129383936-129383936 on Build GRCh38
rs140984747 Chr.3: 129384046-129384046 on Build GRCh38
rs1019076652 Chr.3: 129383849-129383849 on Build GRCh38
rs142664690 Chr.3: 129383794-129383794 on Build GRCh38
rs992116012 Chr.3: 129384187-129384187 on Build GRCh38
rs919733713 Chr.3: 129384241-129384241 on Build GRCh38
rs559217225 Chr.3: 129383780-129383780 on Build GRCh38
rs770175268 Chr.3: 129384198-129384199 on Build GRCh38
rs796339956 Chr.3: 129383821-129383821 on Build GRCh38
rs545845508 Chr.3: 129383956-129383956 on Build GRCh38
rs779539374 Chr.3: 129383986-129383986 on Build GRCh38
rs529816462 Chr.3: 129384160-129384160 on Build GRCh38
rs746168577 Chr.3: 129383864-129383864 on Build GRCh38
rs1047744157 Chr.3: 129384028-129384028 on Build GRCh38
rs1009081743 Chr.3: 129383833-129383833 on Build GRCh38
rs112853945 Chr.3: 129384201-129384202 on Build GRCh38
rs768142842 Chr.3: 129384069-129384069 on Build GRCh38
rs183183208 Chr.3: 129384009-129384009 on Build GRCh38
rs995920302 Chr.3: 129383861-129383861 on Build GRCh38
rs964839176 Chr.3: 129383858-129383858 on Build GRCh38
rs973948784 Chr.3: 129384238-129384238 on Build GRCh38
rs187248484 Chr.3: 129383865-129383865 on Build GRCh38
rs764752886 Chr.3: 129383776-129383776 on Build GRCh38
rs1022855834 Chr.3: 129384079-129384079 on Build GRCh38
rs1040599880 Chr.3: 129384068-129384068 on Build GRCh38
rs1048479450 Chr.3: 129383820-129383820 on Build GRCh38
rs948791503 Chr.3: 129383772-129383772 on Build GRCh38
rs562979809 Chr.3: 129383810-129383810 on Build GRCh38
rs146489949 Chr.3: 129383862-129383862 on Build GRCh38
rs776064203 Chr.3: 129384103-129384103 on Build GRCh38
rs762222761 Chr.3: 129383954-129383954 on Build GRCh38
rs886402339 Chr.3: 129384045-129384045 on Build GRCh38
rs930750173 Chr.3: 129384268-129384268 on Build GRCh38
rs960353387 Chr.3: 129384171-129384171 on Build GRCh38
rs923579931 Chr.3: 129383953-129383953 on Build GRCh38
rs532260951 Chr.3: 129383978-129383978 on Build GRCh38
rs796675960 Chr.3: 129384007-129384007 on Build GRCh38
rs903619991 Chr.3: 129384127-129384127 on Build GRCh38

View Full Product Details