Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 99160736-99161288 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC100507053
Entrez Gene ID
100507053
Gene Name
uncharacterized LOC100507053
Gene Aliases
-
UniGene
Hs.635084
Gene Chromosome Location
Chr.4: 99088857-99301356 on Build GRCh38
Transcripts
NR_037884.1 NR_028270.1

Gene Symbol
PCNAP1
Entrez Gene ID
359806
Gene Name
proliferating cell nuclear antigen pseudogene 1
Gene Aliases
p1PCNA
UniGene
Hs.537400
Gene Chromosome Location
Chr.4: 99160593-99161647 on Build GRCh38
Transcripts
NR_037884.1 NR_028270.1

SNP Detail

SNP ID Chromosome Location
rs191984360 Chr.4: 99161190-99161190 on Build GRCh38
rs1000544730 Chr.4: 99160857-99160857 on Build GRCh38
rs1026623104 Chr.4: 99160817-99160817 on Build GRCh38
rs912091258 Chr.4: 99160786-99160786 on Build GRCh38
rs541597290 Chr.4: 99161026-99161026 on Build GRCh38
rs1009509285 Chr.4: 99160894-99160894 on Build GRCh38
rs912143750 Chr.4: 99161152-99161152 on Build GRCh38
rs559290314 Chr.4: 99160976-99160976 on Build GRCh38
rs1024736530 Chr.4: 99160893-99160893 on Build GRCh38
rs970580783 Chr.4: 99160953-99160953 on Build GRCh38
rs975281536 Chr.4: 99161233-99161233 on Build GRCh38
rs750681934 Chr.4: 99160845-99160845 on Build GRCh38
rs182809955 Chr.4: 99160874-99160874 on Build GRCh38
rs892404864 Chr.4: 99160875-99160875 on Build GRCh38
rs376998076 Chr.4: 99160818-99160818 on Build GRCh38
rs747904613 Chr.4: 99161000-99161000 on Build GRCh38
rs545463246 Chr.4: 99161251-99161251 on Build GRCh38
rs963764961 Chr.4: 99161007-99161007 on Build GRCh38
rs1540053 Chr.4: 99160997-99160997 on Build GRCh38
rs769733240 Chr.4: 99161090-99161090 on Build GRCh38
rs981674457 Chr.4: 99161034-99161034 on Build GRCh38
rs950707558 Chr.4: 99161060-99161060 on Build GRCh38
rs932661956 Chr.4: 99161211-99161211 on Build GRCh38
rs1024882504 Chr.4: 99160909-99160909 on Build GRCh38
rs995159076 Chr.4: 99160800-99160800 on Build GRCh38
rs899541458 Chr.4: 99160840-99160840 on Build GRCh38
rs943532755 Chr.4: 99160816-99160816 on Build GRCh38
rs987716780 Chr.4: 99161150-99161150 on Build GRCh38
rs990378290 Chr.4: 99161121-99161121 on Build GRCh38
rs147512026 Chr.4: 99161147-99161147 on Build GRCh38
rs187095724 Chr.4: 99161016-99161016 on Build GRCh38
rs777878457 Chr.4: 99161259-99161259 on Build GRCh38
rs765950768 Chr.4: 99160960-99160961 on Build GRCh38
rs34743543 Chr.4: 99161211-99161212 on Build GRCh38
rs534848276 Chr.4: 99161046-99161046 on Build GRCh38

View Full Product Details