Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 176156756-176157308 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC643201
Entrez Gene ID
643201
Gene Name
centrosomal protein 192kDa pseudogene
Gene Aliases
-
UniGene
Hs.390285
Gene Chromosome Location
Chr.5: 176143085-176199295 on Build GRCh38
Transcripts
NR_036494.1

SNP Detail

SNP ID Chromosome Location
rs576583510 Chr.5: 176156872-176156873 on Build GRCh38
rs561633609 Chr.5: 176157154-176157154 on Build GRCh38
rs180702867 Chr.5: 176156846-176156846 on Build GRCh38
rs1012347355 Chr.5: 176156962-176156962 on Build GRCh38
rs1001526328 Chr.5: 176157088-176157088 on Build GRCh38
rs765395003 Chr.5: 176156961-176156961 on Build GRCh38
rs186406655 Chr.5: 176156960-176156960 on Build GRCh38
rs73805749 Chr.5: 176157176-176157176 on Build GRCh38
rs116688139 Chr.5: 176157239-176157239 on Build GRCh38
rs1038711485 Chr.5: 176156947-176156947 on Build GRCh38
rs768511061 Chr.5: 176157225-176157225 on Build GRCh38
rs1047526362 Chr.5: 176156905-176156905 on Build GRCh38
rs189736227 Chr.5: 176157046-176157046 on Build GRCh38
rs944095314 Chr.5: 176157251-176157251 on Build GRCh38
rs941715376 Chr.5: 176156945-176156945 on Build GRCh38
rs1052421958 Chr.5: 176156990-176156990 on Build GRCh38
rs560192357 Chr.5: 176157190-176157190 on Build GRCh38
rs540371941 Chr.5: 176157083-176157083 on Build GRCh38
rs908949821 Chr.5: 176156926-176156926 on Build GRCh38
rs1042535339 Chr.5: 176157271-176157271 on Build GRCh38
rs534646346 Chr.5: 176156917-176156917 on Build GRCh38
rs912625377 Chr.5: 176156791-176156791 on Build GRCh38
rs555654802 Chr.5: 176156827-176156827 on Build GRCh38
rs902547151 Chr.5: 176156908-176156908 on Build GRCh38
rs983280960 Chr.5: 176157264-176157264 on Build GRCh38
rs1042377345 Chr.5: 176156861-176156861 on Build GRCh38
rs779994913 Chr.5: 176157045-176157045 on Build GRCh38
rs929101139 Chr.5: 176156873-176156873 on Build GRCh38
rs181846596 Chr.5: 176157062-176157062 on Build GRCh38
rs564914191 Chr.5: 176157213-176157213 on Build GRCh38
rs968773338 Chr.5: 176157063-176157063 on Build GRCh38
rs6871900 Chr.5: 176156959-176156959 on Build GRCh38
rs959313640 Chr.5: 176156802-176156802 on Build GRCh38
rs1001520551 Chr.5: 176156916-176156916 on Build GRCh38
rs114267968 Chr.5: 176157000-176157000 on Build GRCh38
rs992453336 Chr.5: 176156803-176156803 on Build GRCh38
rs954116346 Chr.5: 176157226-176157226 on Build GRCh38
rs537094971 Chr.5: 176157282-176157282 on Build GRCh38
rs997780575 Chr.5: 176156987-176156987 on Build GRCh38
rs926574656 Chr.5: 176156786-176156786 on Build GRCh38
rs961097202 Chr.5: 176157156-176157156 on Build GRCh38
rs912406189 Chr.5: 176157243-176157243 on Build GRCh38
rs545112310 Chr.5: 176156980-176156980 on Build GRCh38
rs1025284374 Chr.5: 176157209-176157209 on Build GRCh38

View Full Product Details