Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 43552763-43553168 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
XRCC1
Entrez Gene ID
7515
Gene Name
X-ray repair cross complementing 1
Gene Aliases
RCC
UniGene
Hs.98493
Gene Chromosome Location
Chr.19: 43543312-43575578 on Build GRCh38
Transcripts
NM_006297.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3823313 c.783A>C p.P261P Chr.19: 43552837-43552837 on Build GRCh38
5390002 c.800C>T p.P267L Chr.19: 43552820-43552820 on Build GRCh38
7191627 c.757G>T p.E253* Chr.19: 43552863-43552863 on Build GRCh38
4078961 c.712-1G>T p.? Chr.19: 43552909-43552909 on Build GRCh38
6824126 c.797C>T p.S266L Chr.19: 43552823-43552823 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs576559582 Chr.19: 43552960-43552960 on Build GRCh38
rs994664781 Chr.19: 43552805-43552805 on Build GRCh38
rs754585462 Chr.19: 43553054-43553054 on Build GRCh38
rs3213361 Chr.19: 43553095-43553095 on Build GRCh38
rs746800877 Chr.19: 43552913-43552913 on Build GRCh38
rs748615713 Chr.19: 43552951-43552951 on Build GRCh38
rs771256506 Chr.19: 43552827-43552827 on Build GRCh38
rs1028375521 Chr.19: 43552928-43552928 on Build GRCh38
rs751127240 Chr.19: 43552930-43552930 on Build GRCh38
rs2307175 Chr.19: 43552967-43552967 on Build GRCh38
rs772402681 Chr.19: 43552992-43552992 on Build GRCh38
rs1799780 Chr.19: 43553124-43553124 on Build GRCh38
rs2307176 Chr.19: 43552966-43552966 on Build GRCh38
rs1030029518 Chr.19: 43552981-43552981 on Build GRCh38
rs747258459 Chr.19: 43553123-43553123 on Build GRCh38
rs148663339 Chr.19: 43553023-43553023 on Build GRCh38
rs777476390 Chr.19: 43552851-43552853 on Build GRCh38
rs761713223 Chr.19: 43553022-43553022 on Build GRCh38
rs745429409 Chr.19: 43553140-43553140 on Build GRCh38
rs371688474 Chr.19: 43553005-43553005 on Build GRCh38
rs754676643 Chr.19: 43552940-43552940 on Build GRCh38
rs767260634 Chr.19: 43553061-43553061 on Build GRCh38
rs147282758 Chr.19: 43552825-43552825 on Build GRCh38
rs915927 Chr.19: 43553075-43553075 on Build GRCh38
rs765125543 Chr.19: 43553024-43553024 on Build GRCh38
rs748549218 Chr.19: 43552800-43552800 on Build GRCh38
rs10407677 Chr.19: 43552969-43552969 on Build GRCh38
rs751089762 Chr.19: 43553050-43553050 on Build GRCh38
rs868134945 Chr.19: 43553056-43553056 on Build GRCh38
rs759826284 Chr.19: 43552873-43552873 on Build GRCh38
rs773424530 Chr.19: 43552808-43552808 on Build GRCh38
rs367988860 Chr.19: 43553141-43553141 on Build GRCh38
rs781347646 Chr.19: 43552945-43552945 on Build GRCh38
rs772158978 Chr.19: 43553112-43553112 on Build GRCh38
rs143917286 Chr.19: 43552802-43552802 on Build GRCh38
rs751654943 Chr.19: 43552976-43552976 on Build GRCh38
rs527933669 Chr.19: 43553114-43553114 on Build GRCh38
rs761650222 Chr.19: 43552907-43552907 on Build GRCh38
rs533735581 Chr.19: 43552829-43552829 on Build GRCh38
rs3213360 Chr.19: 43553101-43553101 on Build GRCh38
rs1048914812 Chr.19: 43553136-43553136 on Build GRCh38
rs1045728604 Chr.19: 43552886-43552886 on Build GRCh38
rs368767486 Chr.19: 43553076-43553076 on Build GRCh38
rs776274682 Chr.19: 43553009-43553009 on Build GRCh38
rs368965282 Chr.19: 43552904-43552904 on Build GRCh38
rs992183301 Chr.19: 43552792-43552792 on Build GRCh38
rs754338621 Chr.19: 43553081-43553081 on Build GRCh38
rs370349287 Chr.19: 43553110-43553110 on Build GRCh38
rs200575088 Chr.19: 43552858-43552858 on Build GRCh38
rs765821033 Chr.19: 43553032-43553032 on Build GRCh38
rs780361763 Chr.19: 43553118-43553118 on Build GRCh38
rs867771214 Chr.19: 43553074-43553074 on Build GRCh38
rs963263091 Chr.19: 43553043-43553043 on Build GRCh38
rs111857343 Chr.19: 43553052-43553052 on Build GRCh38
rs763738997 Chr.19: 43552878-43552878 on Build GRCh38
rs372223449 Chr.19: 43552953-43552953 on Build GRCh38
rs886380545 Chr.19: 43552850-43552850 on Build GRCh38
rs781247196 Chr.19: 43553088-43553088 on Build GRCh38
rs769855188 Chr.19: 43553120-43553120 on Build GRCh38
rs375595185 Chr.19: 43553071-43553071 on Build GRCh38
rs773008470 Chr.19: 43553131-43553131 on Build GRCh38
rs776487682 Chr.19: 43552890-43552890 on Build GRCh38
rs562926147 Chr.19: 43553026-43553026 on Build GRCh38
rs773088616 Chr.19: 43553025-43553025 on Build GRCh38
rs765213529 Chr.19: 43552914-43552914 on Build GRCh38
rs374247969 Chr.19: 43552859-43552859 on Build GRCh38
rs765901398 Chr.19: 43552924-43552924 on Build GRCh38
rs757939800 Chr.19: 43552922-43552922 on Build GRCh38
rs770969968 Chr.19: 43553137-43553137 on Build GRCh38
rs746010419 Chr.19: 43552982-43552982 on Build GRCh38
rs778474464 Chr.19: 43552962-43552962 on Build GRCh38
rs184028231 Chr.19: 43552977-43552977 on Build GRCh38
rs781639877 Chr.19: 43552797-43552797 on Build GRCh38
rs762813899 Chr.19: 43553135-43553135 on Build GRCh38
rs778314777 Chr.19: 43553080-43553080 on Build GRCh38
rs774755296 Chr.19: 43552845-43552845 on Build GRCh38
rs368837757 Chr.19: 43552996-43552996 on Build GRCh38
rs749910091 Chr.19: 43552919-43552919 on Build GRCh38

View Full Product Details