Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 15878747-15879189 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP4F2
Entrez Gene ID
8529
Gene Name
cytochrome P450 family 4 subfamily F member 2
Gene Aliases
CPF2
UniGene
Hs.558423
Gene Chromosome Location
Chr.19: 15878024-15898120 on Build GRCh38
Transcripts
NM_001082.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
992357 c.1408G>A p.G470R Chr.19: 15878926-15878926 on Build GRCh38
992355 c.1520G>A p.R507H Chr.19: 15878814-15878814 on Build GRCh38
6639386 c.1415C>T p.T472M Chr.19: 15878919-15878919 on Build GRCh38
992356 c.1506G>A p.P502P Chr.19: 15878828-15878828 on Build GRCh38
6639387 c.1488C>T p.T496T Chr.19: 15878846-15878846 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201777106 Chr.19: 15879001-15879001 on Build GRCh38
rs1020017105 Chr.19: 15879152-15879152 on Build GRCh38
rs562470633 Chr.19: 15879124-15879124 on Build GRCh38
rs149564867 Chr.19: 15878908-15878908 on Build GRCh38
rs1009949147 Chr.19: 15879005-15879005 on Build GRCh38
rs753224188 Chr.19: 15878971-15878971 on Build GRCh38
rs193114233 Chr.19: 15879055-15879055 on Build GRCh38
rs764924891 Chr.19: 15878829-15878829 on Build GRCh38
rs751855405 Chr.19: 15878845-15878845 on Build GRCh38
rs780277810 Chr.19: 15878853-15878853 on Build GRCh38
rs758747686 Chr.19: 15878976-15878976 on Build GRCh38
rs752587468 Chr.19: 15878872-15878872 on Build GRCh38
rs766239123 Chr.19: 15878783-15878783 on Build GRCh38
rs763963721 Chr.19: 15878915-15878915 on Build GRCh38
rs746343403 Chr.19: 15878846-15878846 on Build GRCh38
rs1049145392 Chr.19: 15878941-15878941 on Build GRCh38
rs766467550 Chr.19: 15878963-15878963 on Build GRCh38
rs150488576 Chr.19: 15878864-15878864 on Build GRCh38
rs1015784839 Chr.19: 15878782-15878782 on Build GRCh38
rs772809433 Chr.19: 15878859-15878859 on Build GRCh38
rs377164041 Chr.19: 15878956-15878956 on Build GRCh38
rs1029355932 Chr.19: 15879056-15879056 on Build GRCh38
rs775460036 Chr.19: 15878913-15878913 on Build GRCh38
rs749993807 Chr.19: 15878773-15878773 on Build GRCh38
rs968395758 Chr.19: 15879038-15879038 on Build GRCh38
rs754992270 Chr.19: 15878828-15878828 on Build GRCh38
rs755536102 Chr.19: 15878929-15878929 on Build GRCh38
rs575928967 Chr.19: 15879126-15879126 on Build GRCh38
rs1048402036 Chr.19: 15879108-15879108 on Build GRCh38
rs777992718 Chr.19: 15878793-15878793 on Build GRCh38
rs895531998 Chr.19: 15878999-15878999 on Build GRCh38
rs547821781 Chr.19: 15878798-15878798 on Build GRCh38
rs553804744 Chr.19: 15879032-15879032 on Build GRCh38
rs777232523 Chr.19: 15878800-15878800 on Build GRCh38
rs758269454 Chr.19: 15878839-15878839 on Build GRCh38
rs189188535 Chr.19: 15878785-15878785 on Build GRCh38
rs748295684 Chr.19: 15878788-15878788 on Build GRCh38
rs762122740 Chr.19: 15878919-15878919 on Build GRCh38
rs879251143 Chr.19: 15879081-15879081 on Build GRCh38
rs772858287 Chr.19: 15878815-15878815 on Build GRCh38
rs3952537 Chr.19: 15878886-15878886 on Build GRCh38
rs769319226 Chr.19: 15878942-15878942 on Build GRCh38
rs944093569 Chr.19: 15879090-15879090 on Build GRCh38
rs3093200 Chr.19: 15878779-15878779 on Build GRCh38
rs765151224 Chr.19: 15878870-15878870 on Build GRCh38
rs530133717 Chr.19: 15879150-15879150 on Build GRCh38
rs761571766 Chr.19: 15878769-15878769 on Build GRCh38
rs904186061 Chr.19: 15879156-15879156 on Build GRCh38
rs957212086 Chr.19: 15878885-15878885 on Build GRCh38
rs752600405 Chr.19: 15878836-15878836 on Build GRCh38
rs766991712 Chr.19: 15878770-15878770 on Build GRCh38
rs367663603 Chr.19: 15878966-15878966 on Build GRCh38
rs781119638 Chr.19: 15878890-15878890 on Build GRCh38
rs775972932 Chr.19: 15878812-15878812 on Build GRCh38
rs376954115 Chr.19: 15878947-15878947 on Build GRCh38
rs879155368 Chr.19: 15879109-15879109 on Build GRCh38
rs779245682 Chr.19: 15878933-15878933 on Build GRCh38
rs879133903 Chr.19: 15879112-15879112 on Build GRCh38
rs911734112 Chr.19: 15879085-15879085 on Build GRCh38
rs749823670 Chr.19: 15878895-15878895 on Build GRCh38
rs571754062 Chr.19: 15878784-15878784 on Build GRCh38
rs746422239 Chr.19: 15878803-15878803 on Build GRCh38
rs998409824 Chr.19: 15878809-15878809 on Build GRCh38
rs146148233 Chr.19: 15878926-15878926 on Build GRCh38
rs765673890 Chr.19: 15878970-15878970 on Build GRCh38
rs143677430 Chr.19: 15878865-15878865 on Build GRCh38
rs138905922 Chr.19: 15878940-15878940 on Build GRCh38
rs138208261 Chr.19: 15878893-15878893 on Build GRCh38
rs747016395 Chr.19: 15878797-15878797 on Build GRCh38
rs767226438 Chr.19: 15878814-15878814 on Build GRCh38
rs906084878 Chr.19: 15879111-15879111 on Build GRCh38
rs557462046 Chr.19: 15878950-15878950 on Build GRCh38
rs774436154 Chr.19: 15878948-15878948 on Build GRCh38
rs879103134 Chr.19: 15879094-15879094 on Build GRCh38
rs765903428 Chr.19: 15878819-15878819 on Build GRCh38
rs912903113 Chr.19: 15878781-15878781 on Build GRCh38
rs776263021 Chr.19: 15878867-15878867 on Build GRCh38
rs749602056 Chr.19: 15878855-15878855 on Build GRCh38
rs931834375 Chr.19: 15878843-15878843 on Build GRCh38
rs866736048 Chr.19: 15878790-15878790 on Build GRCh38
rs774915992 Chr.19: 15878946-15878946 on Build GRCh38
rs764014953 Chr.19: 15878880-15878880 on Build GRCh38
rs762784709 Chr.19: 15878878-15878878 on Build GRCh38
rs763027713 Chr.19: 15878914-15878914 on Build GRCh38
rs758700593 Chr.19: 15878792-15878792 on Build GRCh38
rs3093199 Chr.19: 15879061-15879061 on Build GRCh38
rs769024259 Chr.19: 15878856-15878856 on Build GRCh38
rs751462720 Chr.19: 15878918-15878918 on Build GRCh38
rs544862114 Chr.19: 15879135-15879135 on Build GRCh38
rs878881912 Chr.19: 15879099-15879099 on Build GRCh38
rs377513974 Chr.19: 15878939-15878939 on Build GRCh38
rs750624996 Chr.19: 15878922-15878922 on Build GRCh38
rs896022110 Chr.19: 15879138-15879138 on Build GRCh38
rs537884263 Chr.19: 15878927-15878927 on Build GRCh38
rs778041819 Chr.19: 15878986-15878986 on Build GRCh38
rs1033087360 Chr.19: 15879133-15879133 on Build GRCh38
rs376410257 Chr.19: 15879047-15879047 on Build GRCh38
rs770028371 Chr.19: 15878810-15878810 on Build GRCh38
rs897330784 Chr.19: 15879063-15879063 on Build GRCh38
rs4020346 Chr.19: 15878920-15878920 on Build GRCh38
rs368748953 Chr.19: 15878891-15878891 on Build GRCh38
rs869275818 Chr.19: 15878948-15878948 on Build GRCh38
rs770633305 Chr.19: 15878907-15878907 on Build GRCh38
rs143088581 Chr.19: 15878898-15878898 on Build GRCh38
rs759025188 Chr.19: 15878912-15878912 on Build GRCh38
rs577527241 Chr.19: 15878952-15878952 on Build GRCh38
rs77076703 Chr.19: 15879117-15879117 on Build GRCh38
rs773450268 Chr.19: 15878954-15878954 on Build GRCh38
rs565137459 Chr.19: 15879158-15879158 on Build GRCh38
rs539957683 Chr.19: 15879016-15879016 on Build GRCh38
rs567783827 Chr.19: 15878813-15878813 on Build GRCh38
rs753686777 Chr.19: 15878820-15878820 on Build GRCh38
rs777396221 Chr.19: 15878779-15878779 on Build GRCh38
rs931329896 Chr.19: 15879107-15879107 on Build GRCh38

View Full Product Details