Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 23969663-23970105 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHRS4L2
Entrez Gene ID
317749
Gene Name
dehydrogenase/reductase 4 like 2
Gene Aliases
SDR25C3
UniGene
Hs.743442
Gene Chromosome Location
Chr.14: 23969874-24006408 on Build GRCh38
Transcripts
NM_001193635.1 NM_001193636.1 NM_001193637.1

SNP Detail

SNP ID Chromosome Location
rs112641258 Chr.14: 23969916-23969916 on Build GRCh38
rs945938605 Chr.14: 23969897-23969897 on Build GRCh38
rs995209192 Chr.14: 23969983-23969983 on Build GRCh38
rs558787129 Chr.14: 23969816-23969816 on Build GRCh38
rs976169184 Chr.14: 23969837-23969837 on Build GRCh38
rs754430569 Chr.14: 23969859-23969859 on Build GRCh38
rs894207275 Chr.14: 23969971-23969971 on Build GRCh38
rs771568186 Chr.14: 23969933-23969933 on Build GRCh38
rs893075778 Chr.14: 23969876-23969876 on Build GRCh38
rs61999781 Chr.14: 23969942-23969942 on Build GRCh38
rs560012793 Chr.14: 23969949-23969949 on Build GRCh38
rs751836546 Chr.14: 23970016-23970016 on Build GRCh38
rs755233844 Chr.14: 23970055-23970055 on Build GRCh38
rs141737044 Chr.14: 23970000-23970000 on Build GRCh38
rs778925494 Chr.14: 23970081-23970081 on Build GRCh38
rs1034077201 Chr.14: 23969948-23969948 on Build GRCh38
rs199636983 Chr.14: 23969790-23969790 on Build GRCh38
rs368977129 Chr.14: 23969684-23969684 on Build GRCh38
rs1002567214 Chr.14: 23969701-23969701 on Build GRCh38
rs978709024 Chr.14: 23969780-23969781 on Build GRCh38
rs1034014895 Chr.14: 23969750-23969750 on Build GRCh38
rs781225704 Chr.14: 23970065-23970065 on Build GRCh38
rs575524954 Chr.14: 23969852-23969852 on Build GRCh38
rs531631702 Chr.14: 23970027-23970027 on Build GRCh38
rs538195685 Chr.14: 23969873-23969873 on Build GRCh38
rs886913407 Chr.14: 23969927-23969927 on Build GRCh38
rs908034755 Chr.14: 23969826-23969826 on Build GRCh38
rs199930173 Chr.14: 23969704-23969704 on Build GRCh38
rs562712170 Chr.14: 23970008-23970008 on Build GRCh38
rs763671080 Chr.14: 23969972-23969972 on Build GRCh38
rs774013600 Chr.14: 23969930-23969930 on Build GRCh38
rs577065247 Chr.14: 23969953-23969953 on Build GRCh38
rs747001835 Chr.14: 23969904-23969904 on Build GRCh38
rs61999782 Chr.14: 23969991-23969991 on Build GRCh38
rs920875455 Chr.14: 23969788-23969788 on Build GRCh38
rs116622736 Chr.14: 23969799-23969799 on Build GRCh38
rs971146598 Chr.14: 23969696-23969696 on Build GRCh38
rs1027056185 Chr.14: 23970056-23970056 on Build GRCh38
rs770788787 Chr.14: 23970072-23970072 on Build GRCh38
rs758800812 Chr.14: 23970001-23970001 on Build GRCh38
rs952379914 Chr.14: 23969796-23969796 on Build GRCh38
rs753451295 Chr.14: 23969973-23969973 on Build GRCh38
rs779395918 Chr.14: 23969894-23969894 on Build GRCh38
rs1041583481 Chr.14: 23969898-23969898 on Build GRCh38
rs369340165 Chr.14: 23969856-23969856 on Build GRCh38
rs764650597 Chr.14: 23969858-23969858 on Build GRCh38
rs554529796 Chr.14: 23969901-23969901 on Build GRCh38
rs974119889 Chr.14: 23969785-23969785 on Build GRCh38
rs983669151 Chr.14: 23969812-23969812 on Build GRCh38
rs1002621028 Chr.14: 23969939-23969939 on Build GRCh38
rs748295068 Chr.14: 23970068-23970068 on Build GRCh38
rs768744263 Chr.14: 23969910-23969910 on Build GRCh38
rs765906913 Chr.14: 23969997-23969997 on Build GRCh38
rs540289470 Chr.14: 23969932-23969932 on Build GRCh38
rs780325141 Chr.14: 23969902-23969902 on Build GRCh38
rs932013982 Chr.14: 23969871-23969871 on Build GRCh38
rs373068733 Chr.14: 23969923-23969923 on Build GRCh38
rs750854133 Chr.14: 23969896-23969896 on Build GRCh38

View Full Product Details