Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 61497118-61497448 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADGRL3
Entrez Gene ID
23284
Gene Name
adhesion G protein-coupled receptor L3
Gene Aliases
CIRL3, CL3, LEC3, LPHN3
UniGene
Hs.28391
Gene Chromosome Location
Chr.4: 61201256-62078335 on Build GRCh38
Transcripts
NM_001322246.1 NM_001322402.1 NM_015236.5 XM_011531788.2 XM_011531791.2 XM_017007929.1 XM_017007930.1 XM_017007931.1 XM_017007932.1 XM_017007933.1 XM_017007934.1 XM_017007935.1 XM_017007936.1 XM_017007937.1 XM_017007938.1 XM_017007939.1 XM_017007940.1 XM_017007941.1 XM_017007942.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5866572 c.30G>A p.M10I Chr.4: 61497323-61497323 on Build GRCh38
5866576 c.52C>T p.H18Y Chr.4: 61497345-61497345 on Build GRCh38
3604770 c.27C>T p.F9F Chr.4: 61497320-61497320 on Build GRCh38
3604773 c.43C>T p.P15S Chr.4: 61497336-61497336 on Build GRCh38
3604776 c.55G>A p.A19T Chr.4: 61497348-61497348 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748140786 Chr.4: 61497354-61497355 on Build GRCh38
rs746018239 Chr.4: 61497388-61497388 on Build GRCh38
rs961203156 Chr.4: 61497164-61497169 on Build GRCh38
rs750077514 Chr.4: 61497255-61497255 on Build GRCh38
rs753690153 Chr.4: 61497338-61497338 on Build GRCh38
rs547657076 Chr.4: 61497312-61497312 on Build GRCh38
rs781140447 Chr.4: 61497375-61497375 on Build GRCh38
rs754885643 Chr.4: 61497346-61497346 on Build GRCh38
rs542779581 Chr.4: 61497176-61497176 on Build GRCh38
rs765259443 Chr.4: 61497358-61497358 on Build GRCh38
rs569728663 Chr.4: 61497406-61497406 on Build GRCh38
rs766960254 Chr.4: 61497327-61497327 on Build GRCh38
rs772839558 Chr.4: 61497390-61497391 on Build GRCh38
rs576609204 Chr.4: 61497276-61497276 on Build GRCh38
rs777701538 Chr.4: 61497281-61497281 on Build GRCh38
rs1055028149 Chr.4: 61497401-61497401 on Build GRCh38
rs146841463 Chr.4: 61497396-61497396 on Build GRCh38
rs752606162 Chr.4: 61497369-61497369 on Build GRCh38
rs565098770 Chr.4: 61497306-61497306 on Build GRCh38
rs374166291 Chr.4: 61497417-61497417 on Build GRCh38
rs925151833 Chr.4: 61497193-61497193 on Build GRCh38
rs1015777507 Chr.4: 61497162-61497162 on Build GRCh38
rs766158952 Chr.4: 61497334-61497334 on Build GRCh38
rs1026659709 Chr.4: 61497380-61497380 on Build GRCh38
rs371504666 Chr.4: 61497305-61497305 on Build GRCh38
rs767242068 Chr.4: 61497252-61497252 on Build GRCh38
rs781434551 Chr.4: 61497298-61497298 on Build GRCh38
rs756468990 Chr.4: 61497393-61497393 on Build GRCh38
rs769600185 Chr.4: 61497382-61497384 on Build GRCh38
rs760326778 Chr.4: 61497329-61497329 on Build GRCh38
rs753708754 Chr.4: 61497275-61497275 on Build GRCh38
rs369021307 Chr.4: 61497399-61497399 on Build GRCh38
rs529678104 Chr.4: 61497322-61497322 on Build GRCh38
rs559113755 Chr.4: 61497314-61497314 on Build GRCh38
rs746112623 Chr.4: 61497303-61497303 on Build GRCh38
rs934793430 Chr.4: 61497400-61497400 on Build GRCh38
rs779840510 Chr.4: 61497274-61497274 on Build GRCh38
rs757388371 Chr.4: 61497289-61497289 on Build GRCh38
rs995542166 Chr.4: 61497352-61497352 on Build GRCh38
rs757309845 Chr.4: 61497371-61497371 on Build GRCh38
rs773707258 Chr.4: 61497316-61497316 on Build GRCh38
rs747162650 Chr.4: 61497288-61497288 on Build GRCh38
rs781504255 Chr.4: 61497290-61497290 on Build GRCh38
rs530572768 Chr.4: 61497409-61497409 on Build GRCh38
rs561416087 Chr.4: 61497230-61497230 on Build GRCh38
rs781296932 Chr.4: 61497284-61497284 on Build GRCh38
rs184704156 Chr.4: 61497311-61497311 on Build GRCh38
rs953925993 Chr.4: 61497264-61497264 on Build GRCh38

View Full Product Details