Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 119092021-119092579 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HMBS
Entrez Gene ID
3145
Gene Name
hydroxymethylbilane synthase
Gene Aliases
PBG-D, PBGD, PORC, UPS
UniGene
Hs.82609
Gene Chromosome Location
Chr.11: 119084871-119093549 on Build GRCh38
Transcripts
NM_000190.3 NM_001024382.1 NM_001258208.1 NM_001258209.1 XM_005271531.1 XM_005271532.1 XM_005271533.3 XM_011542796.1 XM_017017629.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2149409 c.614T>G p.I205S Chr.11: 119092126-119092126 on Build GRCh38
6014696 c.677C>T p.A226V Chr.11: 119092429-119092429 on Build GRCh38
923745 c.724G>A p.E242K Chr.11: 119092476-119092476 on Build GRCh38
2106371 c.746C>G p.A249G Chr.11: 119092498-119092498 on Build GRCh38
923744 c.637T>C p.Y213H Chr.11: 119092149-119092149 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs998517128 Chr.11: 119092314-119092314 on Build GRCh38
rs536859676 Chr.11: 119092209-119092209 on Build GRCh38
rs775679798 Chr.11: 119092157-119092157 on Build GRCh38
rs753779066 Chr.11: 119092090-119092090 on Build GRCh38
rs118204111 Chr.11: 119092491-119092491 on Build GRCh38
rs118204114 Chr.11: 119092507-119092507 on Build GRCh38
rs202030678 Chr.11: 119092087-119092087 on Build GRCh38
rs367691838 Chr.11: 119092202-119092202 on Build GRCh38
rs748119639 Chr.11: 119092539-119092539 on Build GRCh38
rs551123195 Chr.11: 119092351-119092351 on Build GRCh38
rs747700022 Chr.11: 119092113-119092113 on Build GRCh38
rs1007859875 Chr.11: 119092137-119092137 on Build GRCh38
rs766381418 Chr.11: 119092390-119092390 on Build GRCh38
rs749610272 Chr.11: 119092116-119092116 on Build GRCh38
rs191708247 Chr.11: 119092093-119092093 on Build GRCh38
rs760527104 Chr.11: 119092548-119092548 on Build GRCh38
rs959501193 Chr.11: 119092059-119092059 on Build GRCh38
rs377382385 Chr.11: 119092088-119092088 on Build GRCh38
rs544561277 Chr.11: 119092350-119092350 on Build GRCh38
rs770285966 Chr.11: 119092114-119092114 on Build GRCh38
rs779121403 Chr.11: 119092231-119092231 on Build GRCh38
rs773159376 Chr.11: 119092354-119092354 on Build GRCh38
rs1784304 Chr.11: 119092106-119092106 on Build GRCh38
rs779696620 Chr.11: 119092100-119092100 on Build GRCh38
rs756844866 Chr.11: 119092505-119092505 on Build GRCh38
rs959814243 Chr.11: 119092190-119092190 on Build GRCh38
rs769925467 Chr.11: 119092543-119092543 on Build GRCh38
rs112806167 Chr.11: 119092210-119092210 on Build GRCh38
rs767293635 Chr.11: 119092402-119092402 on Build GRCh38
rs745830730 Chr.11: 119092545-119092545 on Build GRCh38
rs530739793 Chr.11: 119092071-119092071 on Build GRCh38
rs189033807 Chr.11: 119092470-119092470 on Build GRCh38
rs945463472 Chr.11: 119092306-119092306 on Build GRCh38
rs1016672433 Chr.11: 119092247-119092247 on Build GRCh38
rs986068980 Chr.11: 119092049-119092050 on Build GRCh38
rs118204099 Chr.11: 119092486-119092486 on Build GRCh38
rs546938328 Chr.11: 119092536-119092536 on Build GRCh38
rs980999691 Chr.11: 119092352-119092352 on Build GRCh38
rs761810461 Chr.11: 119092476-119092476 on Build GRCh38
rs1014399089 Chr.11: 119092342-119092342 on Build GRCh38
rs758794172 Chr.11: 119092520-119092520 on Build GRCh38
rs758182016 Chr.11: 119092099-119092099 on Build GRCh38
rs771421211 Chr.11: 119092295-119092295 on Build GRCh38
rs768860225 Chr.11: 119092353-119092353 on Build GRCh38
rs768177122 Chr.11: 119092554-119092554 on Build GRCh38
rs762779875 Chr.11: 119092359-119092359 on Build GRCh38
rs755584861 Chr.11: 119092413-119092413 on Build GRCh38
rs765249502 Chr.11: 119092180-119092180 on Build GRCh38
rs775912581 Chr.11: 119092237-119092237 on Build GRCh38
rs1055330576 Chr.11: 119092391-119092391 on Build GRCh38
rs201909197 Chr.11: 119092489-119092489 on Build GRCh38
rs750969181 Chr.11: 119092514-119092514 on Build GRCh38
rs777084600 Chr.11: 119092075-119092075 on Build GRCh38
rs772049791 Chr.11: 119092547-119092547 on Build GRCh38
rs752355312 Chr.11: 119092408-119092408 on Build GRCh38
rs575608844 Chr.11: 119092133-119092133 on Build GRCh38
rs763844879 Chr.11: 119092497-119092497 on Build GRCh38
rs569520602 Chr.11: 119092455-119092455 on Build GRCh38
rs753280372 Chr.11: 119092502-119092502 on Build GRCh38
rs911130376 Chr.11: 119092291-119092291 on Build GRCh38
rs754597374 Chr.11: 119092204-119092204 on Build GRCh38
rs202067277 Chr.11: 119092475-119092475 on Build GRCh38
rs934529084 Chr.11: 119092493-119092493 on Build GRCh38
rs548857546 Chr.11: 119092079-119092079 on Build GRCh38
rs559595137 Chr.11: 119092300-119092300 on Build GRCh38
rs763928248 Chr.11: 119092428-119092428 on Build GRCh38
rs759384395 Chr.11: 119092401-119092401 on Build GRCh38
rs573522574 Chr.11: 119092282-119092282 on Build GRCh38
rs961414429 Chr.11: 119092254-119092254 on Build GRCh38
rs1018244852 Chr.11: 119092478-119092478 on Build GRCh38
rs768973838 Chr.11: 119092467-119092467 on Build GRCh38
rs780661095 Chr.11: 119092112-119092112 on Build GRCh38
rs757670960 Chr.11: 119092429-119092429 on Build GRCh38
rs140994871 Chr.11: 119092415-119092415 on Build GRCh38
rs557506252 Chr.11: 119092129-119092129 on Build GRCh38
rs985805000 Chr.11: 119092290-119092290 on Build GRCh38
rs771276134 Chr.11: 119092228-119092228 on Build GRCh38
rs993891865 Chr.11: 119092258-119092258 on Build GRCh38
rs771206317 Chr.11: 119092124-119092124 on Build GRCh38
rs759698474 Chr.11: 119092144-119092144 on Build GRCh38
rs755636016 Chr.11: 119092217-119092217 on Build GRCh38
rs770760881 Chr.11: 119092472-119092472 on Build GRCh38
rs751422884 Chr.11: 119092200-119092200 on Build GRCh38
rs373721177 Chr.11: 119092084-119092084 on Build GRCh38
rs895340609 Chr.11: 119092219-119092219 on Build GRCh38
rs118204115 Chr.11: 119092518-119092518 on Build GRCh38
rs114025170 Chr.11: 119092469-119092469 on Build GRCh38
rs1022738604 Chr.11: 119092351-119092351 on Build GRCh38
rs368550811 Chr.11: 119092095-119092095 on Build GRCh38
rs746221527 Chr.11: 119092440-119092440 on Build GRCh38
rs771172103 Chr.11: 119092085-119092085 on Build GRCh38
rs370994091 Chr.11: 119092053-119092053 on Build GRCh38
rs540699633 Chr.11: 119092261-119092261 on Build GRCh38
rs375654742 Chr.11: 119092232-119092232 on Build GRCh38
rs760296083 Chr.11: 119092496-119092496 on Build GRCh38
rs769645562 Chr.11: 119092544-119092544 on Build GRCh38
rs1031415682 Chr.11: 119092179-119092179 on Build GRCh38
rs968385712 Chr.11: 119092056-119092056 on Build GRCh38
rs118204116 Chr.11: 119092159-119092159 on Build GRCh38
rs760900015 Chr.11: 119092176-119092176 on Build GRCh38
rs140572589 Chr.11: 119092488-119092488 on Build GRCh38
rs533119126 Chr.11: 119092325-119092328 on Build GRCh38
rs118204113 Chr.11: 119092506-119092506 on Build GRCh38
rs772414541 Chr.11: 119092235-119092235 on Build GRCh38
rs374052034 Chr.11: 119092268-119092268 on Build GRCh38
rs774013647 Chr.11: 119092399-119092399 on Build GRCh38
rs774594843 Chr.11: 119092127-119092127 on Build GRCh38
rs758847114 Chr.11: 119092442-119092442 on Build GRCh38
rs927970404 Chr.11: 119092315-119092315 on Build GRCh38
rs749793029 Chr.11: 119092220-119092220 on Build GRCh38
rs118204112 Chr.11: 119092500-119092500 on Build GRCh38
rs747263985 Chr.11: 119092466-119092466 on Build GRCh38
rs750338933 Chr.11: 119092186-119092186 on Build GRCh38
rs376554428 Chr.11: 119092461-119092461 on Build GRCh38
rs1005275043 Chr.11: 119092063-119092063 on Build GRCh38
rs963558406 Chr.11: 119092423-119092423 on Build GRCh38
rs747419493 Chr.11: 119092527-119092527 on Build GRCh38
rs28990987 Chr.11: 119092094-119092094 on Build GRCh38
rs151289916 Chr.11: 119092430-119092430 on Build GRCh38
rs118204110 Chr.11: 119092419-119092419 on Build GRCh38
rs968354607 Chr.11: 119092160-119092160 on Build GRCh38
rs142459647 Chr.11: 119092426-119092426 on Build GRCh38
rs747988121 Chr.11: 119092312-119092312 on Build GRCh38
rs540501492 Chr.11: 119092283-119092283 on Build GRCh38
rs911076319 Chr.11: 119092091-119092091 on Build GRCh38
rs998551270 Chr.11: 119092169-119092169 on Build GRCh38
rs752190083 Chr.11: 119092211-119092211 on Build GRCh38
rs1020145031 Chr.11: 119092517-119092517 on Build GRCh38
rs770338960 Chr.11: 119092083-119092083 on Build GRCh38
rs371971227 Chr.11: 119092530-119092530 on Build GRCh38
rs778311186 Chr.11: 119092115-119092115 on Build GRCh38
rs765208153 Chr.11: 119092494-119092494 on Build GRCh38
rs184212174 Chr.11: 119092299-119092299 on Build GRCh38
rs778277582 Chr.11: 119092218-119092218 on Build GRCh38
rs374531252 Chr.11: 119092191-119092191 on Build GRCh38
rs767103817 Chr.11: 119092407-119092407 on Build GRCh38
rs766112542 Chr.11: 119092198-119092198 on Build GRCh38
rs1042448997 Chr.11: 119092311-119092311 on Build GRCh38
rs1045334242 Chr.11: 119092105-119092105 on Build GRCh38
rs370537877 Chr.11: 119092255-119092255 on Build GRCh38
rs369144907 Chr.11: 119092516-119092516 on Build GRCh38
rs747026932 Chr.11: 119092240-119092240 on Build GRCh38
rs772471000 Chr.11: 119092146-119092146 on Build GRCh38

View Full Product Details