Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 204538128-204538299 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MDM4
Entrez Gene ID
4194
Gene Name
MDM4, p53 regulator
Gene Aliases
HDMX, MDMX, MRP1
UniGene
Hs.497492
Gene Chromosome Location
Chr.1: 204516379-204558120 on Build GRCh38
Transcripts
NM_001204171.1 NM_001204172.1 NM_001278516.1 NM_001278517.1 NM_001278518.1 NM_001278519.1 NM_002393.4 XM_017001311.1 XM_017001312.1 XM_017001313.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6933265 c.79-9A>T p.? Chr.1: 204538212-204538212 on Build GRCh38
7339474 c.484G>C p.E162Q Chr.1: 204538281-204538281 on Build GRCh38
3943353 c.127C>G p.L43V Chr.1: 204538269-204538269 on Build GRCh38
6933264 c.415A>T p.S139C Chr.1: 204538212-204538212 on Build GRCh38
6710377 c.79-8G>A p.? Chr.1: 204538213-204538213 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759826464 Chr.1: 204538253-204538253 on Build GRCh38
rs748726868 Chr.1: 204538202-204538203 on Build GRCh38
rs776550793 Chr.1: 204538248-204538248 on Build GRCh38
rs752213278 Chr.1: 204538238-204538238 on Build GRCh38
rs535411740 Chr.1: 204538193-204538193 on Build GRCh38
rs536731861 Chr.1: 204538192-204538192 on Build GRCh38
rs541354806 Chr.1: 204538151-204538151 on Build GRCh38
rs758910592 Chr.1: 204538176-204538176 on Build GRCh38
rs116197192 Chr.1: 204538255-204538255 on Build GRCh38
rs778877475 Chr.1: 204538239-204538239 on Build GRCh38
rs779925757 Chr.1: 204538212-204538212 on Build GRCh38
rs878913118 Chr.1: 204538229-204538229 on Build GRCh38
rs771038153 Chr.1: 204538242-204538242 on Build GRCh38
rs755704621 Chr.1: 204538210-204538210 on Build GRCh38
rs759645397 Chr.1: 204538165-204538165 on Build GRCh38
rs763354177 Chr.1: 204538258-204538258 on Build GRCh38
rs3761687 Chr.1: 204538163-204538163 on Build GRCh38
rs555716350 Chr.1: 204538266-204538266 on Build GRCh38
rs780847647 Chr.1: 204538195-204538195 on Build GRCh38
rs748966155 Chr.1: 204538230-204538230 on Build GRCh38
rs763547550 Chr.1: 204538183-204538183 on Build GRCh38
rs756009753 Chr.1: 204538165-204538166 on Build GRCh38
rs751129797 Chr.1: 204538192-204538192 on Build GRCh38
rs986915016 Chr.1: 204538177-204538177 on Build GRCh38
rs775928214 Chr.1: 204538256-204538256 on Build GRCh38
rs377079920 Chr.1: 204538190-204538190 on Build GRCh38
rs752955441 Chr.1: 204538180-204538180 on Build GRCh38
rs748072156 Chr.1: 204538240-204538240 on Build GRCh38
rs573980569 Chr.1: 204538186-204538186 on Build GRCh38
rs750134262 Chr.1: 204538196-204538196 on Build GRCh38

View Full Product Details