Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 3728125-3728259 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CREBBP
Entrez Gene ID
1387
Gene Name
CREB binding protein
Gene Aliases
CBP, KAT3A, RSTS
UniGene
Hs.459759
Gene Chromosome Location
Chr.16: 3725054-3880727 on Build GRCh38
Transcripts
NM_001079846.1 NM_004380.2 XM_005255124.4 XM_005255125.4 XM_006720848.3 XM_011522381.2 XM_017022944.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6942789 c.6875A>G p.Q2292R Chr.16: 3728172-3728172 on Build GRCh38
6492954 c.6825_6826insG p.Q2276fs*65 Chr.16: 3728221-3728222 on Build GRCh38
6964217 c.6844G>C p.D2282H Chr.16: 3728203-3728203 on Build GRCh38
1128962 c.6795G>A p.A2265A Chr.16: 3728252-3728252 on Build GRCh38
6635346 c.6846C>T p.D2282D Chr.16: 3728201-3728201 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781438241 Chr.16: 3728208-3728208 on Build GRCh38
rs372866305 Chr.16: 3728213-3728213 on Build GRCh38
rs749660119 Chr.16: 3728217-3728217 on Build GRCh38
rs761951767 Chr.16: 3728235-3728235 on Build GRCh38
rs201161108 Chr.16: 3728195-3728195 on Build GRCh38
rs753192214 Chr.16: 3728209-3728209 on Build GRCh38
rs750251074 Chr.16: 3728162-3728162 on Build GRCh38
rs763223282 Chr.16: 3728167-3728167 on Build GRCh38
rs956984557 Chr.16: 3728159-3728159 on Build GRCh38
rs148904096 Chr.16: 3728198-3728198 on Build GRCh38
rs949743163 Chr.16: 3728202-3728202 on Build GRCh38
rs779528907 Chr.16: 3728234-3728234 on Build GRCh38
rs368174171 Chr.16: 3728194-3728194 on Build GRCh38
rs759722245 Chr.16: 3728223-3728240 on Build GRCh38
rs866381658 Chr.16: 3728206-3728206 on Build GRCh38
rs771721926 Chr.16: 3728218-3728218 on Build GRCh38
rs778189658 Chr.16: 3728216-3728216 on Build GRCh38

View Full Product Details