Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 3727677-3727840 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CREBBP
Entrez Gene ID
1387
Gene Name
CREB binding protein
Gene Aliases
CBP, KAT3A, RSTS
UniGene
Hs.459759
Gene Chromosome Location
Chr.16: 3725054-3880727 on Build GRCh38
Transcripts
NM_001079846.1 NM_004380.2 XM_005255124.4 XM_005255125.4 XM_006720848.3 XM_011522381.2 XM_017022944.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5019178 c.7212A>G p.E2404E Chr.16: 3727835-3727835 on Build GRCh38
3509547 c.7281C>T p.V2427V Chr.16: 3727766-3727766 on Build GRCh38
4060689 c.7285G>A p.D2429N Chr.16: 3727762-3727762 on Build GRCh38
4770345 c.7329G>T p.*2443Y Chr.16: 3727718-3727718 on Build GRCh38
2919888 c.7262C>T p.S2421F Chr.16: 3727785-3727785 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767532168 Chr.16: 3727781-3727781 on Build GRCh38
rs752612004 Chr.16: 3727782-3727782 on Build GRCh38
rs895608889 Chr.16: 3727736-3727736 on Build GRCh38
rs958995662 Chr.16: 3727813-3727813 on Build GRCh38
rs966123342 Chr.16: 3727820-3727820 on Build GRCh38
rs1004921860 Chr.16: 3727801-3727801 on Build GRCh38
rs1027596396 Chr.16: 3727790-3727790 on Build GRCh38
rs749538388 Chr.16: 3727742-3727742 on Build GRCh38
rs751483136 Chr.16: 3727702-3727702 on Build GRCh38
rs772690401 Chr.16: 3727755-3727755 on Build GRCh38
rs1007567958 Chr.16: 3727711-3727711 on Build GRCh38
rs144609433 Chr.16: 3727745-3727745 on Build GRCh38
rs587783514 Chr.16: 3727754-3727754 on Build GRCh38
rs145623010 Chr.16: 3727729-3727729 on Build GRCh38
rs749448607 Chr.16: 3727804-3727804 on Build GRCh38
rs761138453 Chr.16: 3727765-3727765 on Build GRCh38
rs990512518 Chr.16: 3727817-3727817 on Build GRCh38
rs368297611 Chr.16: 3727792-3727792 on Build GRCh38
rs750047644 Chr.16: 3727770-3727770 on Build GRCh38
rs748024973 Chr.16: 3727721-3727721 on Build GRCh38
rs764396354 Chr.16: 3727766-3727766 on Build GRCh38
rs147915719 Chr.16: 3727809-3727809 on Build GRCh38
rs757539209 Chr.16: 3727808-3727808 on Build GRCh38
rs200566758 Chr.16: 3727791-3727791 on Build GRCh38
rs545097572 Chr.16: 3727773-3727773 on Build GRCh38
rs745992971 Chr.16: 3727811-3727811 on Build GRCh38
rs367899910 Chr.16: 3727746-3727746 on Build GRCh38
rs775691341 Chr.16: 3727756-3727756 on Build GRCh38
rs372173306 Chr.16: 3727772-3727772 on Build GRCh38
rs763197100 Chr.16: 3727701-3727702 on Build GRCh38
rs374635162 Chr.16: 3727724-3727724 on Build GRCh38
rs368649361 Chr.16: 3727730-3727730 on Build GRCh38
rs780837684 Chr.16: 3727708-3727708 on Build GRCh38
rs754944719 Chr.16: 3727780-3727780 on Build GRCh38
rs370168199 Chr.16: 3727739-3727739 on Build GRCh38
rs375448134 Chr.16: 3727775-3727775 on Build GRCh38
rs755034771 Chr.16: 3727706-3727706 on Build GRCh38

View Full Product Details