Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 55485260-55485418 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MMP2
Entrez Gene ID
4313
Gene Name
matrix metallopeptidase 2
Gene Aliases
CLG4, CLG4A, MMP-2, MMP-II, MONA, TBE-1
UniGene
Hs.513617
Gene Chromosome Location
Chr.16: 55478830-55506691 on Build GRCh38
Transcripts
NM_001127891.2 NM_001302508.1 NM_001302509.1 NM_001302510.1 NM_004530.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6079382 c.556G>C p.G186R Chr.16: 55485325-55485325 on Build GRCh38
4702697 c.555C>T p.D185D Chr.16: 55485324-55485324 on Build GRCh38
6079381 c.589C>A p.P197T Chr.16: 55485358-55485358 on Build GRCh38
191658 c.627C>T p.D209D Chr.16: 55485396-55485396 on Build GRCh38
5732029 c.568C>T p.L190F Chr.16: 55485337-55485337 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761562624 Chr.16: 55485284-55485284 on Build GRCh38
rs886052125 Chr.16: 55485357-55485357 on Build GRCh38
rs767160206 Chr.16: 55485288-55485288 on Build GRCh38
rs778038179 Chr.16: 55485396-55485396 on Build GRCh38
rs786205497 Chr.16: 55485307-55485307 on Build GRCh38
rs763687510 Chr.16: 55485282-55485282 on Build GRCh38
rs1057518712 Chr.16: 55485308-55485308 on Build GRCh38
rs111590299 Chr.16: 55485316-55485316 on Build GRCh38
rs140984687 Chr.16: 55485360-55485360 on Build GRCh38
rs766292429 Chr.16: 55485322-55485322 on Build GRCh38
rs773941696 Chr.16: 55485283-55485283 on Build GRCh38
rs753761872 Chr.16: 55485324-55485324 on Build GRCh38
rs754848720 Chr.16: 55485333-55485333 on Build GRCh38
rs760486439 Chr.16: 55485318-55485318 on Build GRCh38
rs778965210 Chr.16: 55485340-55485340 on Build GRCh38
rs151071926 Chr.16: 55485306-55485306 on Build GRCh38
rs199814532 Chr.16: 55485354-55485354 on Build GRCh38

View Full Product Details